Chemistry > Chemical Synthesis > Technology Spotlights >  Symphos Ligands
Chemical Synthesis
Symphos Ligands
A new class of phenyl-backbone derived P,O-ligands (Symphos) were developed by Symyx Technologies in collaboration with professor Fu and co-workers.1 These new ligands proved to be suitable for general palladium/ligand catalyzed Suzuki transformations of aryl halides. This new system demonstrated its efficiency by catalyzing the cross-coupling of aryl boronic acids and aryl chlorides containing electron poor and electron rich substituents with good overall yields. The catalysis was carried out with Pd(dba)2 and Symphos with CsF in toluene at 105 °C with yields up to 97%.
Symphos Ligand for Efficient Suzuki Cross Coupling
Entry
R-B(OH)2
Ar-Cl
Yield (%)
1
Symphos Tech Spotlight 03
Symphos Tech Spotlight 04
96
2
Symphos Tech Spotlight 05
Symphos Tech Spotlight 06
97
3
Symphos Tech Spotlight 07
Symphos Tech Spotlight 08
91
4
Symphos Tech Spotlight 09
Symphos Tech Spotlight 10
94
Reference:
  1. Bei, X. et al. J. Org. Chem. 1999, 64, 6797.
Product No.
Product Name
Structure
Add to Cart
SYMPHOS
Symphos Tech Spotlight 11
2-(2-Dicyclohexylphosphinophenyl)-1,3-dioxolane
Symphos Tech Spotlight 12