Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Chemical Synthesis > Technology Spotlights > Potassium Organotrifluoroborates
Technology Spotlights
Potassium Organotrifluoroborates
Organotrifluoroborates: Versatile and Stable Boronic Acid Surrogates
Prof. Gary Molander has extensively developed the chemistry of organotrifluoroborates. These bench stable boronic acid surrogates are useful for Suzuki-Miyaura cross-coupling reactions and have also been used for a variety of other C-C bond forming reactions. Importantly, these reagents are compatible with a wide range of functional groups and are stable to many commonly used and harsh reaction conditions.
Benefits of Organtrifluoroborates:
  • Bench stable reagents that can be stored indefinitely
  • Can be effectively coupled with a wide range of organic electrophiles
  • Exist as tetracoordinated monomeric species
  • Are less prone to protodeboronation than their boronic acid or boronate ester counterparts
Suzuki-Miyaura Cross-Coupling with Organotrifluoroborates
Suzuki-Miyaura Cross-Coupling with Organotrifluoroborates
The Versatility of Organotrifluoroborates
The Versatility of Organotrifluoroborates
Organotrifluoroborates from Aldrich
Product # Description Molecular Formula
684961 Potassium 2-furantrifluoroborate 684961
711101 Potassium pyridine-3-trifluoroborate 711101
711144 Potassium 1H-pyrazole-3-trifluoroborate 711144
681342 Potassium (bromomethyl)trifluoroborate
718947 Potassium (iodomethyl)trifluoroborate
655228 Potassium vinyltrifluoroborate
730432 Potassium propynyltrifluoroborate 730432
659274 Potassium allyltrifluoroborate 659274
662984 Potassium cyclopropyltrifluoroborate 662984
711098 Potassium pyrimidine-5-trifluoroborate 711098
723797 Potassium thiophene-2-trifluoroborate 723797
667145 Potassium sec-butyltrifluoroborate 667145
728799 Potassium 2-bromopyridine-3-trifluoroborate 728799
717517 Potassium 6-bromopyridine-3-trifluoroborate 717517
728802 Potassium 2-chloropyridine-3-trifluoroborate 728802
723568 Potassium pyridine-4-trifluoroborate 723568
666017 Potassium cyclopentyltrifluoroborate 666017
710075 (Thiomorpholinium-4-ylmethyl)trifluoroborate - internal salt 710075
706116 Potassium benzofurazan-5-trifluoroborate 706116
723592 Potassium (2-nitrophenyl)trifluoroborate 723592
719013 (Piperidinium-1-ylmethyl)trifluoroborate - internal salt 719013
710083 [(4-methylpiperazin-1-ium-1-yl)methyl]trifluoroborate - internal salt 710083
723991 Potassium 2-fluoro-5-formylphenyltrifluoroborate 723991
659789 Potassium 3-carboxyphenyltrifluoroborate 659789
563056 Potassium benzyltrifluoroborate
710881 Potassium phenoxymethyltrifluoroborate 710881
For a complete list of organotrifluoroborates from Aldrich, visit Aldrich.com/tfb.
References:
  1.  Molander, G. A.; Figueroa, R. Aldrichimica Acta . 2005, 38, 49-56.
  2.  Genêt, J.-P.; Darses, S. Eur. J. Org. Chem. 2003, 4313-4327.