Chemistry > Drug Discovery > Coated Multiwell Plates > Specification Profiles
Coated Multiwell Plates
Specification Profiles
    Biotinylated Biomolecule Capture
    Cell-Based Assays
    Epitope Tag and Protein Capture Plates
    Back to Coated Multiwell Plates home
Biotinylated Biomolecule Capture
SigmaScreen™ Streptavidin Coated Plates
Description sg_dd_hts_tbicon
Biotin
Binding Capacity
%CV
well to well
%CV
plate to plate
DNase
Protease
96-well white, M 5557  n/a
>25 pmole
biotin per well
<10%
<10%
None
detected
None
detected
384-well clear, S 8686
384-well white, S 8561
384-well black, S 8811
>4 pmole
biotin per well
<10%
<10%
None
detected
None
detected
SigmaScreen™ Streptavidin High Capacity Coated Plates
Description sg_dd_hts_tbicon
Biotin
Binding Capacity
%CV
well to well
%CV
plate to plate
DNase
Protease
96-well clear, S 6940
>300 pmole
biotin per well
<15%
<15%
None
detected
None
detected
96-well black, S 8940 n/a
>200 pmole
biotin per well
<15%
<15%
None
detected
None
detected
96-well white, S 9065 n/a
>250 pmole
biotin per well
<15%
<15%
None
detected
None
detected
384-well clear, S 8815
>150 pmole
biotin per well
<15%
<15%
None
detected
None
detected
384-well black, S 9190
>100 pmole
biotin per well
<15%
<15%
None
detected
None
detected
384-well white, S 9315
>125 pmole
biotin per well
<15%
<15%
None
detected
None
detected
SigmaScreen™ ExtrAvidin Coated Plates
Description sg_dd_hts_tbicon
Biotin
Binding Capacity
%CV
well to well
%CV
plate to plate
DNase
Protease
96-well clear, E 3027  
>25 pmole
biotin per well
<10%
<10%
None
detected
None
detected
Back to Top
Cell-Based Assays
SigmaScreen™ Poly-D-Lysine Coated Plates
Description sg_dd_hts_tbicon
Bioburden
Endotoxin
Plate Performance
96-well clear, M 3555
96-well white, M 5682
96-well black, M 5307
<0.52% wells contaminated
(<10 wells out of 20 plates)
<0.5 EU/mL for
all wells tested
Tested wells show >2X signal of negative control wells. Visual confirmation of greater number of cells (BHK 21) adhered to test wells than negative control wells.
384-well clear, S 7436
384-well white, S 7561
384-well black, S 7686
<0.52% wells contaminated
(<40 wells out of 20 plates)
<0.5 EU/mL for
all wells tested
Tested wells show >1.5X signal of negative control wells. Visual confirmation of greater number of cells (BHK 21) adhered to test wells than negative control wells.
SigmaScreen™ Collagen I Coated Plates
Description sg_dd_hts_tbicon
Bioburden
Endotoxin
Plate Performance
96-well clear, S 8813
96-well white, S 3315
96-well black, S 3190
<0.52% wells contaminated
(<10 wells out of 20 plates)
<0.5 EU/mL for
all wells tested
Test wells show approximately 90% of HT 1080 cells spread, as compared to negative control wells which demonstrate rounded cellular morphology and poor spreading.
384-well clear, S 3440
384-well white, S 3565
384-well black, S 3690
<0.52% wells contaminated
(<40 wells out of 20 plates)
<0.5 EU/mL for
all wells tested
Test wells show approximately 90% of HT 1080 cells spread, as compared to negative control wells which demonstrate rounded cellular morphology and poor spreading.
Back to Top
Epitope Tag and Protein Capture Plates
HIS-Select™ HC Nickel Coated Plate
Description sg_dd_hts_tbicon
%CV well to well
%CV plate to plate
Binding Capacity
96-well clear, S 5563
<15%
<15%
>4 5g protein per well
384-well clear, H 1661  
<15%
<15%
>2 5g protein per well
HIS-Select™ HS Nickel Coated Plate
Description sg_dd_hts_tbicon
%CV well to well
%CV plate to plate
Minimum Detectable
Protein Binding
96-well strip, S 5688
<10%
<10%
Absorbance of 1 ng/well
>0.20
Anti-FLAG™ HS M2 Coated Plate
Description sg_dd_hts_tbicon
%CV well to well
%CV plate to plate
Binding Sensitivity
96-well clear, P 2983
<10%
<10%
Detection of <1 ng of FLAG-BAP-His Control Protein above background
Protein G HS Coated Plate
Description sg_dd_hts_tbicon
%CV well to well
%CV plate to plate
Binding Sensitivity
(signal/noise ratio)
96-well clear, S 2063
<10%
<10%
>1.5 (test wells show detection of an antibody Alkaline Phosphatase conjugate of <1 ng above background)
Glutathione Coated Plate
Description sg_dd_hts_tbicon
%CV well to well
%CV plate to plate
Binding Capacity
Binding Sensitivity
96-well clear, P 3233 n/a
<10%
<10%
>10 ng purified GST
<1 ng purified GST detected
Back to Top
Back to Assay Development and Screening Home
Sigma’s line of coated plates utilize Corning ® base plates.
Corning is a registered trademark of Corning Incorporated, Corning, NY 14831-0001.
SigmaScreen is a trademark of Sigma-Aldrich Corporation, St. Louis, MO 63103.