Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Coated Multiwell Plates
Product Lines
    Biotinylated Biomolecule Capture
    Cell-Based Assays
    Epitope Tag and Protein Capture Plates
    Back to Coated Multiwell Plates home
Click here for a complete specification profile of Sigma's coated multiwell plates.
Biotinylated Biomolecule Capture
For superior binding capacity of biotinylated molecules, SigmaScreen™ Streptavidin Coated Plates offer up to 2X the binding capacity of other commercial suppliers for standard streptavidin coated plates.
Product
Code
Description Pack Size
M4058 SigmaScreen Streptavidin Coated Plates, 96-well, black
5 each
4x25 each
S8686 SigmaScreen Streptavidin Coated Plates, 384-well, clear
5 each
4x25 each
M5432 SigmaScreen Streptavidin Coated Plates, 96-well, clear
5 each
4x25 each
M3433 SigmaScreen Streptavidin Coated Strip-Well Plates, 96-well
1 each
5 each
Back to Top
Patent pending technology allows for the highest binding capacity available today. With up to 10X more than standard streptavidin and 3X greater than other high-binding plates, Sigma's Streptavidin High Capacity Coated Plates provide a platform to capture enough peptide or protein for elution and various downstream applications.
Product
Code
Description Pack Size
S6940 SigmaScreen Streptavidin High Capacity Coated Plates, 96-well, clear
1 each
5 each
Back to Top
Cell-Based Assays
Get up to 60% greater cell adherence with SigmaScreen™ Poly-D-Lysine Coated Plates.
Product
Code
Description Pack Size
S7686 SigmaScreen Poly-D-Lysine Coated Plates, 384-well, black
5 each
5x20 each
M5307 SigmaScreen Poly-D-Lysine Coated Plates, 96-well, black
5 each
5x20 each
Z382493 SigmaScreen™ Poly-D-Lysine coated plates 96-well, clear
5 each
5x20 each
M5682 SigmaScreen Poly-D-Lysine Coated Plates, 96-well, white
5 each
5x20 each
Back to Top
All PDL plate formats are clear bottom and include lids.
SigmaScreen™ Collagen I Coated Plates offer rapid and uniform cell proliferation.
Product
Code
Description Pack Size
S3690 SigmaScreen Collagen I Coated Plates, 384-well, black
5 each
10x5 each
S3190 SigmaScreen Collagen I Coated Plates, 96-well, black
5 each
10x5 each
S3565 SigmaScreen Collagen I Coated Plates, 384-well, white
5 each
10x5 each
S3315 SigmaScreen Collagen I Coated Plates, 96-well, white
5 each
10x5 each
Back to Top
All Collagen I plate formats are clear bottom and include lids.
Epitope Tag and Protein Capture Plates
Product
Code
Description Pack Size
S5563 HIS-Select™ HC Nickel Coated 96-well, clear
1 each
5 each
S5688 HIS-Select™ HS Nickel Coated 96-well, strip
1 each
5 each
P2983 Anti-FLAG™ HS M2 Coated 96-well, clear
1 each
5 each
H9412 HIS-Select™ iLAP High Capacity (HC) Nickel Coated 96 well, clear 1 each
5 each
Back to Top
Sigma’s line of coated plates utilize Corning® base plates.
To Order Call: 1-800-325-3010
Technical Service: 1-800-325-5832
Corning is a registered trademark of Corning Incorporated, Corning, NY 14831-0001.
SigmaScreen is a trademark of Sigma-Aldrich Corporation, St. Louis, MO 63103.