|
Peptides Synthesis
Building Blocks for Peptide Synth.
|
Sigma-Aldrich offers a comprehensive portfolio of high quality building blocks for peptide synthesis. All proteinacious amino acids are available with multiple combinations of different protecting groups and schemes — from standard building blocks for Boc-/Bzl- or Fmoc-/tBu- Solid Phase Peptide Synthesis to Z- Na-protected compounds for Solution Phase Synthesis. Orthogonal and other selectively removable protecting groups extend the scope of peptide synthesis to include unnatural, cyclic, phosphorylated, and labeled peptides.