Chemistry > Drug Discovery > Resin Explorer > Resins for Solid-Phase Synthesis
Resin Explorer
Resins for Solid-Phase Synthesis
  Alkenyl Resins
Amine-Functionalized Resins
BHA Resins
Br-Functionalized Resins
Chloromethyl Resins
CHO-Functionalized Resins
Cl-Functionalized Resins
COOH-Functionalized Resins
HypoGel® Resins
I-Functionalized Resins
JandaJels™
MBHA Resins
Miscellaneous Resins
NH2-Functionalized Resins
Nitrophenyl Carbonates
OH-Functionalized Resins
Oxime Resins
Peg Cross-Linked
Boc-/Bzl Peptide Synthesis
Fmoc-/tBu Peptide Synthesis
Phoxime Resin
Resin Matrices
Rink Acid Resins
Rink Resins
Safety Catch Resins
TentaGel® Resins
Thiol-Functionalized Resins
Triazine-Based Resins
Trityl Amines
Wang Resins
Wang Derivatives
Alkenyl Resins        
Product Name Loading Mesh Size Polymer Matrix Product # 
REM resin 0.1-0.5 mmol/g 200-400 PS crosslinked with 1% acrylate ester 471739
REM resin ca.1.0 mmol/g 200-400 PS crosslinked with 1% DVB 510173
REM resin 1.1 mmol/g 100-200 PS crosslinked with 1% DVB 537608
REM resin ca. 0.5 mmol/g 70-90 PS crosslinked with 1 % DVB 537381
REM resin ~ 0.9 mmol/g 100-300 PS crosslinked with 1 % DVB 83558
Vinyl sulfone, polymer-bound 0.4-1.2 mmol/g 100-200 PS crosslinked with 1% DVB 564737
Vinyl-polystyrene Capacity ~1.5 mmol/g, ~ 1.5 mmol/g 200-400 PS crosslinked with 1 % DVB 17761
Amine-Functionalized Resins        
Product Name Loading Mesh Size Polymer Matrix Product # 
Amidine resin ~ 0.9 mmol/g 100-300 PS crosslinked with 2% DVB 06565
N-(4-Benzyloxybenzyl)hydroxylamine polymer bound Capacity ~0.5 mmol/g, resin ~ 0.5 mmol/g 200-400 PS crosslinked with 1 % DVB 17762
(Aminomethyl)polystyrene 1.0-1.5 mmol/g 200-400 PS crosslinked with 1% DVB 474045
(Aminomethyl)polystyrene 0.5-1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 514772
(Aminomethyl)polystyrene 1.0 mmol/g 100-200 PS crosslinked with 1% DVB 522759
(Aminomethyl)polystyrene 0.5-1.0 mmol/g 70-90 PS crosslinked with 1% DVB 515612
(Aminomethyl)polystyrene Particle size 70-90 mesh 1.0-1.5 mmol/g 70-90 PS crosslinked with 1% DVB 515620
(Aminomethyl)polystyrene 1.5-2.0 mmol/g 70-90 PS crosslinked with 1% DVB 515639
(R)-(+)-a-methylbenzylamine, polymer-bound ca.2.0 mmol/g 100-200 PS crosslinked with 1 % DVB 576395
2-Chlorotrityl Knorr resin 0.5-1.0 mmol/g 100-200 PS crosslinked with 1% DVB 549401
2-N-Fmoc-Amino-dibenzocyclohepta-1,4-diene, polymer-bound 0.5-1.3 mmol/g 100-200 PS crosslinked with 1% DVB 564672
4-[4-(1-Fmoc-aminoethyl)-2-methoxy-5-nitrophenoxy]butyramidomethyl-polystyrene 0.6-1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 564753
4-Benzyloxybenzylamine, polymer-bound  0.5-1.3 mmol/g 100-200 PS crosslinked with 1 % DVB 564818
4-Carboxybenzenesulfonamide, polymer-bound 0.8-1.0 50-90 PS crosslinked with 1% DVB 534625
4-Carboxybenzenesulfonamide, polymer-bound 0.7-1.3 mmol/g 100-200 PS crosslinked with 1% DVB 549347
(Aminomethyl)polystyrene macroporous 1.5-3.0 mmol/g 30-60 PS crosslinked with 1% DVB 564095
(Aminomethyl)polystyrene macroporous 1.5-3.0 mmol/g 70-90 PS crosslinked with 1% DVB 564109
Bis(tert-butoxycarbonyl)thiopseudourea, polymer-bound ca. 1.0 mmol/g 100-200 PS crosslinked with 1% DVB 540382
Dimethylaminomethyl-polystyrene 1.0-1.5 mmol/g 20-50 PS crosslinked with 1% DVB 426997
Fmoc-3-amino-3-(2-nitrophenyl)propionic acid, polymer-bound 0.5-1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 564745
N-Methyl aminomethylated polystyrene 1.0-1.5 mmol/g 100-200 PS crosslinked with 1% DVB 538779
PAL resin 0.4-0.8 mmol/g 100-200 PS crosslinked with 1% DVB 549444
Sieber amide resin 0.3-0.6 mmol/g 50-100 PS crosslinked with 1% DVB 533955
tert-Butyl N-(2-mercaptoethyl)carbamate, polymer-bound ca. 2 mmol/g 100-200 PS crosslinked with 1% DVB 579378
Triphenylchloromethane-4-carboxamide polymer bound 1.1-1.3 mmol/g 200-400 PS cross-linked with 1% DVB 00481
BHA Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
2-Chlorobenzhydryl chloride, polymer-bound 1.6-2.2 mmol/g 100-200 PS cross-linked with 1% DVB 572632
2-Chlorobenzhydryl chloride, polymer-bound 1.6-2.2 mmol/g 100-200 PS cross-linked with 1% DVB 572632
HMPB-benzhydrylamine polymer bound 0.7-1.3 mmol/g 100-200 PS cross-linked with 1% DVB 564850
HMPB-benzhydrylamine polymer bound 0.7-1.3 mmol/g 100-200 PS cross-linked with 1% DVB 564850
4-Methylbenzhydrol, polymer-bound 0.5-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 564206
4-Methylbenzhydrol, polymer-bound 0.5-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 564206
4-Methylbenzhydryl alcohol, polymer-bound 1.6-2.0 mmol/g 100-200 PS cross-linked with 1% DVB 572624
4-Methylbenzhydryl alcohol, polymer-bound 1.6-2.0 mmol/g 100-200 PS cross-linked with 1% DVB 572624
4-Methylbenzhydryl chloride, polymer-bound 1.6-2.2 mmol/g 100-200 PS cross-linked with 1% DVB 572640
4-Methylbenzhydryl chloride, polymer-bound 1.6-2.2 mmol/g 100-200 PS cross-linked with 1% DVB 572640
Benzhydrol, polymer-bound 0.5-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 564192
Benzhydryl chloride, polymer-bound 1.6-2.2 mmol/g 100-200 PS cross-linked with 1% DVB 572616
Benzhydryl chloride, polymer-bound 1.6-2.2 mmol/g 100-200 PS cross-linked with 1% DVB 572616
Benzhydrylamine polymer-bound hydrochloride 0.7-1.4 mmol/g 100-200 PS cross-linked with 1% DVB 532460
Benzhydrylamine polymer-bound hydrochloride 0.7-1.4 mmol/g 200-400 PS cross-linked with 1% DVB 532479
Benzhydrylamine polymer-bound hydrochloride 0.7-1.4 mmol/g 100-200 PS cross-linked with 2% DVB 532436
Benzhydrylamine hydrochloride polymer bound ~ 1.1 mmol/g 200-400 PS cross-linked with 1% DVB 12106
Benzhydrylamine hydrochloride polymer bound ~ 1.5 mmol/g 100-200 PS cross-linked with 2% DVB 12110
Br-Functionalized Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
4-(Benzyloxy)benzyl bromide polymer bound 0.5-1.0 mmol/g 100-200 PS cross-linked with 1% DVB 534854
4-Bromopolystyrene 0.5-2.0 mmol/g 100-200 PS cross-linked with 1% DVB 549355
Brominated PPOA resin 0.5-1.0 mmol/g 100-200 PS cross-linked with 1% DVB 533920
Brominated Wang resin 0.5-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 534471
Brominated Wang resin ~ 1.0 mmol/g 70-90 PS cross-linked with 1% DVB 10984
Bromoacetal, polymer-bound 1.0-1.5 mmol/g 75-100 PS cross-linked with 1% DVB 564869
Bromoacetic acid, polymer-bound 0.85-1.2 mmol/g 100-200 PS cross-linked with 1% DVB 537500
Bromopolystyrene 1.2-1.3 mmol/g 100-400 PS cross-linked with 2% DVB 18106
Bromopolystyrene ~ 2.5 mmol/g 100-400 PS cross-linked with 2% DVB 18099
Bromopolystyrene ~4 mmol/g 200-400 PS cross-linked with 1% DVB 18100
Bromopolystyrene ~ 4 mmol/g 200-400 PS cross-linked with 2% DVB 18104
Bromopolystyrene ~ 5.5 mmol/g 200-400 PS cross-linked with 2% DVB 18102
Bromopolystyrene ~ 5.5 mmol/g 100-400 PS cross-linked with 2% DVB 18101
HypoGel® 200 Br ~ 0.8 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB;) PS beads 79709
Polystyrene A-Br for peptide synthesis 0.8-1.2 mmol/g 160-200 mm PS cross-linked with 1% DVB 81534
Polystyrene A-Br for peptide synthesis 0.8-1.2 mmol/g 500-560 mm PS cross-linked with 1% DVB 81535
Selenium bromide, polymer-bound       535958
TentaGel HL-Br ~ 0.48 mmol/g ~ 110 mm PEG grafted on low cross-linked PS beads 86331
TentaGel MB-Br ~ 0.40 mmol/g 140-170 mm PEG grafted on low cross-linked PS beads 86353
TentaGel S-Br ~ 0.26 mmol/g ~ 90 mm PEG grafted on low cross-linked PS beads 86363
TentaGel S-Br ~ 0.25 mmol/g ~ 130 mm PEG grafted on low cross-linked PS beads 86357
Chloromethyl Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
[5-[4-(Chloromethyl)phenyl]pentyl]styrene, polymer-bound 0.75-1.25 mmol/g 100-200 PS cross-linked with 1% DVB 513776
4-(Benzyloxy)benzyl chloride polymer bound 0.5-1.0 mmol/g 100-200 PS cross-linked with 1% DVB 537276
4-Methoxybenzhydryl chloride, polymer-bound ca.1.0 mmol/g 50-90 PS cross-linked with 1% DVB 534692
CHO-Functionalized Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
(4-Formyl-3-methoxyphenoxymethyl)polystyrene 1.0-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 516449
(4-Formyl-3-methoxyphenoxymethyl)polystyrene ~ 1.2 mmol/g 100-300 PS crosslinked with 2% DVB 09942
3-Benzyloxybenzaldehyde, polymer-bound 1.5-2.0 mmol/g 200-400 PS cross-linked with 1% DVB 477885
4-(4-Formyl-3-methoxyphenoxy)butyryl AM resin 0.8-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 541583
4-Benzyloxy-2,6-dimethoxybenzaldehyde,polymer-bound 1.0-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 538213
Formylpolystyrene 0.7-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 532401
Formylpolystyrene 2.0-3.0 mmol/g 100-200 PS cross-linked with 2% DVB 532428
HypoGel® 200 CHO ~ 0.8 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB;) PS beads 75401
HypoGel® 200 FP ~ 0.8 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB;) PS beads 78528
Indole resin ca. 0.5 mmol/g 200-400 PS crosslinked with 1 % DVB 516279
Indole resin ~ 1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 20529
Polystyrene A-CH(OEt)2 0.8-1.2 mmol/g 160-200 mm PS crosslinked with 1 % DVB 81537
Polystyrene A-CH(OEt)2 0.8-1.2 mmol/g 500-560 mm PS crosslinked with 1 % DVB 81538
Polystyrene AM-CH(OEt)2 Capacity (N) 0.8-1.2 mmol/g, resin 0.8-1.2 mmol/g 100-200 PS crosslinked with 1 % DVB 81543
Polystyrene AM-CH(OEt)2 for peptide synthesis Capacity (N) 0.8-1.2 mmol/g, resin 0.8-1.2 mmol/g 200-400 PS crosslinked with 1 % DVB 81544
Polystyrene AM-CH(OEt)2 Capacity (N) 0.8-1.2 mmol/g, resin 0.8-1.2 mmol/g 160-200 mm PS crosslinked with 1 % DVB 81545
TentaGel HL-CH(OEt)2 Capacity ~0.40 mmol/g, ~ 0.40 mmol/g ~ 110 mm PEG grafted on low cross-linked PS beads 86332
TentaGel MB-CH(OEt)2 Capacity ~0.40 mmol/g, ~ 0.40 mmol/g 140-170 mm PEG grafted on low cross-linked PS beads 86354
Cl-Functionalized Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
Benzoyl chloride polymer bound ~ 2.1 mmol/g 100-300 PS crosslinked with 2% DVB 12973
(Chloromethyl)polystyrene ~ 0.8 mmol/g 200-400 PS cross-linked with 1% DVB 63865
(Chloromethyl)polystyrene ~ 1.1 mmol/g 200-400 PS cross-linked with 1% DVB 63852
(Chloromethyl)polystyrene ~ 1.7 mmol/g 200-400 PS cross-linked with 1% DVB 63866
(Chloromethyl)polystyrene ~ 4.5 mmol/g 200-400 PS cross-linked with 1% DVB 63863
(Chloromethyl)polystyrene ~ 1.5 mmol/g 70-90 PS cross-linked with 1% DVB 10983
(Chloromethyl)polystyrene ~ 2 mmol/g 250-300 mm PS cross-linked with 1% DVB 17296
(Chloromethyl)polystyrene ~ 1.0 mmol/g 400-500 mm PS cross-linked with 1% DVB 17298
(Chloromethyl)polystyrene ~ 2 mmol/g 400-500 mm PS cross-linked with 1% DVB 17297
(Chloromethyl)polystyrene gel type ~ 2 mmol/g 70-150 PS cross-linked with 1.5% DVB 63858
(Chloromethyl)polystyrene ~ 1.4 mmol/g 200-400 PS cross-linked with 2% DVB 63872
(Chloromethyl)polystyrene ~ 2.1 mmol/g 200-400 PS cross-linked with 2% DVB 63874
(Chloromethyl)polystyrene >= 4.0 mmol/g 100-200 PS crosslinked with 2% DVB 75063
(Chloromethyl)polystyrene ~ 4.3 mmol/g 200-400 PS cross-linked with 2% DVB 63873
(Chloromethyl)polystyrene gel type ~ 2 mmol/g 70-150 PS cross-linked with 2.5% DVB 63861
(Chloromethyl)polystyrene gel type ~ 2 mmol/g 18-70 PS cross-linked with 2.5% DVB 63862
(Chloromethyl)polystyrene gel type ~ 5 mmol/g 16-50 PS cross-linked with 3.5% DVB 63864
(Chloromethyl)polystyrene gel type ~ 5 mmol/g 16-50 PS cross-linked with 4.5% DVB 63867
(Chloromethyl)polystyrene ~ 5.5 mmol/g 16-50 PS cross-linked with 5.5% DVB 63868
(Chloromethyl)polystyrene gel type ~ 5.5 mmol/g 16-50 PS cross-linked with 5.8% DVB 63875
(Chloromethyl)polystyrene ~ 0.7 mmol/g 200-400 PS cross-linked with 2% DVB 63871
Chloroacetyl, polymer-bound 2.5-5.0 mmol/g 100-200 PS crosslinked with 1 % DVB 564915
Merrifield's resin 2.5-4.0 mmol/g 50-100 PS crosslinked with 1 % DVB 577901
Merrifield's resin 1.0-1.5 mmol/g 200-400 PS crosslinked with 1 % DVB 221481
Merrifield's resin 1.0-1.5 mmol/g 200-400 PS crosslinked with 2 % DVB 221503
Merrifield's resin 1.5-2.0 mmol/g 200-400 PS crosslinked with 1 % DVB 437956
Merrifield's resin 2.0-2.5 mmol/g 200-400 PS crosslinked with 2 % DVB 449113
Merrifield's resin 3.0-3.5 mmol/g 200-400 PS crosslinked with 1 % DVB 456969
Merrifield's resin 3.5-4.5 mmol/g 200-400 PS crosslinked with 1 % DVB 474517
Merrifield's resin 0.5-1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 516074
Merrifield's resin 0.8-1.2 mmol/g 100-200 PS crosslinked with 1 % DVB 522716
Merrifield's resin 1.0-1.5 mmol/g 100-200 PS crosslinked with 1 % DVB 515957
Merrifield's resin 1.5-2.0 mmol/g 100-200 PS crosslinked with 1 % DVB 497045
Merrifield's resin 2.0-3.0 mmol/g 100-200 PS crosslinked with 1 % DVB 49,705-3
Merrifield's resin 3.5-4.5 mmol/g 100-200 PS crosslinked with 1 % DVB 49,706-1
Merrifield's resin 1.0-1.5 mmol/g 70-90 PS crosslinked with 1 % DVB 49,919-6
Merrifield's resin 1.5-2.0 mmol/g 70-90 PS crosslinked with 1 % DVB 49,918-8
Merrifield's resin 1.7-2.2 mmol/g 50-100 PS crosslinked with 1 % DVB 52,272-4
Merrifield's resin 1.7-2.2 mmol/g 50-100 PS crosslinked with 1 % DVB 52,273-2
Merrifield's resin 3.6-4.2 mmol/g 50-100 PS crosslinked with 1 % DVB 52,274-0
COOH Functionalized Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
Carboxyethylpolystryrene 0.8-1.5 mmol/g 100-200 PS crosslinked with 1 % DVB 53,862-0
Carboxypolystyrene 0.5-1.5 mmol/g 100-200 PS crosslinked with 1 % DVB 53,237-1
Carboxypolystyrene 1.6-3.0 mmol/g 100-200 PS crosslinked with 1 % DVB 53,239-8
HypoGel® 200 COOH ~ 0.8 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB) PS beads 78967
Polystyrene A-COOH 0.8-1.2 mmol/g 160-200 mm PS crosslinked with 1 % DVB 81536
Polystyrene A-COOH 0.8-1.2 mmol/g 500-560 mm PS crosslinked with 1 % DVB 81539
Polystyrene AM-COOH 0.8-1.2 mmol/g 100-200 PS crosslinked with 1 % DVB 81546
Polystyrene AM-COOH for peptide synthesis 0.8-1.2 mmol/g 200-400 PS crosslinked with 1 % DVB 81547
Polystyrene AM-COOH 0.6-0.8 mmol/g 160-200 mm PS crosslinked with 1 % DVB 81548
TentaGel HL-COOH ~ 0.40 mmol/g ~ 110 mm PEG grafted on low cross-linked PS beads 86333
TentaGel MB-COOH ~ 0.40 mmol/g 140-170 mm PEG grafted on low cross-linked PS beads 86355
TentaGel S-COOH ~ 0.25 mmol/g ~ 90 mm PEG grafted on low cross-linked PS beads 86336
TentaGel S-COOH ~ 0.25 mmol/g ~ 130 mm PEG grafted on low cross-linked PS beads 86358
HypoGel® Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
HypoGel® 200 FMP ~ 0.6 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB) PS beads 74911
HypoGel® 200 PHB ~ 0.7 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB) PS beads 75513
HypoGel® 200 Trt-OH ~ 0.7 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB) PS beads 75040
HypoGel® 200 HMB ~ 0.8 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB) PS beads 78619
I-Functionalized Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
4-Iodophenol, polymer-bound 1.0-2.0 mmol/g 50-100 PS crosslinked with 1 % DVB 570788
Iodopolystyrene 3.5-4.0 mmol/g 50-100 PS crosslinked with 1 % DVB 570796
JandaJels™       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
Amine-Functionalized      
JandaJelä- Rink amide 0.3-1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 570508
JandaJelä-NH2 1.0 mmol/g 50-100 PS crosslinked with 2 % DVB 524603
JandaJelä-NH2 1.0 mmol/g 100-200 PS crosslinked with 2 % DVB 524611
JandaJelä-NH2 1.0 mmol/g 200-400 PS crosslinked with 2 % DVB 524638
Cl-Functionalized      
JandaJelä-Cl Particle size 50-100 mesh 0.45-0.70 mmol/g 50-100 PS crosslinked with 2 % DVB 524530
JandaJelä-Cl Particle size 100-200 mesh 0.45-0.70 mmol/g 100-200 PS crosslinked with 2 % DVB 524549
JandaJelä-Cl Particle size 200-400 mesh 0.45-0.70 mmol/g 200-400 PS crosslinked with 2 % DVB 524557
OH-Functionalized      
JandaJelä-4-Mercaptophenol ca. 2.0 mmol/g 100-200 PS crosslinked with 1 % DVB 569674
JandaJelä-OH Particle size 50-100 mesh 1.0 mmol/g 50-100 PS crosslinked with 2 % DVB 524565
JandaJelä-OH Particle size 200-400 mesh 1.0 mmol/g 200-400 PS crosslinked with 2 % DVB 524581
JandaJelä-OH Particle size 100-200 mesh 1.0 mmol/g 100-200 PS crosslinked with 2 % DVB 524573
Reagents      
JandaJelä- 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide 1.5-2.0 mmol/g 50-100 PS crosslinked with 2 % DVB 567248
JandaJelä- 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide 1.5-2.0 mmol/g 100-200 PS crosslinked with 2 % DVB 567256
JandaJelä- 1,3,4,6,7,8-hexahydro-2H-pyrimido-[1,2-a] pyrimidine 2.3 mmol/g 100-200 PS crosslinked with 1 % DVB 570494
JandaJelä-morpholine ca. 3.6 mmol/g 50-100 PS crosslinked with 1 % DVB 578819
JandaJelä polypyridine ca. 8.0 mmol/g 100-200 PS crosslinked with 1 % DVB 576387
JandaJelä polypyridine ca. 8.0 mmol/g 50-100 PS crosslinked with 1 % DVB 578290
JandaJelä-Triphenylphosphine 3.0 mmol/g 50-100 PS crosslinked with 2 % DVB 533416
Wang      
JandaJelä-Wang 1.0 mmol/g 50-100 PS crosslinked with 2 % DVB 524646
JandaJelä-Wang 1.0 mmol/g 100-200 PS crosslinked with 2 % DVB 524654
JandaJelä-Wang 1.0 mmol/g 200-400 PS crosslinked with 2 % DVB 524662
MBHA Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
3[4'-(Hydroxymethyl)phenoxy] propionic acid-4-methylbenzhydrylamine resin 0.7-1.3 mmol/g 200-400 PS crosslinked with 2 % DVB 564702
4-(Hydroxymethyl)phenoxyacetic acid polymer-bound to MBHA resin 0.6-1.2 mmol/g 100-200 PS crosslinked with 1 % DVB 564133
4-(Hydroxymethyl)phenoxyacetic acid polymer-bound to MBHA resin 0.6-1.2 mmol/g 100-200 PS crosslinked with 1 % DVB 564141
HMBA-4-methylbenzhydrylamine polymer bound ca. 1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 564710
4-Methylbenzhydrylamine hydrochloride polymer bound Capacity (amine) ~0.7 mmol/g, resin ~ 0.7 mmol/g 100-200 PS cross-linked with 1% DVB 65837
4-Methylbenzhydrylamine hydrochloride polymer bound Capacity (amine) ~1.4 mmol/g, resin ~ 1.4 mmol/g 200-400 PS cross-linked with 1% DVB 65836
4-Methylbenzhydrylamine hydrochloride polymer bound 1 % cross-linked Pore Size 200-400 mesh 0.5-1.0 mmol/g 200-400 PS crosslinked with 1 % DVB 533173
4-Methylbenzhydrylamine hydrochloride polymer bound 2 % cross-linked Pore Size 200-400 mesh 0.7-1.4 mmol/g 200-400 PS crosslinked with 2 % DVB 532452
4-Methylbenzhydrylamine hydrochloride polymer bound 2 % cross-linked Pore Size 100-200 mesh 0.7-1.4 mmol/g 100-200 PS crosslinked with 2 % DVB 532444
4-Methylbenzhydrylamine hydrochloride polymer bound Polymer is polystyrene 0.5-1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 513229
4-Methylbenzhydrylamine polymer bound Capacity (amine) ~0.8 mmol/g, resin (CHN) ~ 0.8 mmol/g 100-200 PS cross-linked with 1% DVB 65835
Misc Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
(4-Diethylsilylbutyl)polystyrene Capacity ~1.5 mmol/g, ~ 1.5 mmol/g 200-400 PS crosslinked with 1 % DVB 17019
[3-(4-Methoxyphenyl-dimethylsilyl)propyl]polystyrene Capacity ~1.1 mmol/g, ~ 1.1 mmol/g 100-400 PS crosslinked with 2% DVB 02585
[4-(Diazomethyl)phenoxymethyl]polystyrene Capacity ~1.4 mmol/g, ~ 1.4 mmol/g 100-300 PS crosslinked with 2% DVB 17338
3,4-Dihydro-2H-pyran polymer bound Capacity ~1.0 mmol/g, resin ~ 1.0 mmol/g 100-300 PS crosslinked with 1 % DVB 17235
3,4-Dihydro-2H-pyran polymer bound binding capacity ~2 mmol/g, resin ~ 2 mmol/g 100-300 PS crosslinked with 2% DVB 37352
NH2-Functionalized Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
(Aminomethyl)polystyrene Capacity (amine) ~0.5 mmol/g, resin ~ 0.5 mmol/g 100-200 PS cross-linked with 1% DVB 08558
(Aminomethyl)polystyrene Capacity ~2 mmol/g, resin ~ 2 mmol/g 200-400 PS crosslinked with 1 % DVB 17026
(Aminomethyl)polystyrene Capacity (amine) ~1.0 mmol/g, resin ~ 1.0 mmol/g 400-500 mm PS cross-linked with 1% DVB 17301
(Aminomethyl)polystyrene Capacity (amine) ~2.0 mmol/g, resin ~ 2.0 mmol/g 400-500 mm PS cross-linked with 1% DVB 17302
(Aminomethyl)polystyrene Capacity (amine) ~1.5 mmol/g, resin ~ 1.5 mmol/g 200-400 PS cross-linked with 2% DVB 08564
(Aminomethyl)polystyrene Capacity (amine) ~0.5 mmol/g, resin ~ 0.5 mmol/g 100-200 PS cross-linked with 3% DVB 08555
(Aminomethyl)polystyrene Capacity (amine) ~1.4 mmol/g, resin ~ 1.4 mmol/g 100-200 PS cross-linked with 3% DVB 08556
(Aminomethyl)polystyrene Capacity (amine) ~1.1 mmol/g, resin ~ 1.1 mmol/g 100-200 PS cross-linked with 1% DVB 08559
(Aminomethyl)polystyrene Capacity (amine) ~0.6 mmol/g, resin ~ 0.6 mmol/g 200-400 PS cross-linked with 1% DVB 08566
  ~ 1.1 mmol/g 200-400 PS cross-linked with 1% DVB 09669
(Aminomethyl)polystyrene Capacity (amine) ~2 mmol/g, resin ~ 2 mmol/g 100-200 PS cross-linked with 3% DVB 08557
(Aminomethyl)polystyrene hydrochloride Capacity (amine) ~0.6 mmol/g, resin ~ 0.6 mmol/g 100-200 PS cross-linked with 1% DVB 08568
HypoGel® 200 NH2 ~ 0.9 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB) PS beads 76034
Polystyrene AM-NH2 Capacity (N) 0.8-1.2 mmol/g, resin 0.8-1.2 mmol/g 160-200 mm PS cross-linked with 1% DVB 81553
Polystyrene A-NH2 Capacity 0.8-1.2 mmol/g, 0.8-1.2 mmol/g 160-200 mm PS cross-linked with 1% DVB 81558
Polystyrene A-NH2 Capacity 0.8-1.2 mmol/g, 0.8-1.2 mmol/g 500-560 mm PS cross-linked with 1% DVB 81559
Polystyrene Microspheres 2-aminoethylated Capacity ~0.8 mmol/g, ~ 0.8 mmol/g ~ 5 mm PS crosslinked with 1 % DVB 53532
Polystyrol Microspheres 2-bromoethylated Capacity ~1.2 mmol/g, ~ 1.2 mmol/g ~ 5 mm PS crosslinked with 1 % DVB 13292
Polystyrol Microspheres 2-hydroxyethylated Capacity ~1.3 mmol/g, ~ 1.3 mmol/g ~ 5 mm PS crosslinked with 1 % DVB 15647
TentaGel HL-NH2 Capacity ~0.40 mmol/g, ~ 0.40 mmol/g ~ 110 mm PEG grafted on low cross-linked PS beads 86334
Tentagel M Br Microspheres Capacity ~0.2 mmol/g, ~ 0.2 mmol/g ~ 10 mm PEG grafted on low cross-linked PS beads 95677
Tentagel M NH2 Microspheres Capacity ~0.2 mmol/g, ~ 0.2 mmol/g ~ 10 mm PEG grafted on low cross-linked PS beads 18186
Tentagel M OH Microspheres Capacity ~0.2 mmol/g, ~ 0.2 mmol/g ~ 10 mm PEG grafted on low cross-linked PS beads 91924
TentaGel MB-NH2 Capacity ~0.40 mmol/g, ~ 0.40 mmol/g 140-170 mm PEG grafted on low cross-linked PS beads 86356
TentaGel S-NH2 Capacity (amine) ~0.26 mmol/g, ~ 0.26 mmol/g ~ 90 mm PEG grafted on low cross-linked PS beads 86364
TentaGel S-NH2 Capacity (N) ~0.45 mmol/g, resin ~ 0.45 mmol/g 150-200 mm PEG grafted on low cross-linked PS beads 86359
Nitrophenyl Carbonates       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
4-Nitrophenyl carbonate, polymer-bound 1 % cross-linked 1.5-2.5 mmol/g 100-200 PS crosslinked with 1 % DVB 494836
4-Nitrophenyl carbonate, polymer-bound 1 % cross-linked Pore Size 100-200 mesh 0.5-1.1 mmol/g 100-200 PS crosslinked with 1 % DVB 538485
4-Nitrophenyl carbonate, polymer-bound 1.0-1.5 mmol/g 200-400 PS crosslinked with 1 % DVB 515299
4-Nitrophenyl carbonate Merrifield resin Capacity (N) ~0.8 mmol/g, resin ~ 0.8 mmol/g 100-200 PS crosslinked with 1 % DVB 13587
4-Nitrophenyl carbonate-Wang resin Capacity (N) ~0.6 mmol/g, resin ~ 0.6 mmol/g 100-200 PS crosslinked with 1 % DVB 13649
OH-Functionalized Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
HMBA Resin      
4-Hydroxymethylbenzoic acid, polymer-bound 0.5-1.2 mmol/g 50-90 PS crosslinked with 1 % DVB 534536
Hydroxymethyl Resins      
Benzyl alcohol, polymer-bound Particle size 100-200 mesh 1.0-2.0 mmol/g 100-200 PS crosslinked with 1 % DVB 485128
Benzyl alcohol, polymer-bound Particle size 100-200 mesh 0.5-1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 514764
Benzyl alcohol, polymer-bound Particle size 70-90 mesh 0.5-1.0 mmol/g 70-90 PS crosslinked with 1 % DVB 515663
Benzyl alcohol, polymer-bound 1 % cross-linked Particle size 70-90 mesh 1.0-1.5 mmol/g 70-90 PS crosslinked with 1 % DVB 515671
Benzyl alcohol, polymer-bound 1 % cross-linked 2.0-3.0 mmol/g 70-90 PS crosslinked with 1 % DVB 499226
Wang Resins      
2-Pyridine-co-4-benzyloxybenzyl alcohol, polymer-bound 1 % cross-linked with divinylbenzene 0.4-0.9 mmol/g 100-200 PS crosslinked with 1 % DVB 564680
4-Benzyloxybenzyl alcohol, polymer-bound Particle size 200-400 mesh 0.5-1.0 mmol/g 200-400 PS crosslinked with 1 % DVB 514756
4-Benzyloxybenzyl alcohol, polymer-bound Particle size 200-400 mesh 1.0-1.5 mmol/g 200-400 PS crosslinked with 1 % DVB 477036
4-Benzyloxybenzyl alcohol, polymer-bound Particle size 200-400 mesh 1.5-2.0 mmol/g 200-400 PS crosslinked with 1 % DVB 520527
4-Benzyloxybenzyl alcohol, polymer-bound Particle size 100-200 mesh 0.5-1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 470635
4-Benzyloxybenzyl alcohol, polymer-bound Particle size 100-200 mesh 1.0-1.5 mmol/g 100-200 PS crosslinked with 1 % DVB 520608
4-Benzyloxybenzyl alcohol, polymer-bound Particle size 100-200 mesh 1.5-2.0 mmol/g 100-200 PS crosslinked with 1 % DVB 514748
4-Benzyloxybenzyl alcohol, polymer-bound Particle size 100-200 mesh 0.9 mmol/g 100-200 PS crosslinked with 1 % DVB 522775
4-Benzyloxybenzyl alcohol, polymer-bound Particle size 70-90 mesh 0.5-1.0 mmol/g 70-90 PS crosslinked with 1 % DVB 515647
4-Benzyloxybenzyl alcohol, polymer-bound Particle size 70-90 mesh 1.0-1.5 mmol/g 70-90 PS crosslinked with 1 % DVB 515655
4-Benzyloxybenzyl alcohol, polymer-bound Particle size 70-90 mesh 1.5-2.0 mmol/g 70-90 PS crosslinked with 1 % DVB 510335
Wang Derivatives      
Imidazole carbonate, polymer-bound 0.5-1.5 mmol/g 200-400 PS crosslinked with 1 % DVB 498238
Imidazole carbonate, polymer-bound 0.7-1.5 mmol/g 100-200 PS crosslinked with 1 % DVB 549363
Wang trichloroacetimidate resin Particle size 200-400 mesh 0.75-1.25 mmol/g 200-400 PS crosslinked with 1 % DVB 494798
Wang trichloroacetimidate resin Particle size 100-200 mesh 0.75-1.25 mmol/g 100-200 PS crosslinked with 1 % DVB 510327
Miscellaneous OH-Functionalized Resins      
(Hydroxymethyl)polystyrene Capacity ~1.1 mmol/g, resin ~ 1 mmol/g 100-200 PS cross-linked with 1% DVB 55761
(Hydroxymethyl)polystyrene Capacity ~0.6 mmol/g, resin ~ 0.6 mmol/g 200-400 PS cross-linked with 1% DVB 55763
(Hydroxymethyl)polystyrene Capacity ~1.1 mmol/g, resin ~ 1.1 mmol/g 200-500 PS cross-linked with 1% DVB 55761
(Hydroxymethyl)polystyrene Capacity ~1.7 mmol/g, ~ 1.7 mmol/g 100-300 PS cross-linked with 1% DVB 00084
Fmoc-Gly-HMPB-4-methyl-benzhydrylamine polymer bound Capacity ~0.5 mmol/g, resin ~ 0.5 mmol/g 200-400 PS cross-linked with 1% DVB 47557
Fmoc-Gly-HMPB-4-methyl-benzhydrylamine polymer bound Capacity ~0.7 mmol/g, resin ~ 0.7 mmol/g 200-400 PS cross-linked with 1% DVB 47558
HMBA-4-methylbenzhydrylamine polymer bound Capacity ~0.7 mmol/g, resin ~ 0.7 mmol/g 100-200 PS cross-linked with 1% DVB 53417
HMPA-(aminomethyl)polystyrene Capacity 0.3-0.7 mmol/g, resin 0.3-0.7 mmol/g 200-400 PS cross-linked with 1% DVB 53423
HMPA-(aminomethyl)polystyrene Capacity ~1.1 mmol/g, resin ~ 1.1 mmol/g 100-200 PS cross-linked with 1% DVB 53419
HMPA-(aminomethyl)polystyrene Capacity ~1.1 mmol/g, resin ~ 1.1 mmol/g 200-400 PS cross-linked with 1% DVB 53422
HMPB-(aminomethyl)polystyrene Capacity ~1.1 mmol/g, resin ~ 1.1 mmol/g 100-200 PS cross-linked with 1% DVB 53421
HMPB-(aminomethyl)polystyrene Capacity ~1.1 mmol/g, resin ~ 1.1 mmol/g 200-400 PS cross-linked with 1% DVB 53424
HMPB-(aminomethyl)polystyrene Capacity ~0.5 mmol/g, resin ~ 0.5 mmol/g 200-400 PS cross-linked with 1% DVB 53425
HMPB-4-methylbenzhydrylamine polymer bound Capacity ~0.5 mmol/g, resin ~ 0.5 mmol/g 200-400 PS cross-linked with 1% DVB 53429
HMPB-benzhydrylamine polymer bound Capacity ~0.5 mmol/g, resin ~ 0.5 mmol/g 200-400 PS cross-linked with 1% DVB 53427
HMPB-benzhydrylamine polymer bound Capacity ~0.9 mmol/g, resin ~ 0.9 mmol/g 200-400 PS cross-linked with 1% DVB 53426
HMPEA-(aminomethyl)polystyrene Capacity ~1.1 mmol/g, resin ~ 1.1 mmol/g 100-200 PS cross-linked with 1% DVB 53431
HypoGel® 200 OH ~ 0.8 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB;) PS beads 78482
Polystyrene A-HMPA Capacity (N) 0.7-1.0 mmol/g, resin 0.7-1.0 mmol/g 160-200 mm PS cross-linked with 1% DVB 81541
Polystyrene A-HMPA Capacity 0.7-1.0 mmol/g, 0.7-1.0 mmol/g 500-560 mm PS cross-linked with 1% DVB 81542
Polystyrene AM-HMPA Capacity 0.7-1.0 mmol/g, 0.7-1.0 mmol/g 160-200 mm PS cross-linked with 1% DVB 81549
TentaGel HL-OH Capacity ~0.40 mmol/g, ~ 0.40 mmol/g ~ 110 mm PEG grafted on low cross-linked PS beads 86337
TentaGel MB-OH Capacity ~0.40 mmol/g, ~ 0.40 mmol/g 140-170 mm PEG grafted on low cross-linked PS beads 86371
TentaGel S-OH Capacity ~0.26 mmol/g, ~ 0.26 mmol/g 100-200 mm PEG grafted on low cross-linked PS beads 86361
TentaGel S-OH Capacity ~0.26 mmol/g, ~ 0.26 mmol/g ~ 90 mm PEG grafted on low cross-linked PS beads 86365
Wang resin Capacity 0.6-1.0 mmol/g 0.6-1.0 mmol/g 200-400 PS cross-linked with 1% DVB 13618
Wang resin Capacity ~1 mmol/g ~ 1.0 mmol/g 200-400 PS cross-linked with 1% DVB 13611
Wang resin Capacity ~1.1 mmol/g, resin ~ 1.1 mmol/g 100-200 PS cross-linked with 1% DVB 13609
Wang resin Capacity ~2.9 mmol/g, resin ~ 2.9 mmol/g 100-200 PS cross-linked with 1% DVB 17016
Wang resin Capacity 1.0 mmol/g, 1.0 mmol/g 75-100 PS cross-linked with 1% DVB 17095
Wang resin Capacity 1.0-1.5 mmol/g, 1.0-1.5 mmol/g 70-90 PS cross-linked with 1% DVB 10982
Wang resin Capacity ~1.7 mmol/g, capacity : ~ 1.7 mmol/g 250-300 mm PS cross-linked with 1% DVB 17299
2-Chlorobenzhydrol, polymer-bound 1.0-2.0 mmol/g 100-200 PS crosslinked with 1 % DVB 543411
4-Hydroxy-2-methoxybenzyl alcohol, polymer-bound 0.6-1.1 mmol/g 50-90 PS crosslinked with 1 % DVB 560553
4-Hydroxy-2-methoxybenzyl alcohol, polymer-bound 0.3-0.6 mmol/g 50-90 PS crosslinked with 1 % DVB 540730
HMPA-(aminomethyl)polystyrene 0.7-1.3 mmol/g 100-200 PS crosslinked with 1 % DVB 564788
4-Mercaptophenol, polymer-bound 1.0-1.5 mmol/g 100-200 PS crosslinked with 1 % DVB 511714
4-Methoxybenzhydrol, polymer-bound ca.1.0 mmol/g 50-90 PS crosslinked with 1 % DVB 549932
4-Nitrobenzophenone oxime, polymer-bound 1.0-1.5 mmol/g 200-400 PS crosslinked with 1 % DVB 477044
PAM resin 0.7-1.3 mmol/g   PS crosslinked with 1 % DVB 549428
Sulfonylamido ethanol, polymer-bound 0.5-1.5 mmol/g 100-200 PS crosslinked with 1 % DVB 564419
4-Nitrobenzophenone oxime polymer bound ~ 1.1 mmol/g 100-200 PS cross-linked with 1% DVB 73083
Oxime Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
4-Chlorobenzophenone oxime, polymer-bound, 1.0-1.5 mmol/g 100-200 PS crosslinked with 1 % DVB 564184
4-Methoxybenzophenone oxime, polymer-bound 1.0-1.5 mmol/g 100-200 PS crosslinked with 1 % DVB 564168
Benzophenone oxime, polymer-bound 1.0-1.5 mmol/g 100-200 PS crosslinked with 1 % DVB 564176
Peg Cross-Linked       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
Ethylene glycol polymer bound ~ 2.8 mmol/g 200-400 PS crosslinked with 2% DVB 03773
Boc-/Bzl Peptide Synthesis        
Product Name Mesh Size Loading Polymer Matrix Product # 
MAP Core Resins      
Boc-Lys(Boc)-Lys[Boc-Lys(Boc)]-Cys(Acm)-b-Ala-O-PAM resin 200-400 ~ 1.6 mmol/g PS crosslinked with 1 % DVB 09852
Boc-Lys(Fmoc)-Lys[Boc-Lys(Fmoc)]-b-Ala-O-Pam resin 200-400 ~ 1.2 mmol/g PS crosslinked with 1 % DVB 09853
Boc-Lys(Boc)-Lys[Boc-Lys(Boc)]-Lys{Boc-Lys(Boc)-Lys[Boc-Lys(Boc)]}-b-Ala-O-PAM resin 200-400 ~ 1.8 mmol/g PS crosslinked with 1 % DVB 09854
Boc-Lys(Fmoc)-Lys[Boc-Lys(Fmoc)]-Lys{Boc-Lys(Fmoc)-Lys[Boc-Lys(Fmoc)]}-b-Ala-O-PAM resin 200-400 ~ 1.9 mmol/g PS crosslinked with 1 % DVB 09863
Boc-Lys(Boc)-Lys[Boc-Lys(Boc)]-Lys{Boc-Lys(Boc)-Lys[Boc-Lys(Boc)]}-Cys(Acm)-b-Ala-O-PAM resin 200-400 ~ 2.0 mmol/g PS crosslinked with 1 % DVB 09864
Preloaded PAM      
Boc-Ala-PAM resin 75-150 mm; 100-200 ~ 0.7 mmol/g PS cross-linked with 1% DVB 89336
Boc-D-alanine esterified with PAM resin 75-150 mm; 100-200 ~ 0.7 mmol/g PS cross-linked with 1% DVB 08936
Boc-b-Ala-PAM resin 75-150 mm; 100-200 ~ 0.5 mmol/g PS cross-linked with 1% DVB 09846
Boc-Arg(Tos)-PAM resin 75-150 mm; 100-200 ~ 0.6 mmol/g PS cross-linked with 1% DVB 43750
Boc-Asn-PAM resin 75-150 mm; 100-200 ~ 0.7 mmol/g PS cross-linked with 1% DVB 00994
Boc-Asp(OcHx)-PAM resin 75-150 mm; 100-200 ~ 0.6 mmol/g PS cross-linked with 1% DVB 38712
Boc-Cys(Acm)-PAM resin 75-150 mm; 100-200 ~ 0.7 mmol/g PS cross-linked with 1% DVB 61254
Boc-Cys(Mbzl)-PAM resin 75-150 mm; 100-200 ~ 0.6 mmol/g PS cross-linked with 1% DVB 96728
Boc-Gln-PAM resin 75-150 mm; 100-200 ~ 0.7 mmol/g PS cross-linked with 1% DVB 05628
Boc-Glu(OcHx)-PAM resin 75-150 mm; 100-200 ~ 0.7 mmol/g PS cross-linked with 1% DVB 56313
Boc-Gly-PAM resin 75-150 mm; 100-200 ~ 0.7 mmol/g PS cross-linked with 1% DVB 51591
Boc-His(Tos)-PAM resin 38-75 mm; 200-400 ~ 0.5 mmol/g PS cross-linked with 1% DVB 41614
Boc-Ile-PAM resin 75-150 mm; 100-200 ~ 0.7 mmol/g PS cross-linked with 1% DVB 73324
Boc-Leu-PAM resin 75-150 mm; 100-200 ~ 0.6 mmol/g PS cross-linked with 1% DVB 11883
Boc-D-Leu-PAM resin 75-150 mm; 100-200 ~ 0.7 mmol/g PS cross-linked with 1% DVB 40954
Boc-Lys(2-Cl-Z)-PAM resin 75-150 mm; 100-200 ~ 0.6 mmol/g PS cross-linked with 1% DVB 44089
Boc-Met(O)-PAM resin 75-150 mm; 100-200 ~ 0.6mmol/g PS cross-linked with 1% DVB 18752
Boc-Met-PAM resin 75-150 mm; 100-200 ~ 0.7 mmol/g PS cross-linked with 1% DVB 79164
Boc-Phe-PAM resin 75-150 mm; 100-200 ~ 0.6 mmol/g PS cross-linked with 1% DVB 50659
Boc-D-Phe-PAM resin 75-150 mm; 100-200 ~ 0.7 mmol/g PS cross-linked with 1% DVB 39562
Boc-Pro-PAM resin 75-150 mm; 100-200 ~ 0.7 mmol/g PS cross-linked with 1% DVB 23908
Boc-Ser(Bzl)-PAM resin 75-150 mm; 100-200 ~ 0.6 mmol/g PS cross-linked with 1% DVB 08939
Boc-Thr(Bzl)-PAM resin 75-150 mm; 100-200 ~ 0.6 mmol/g PS cross-linked with 1% DVB 00839
Boc-Trp(For)-PAM resin 75-150 mm; 100-200 ~ 0.6 mmol/g PS cross-linked with 1% DVB 56932
Boc-Tyr(2-Br-Z)-PAM resin 75-150 mm; 100-200 ~ 0.6 mmol/g PS cross-linked with 1% DVB 53626
Boc-Val-PAM resin 75-150 mm; 100-200 ~ 0.7 mmol/g PS cross-linked with 1% DVB 50554
Boc-D-Val-PAM resin 75-150 mm; 100-200 ~ 0.7 mmol/g PS cross-linked with 1% DVB 04371
Fmoc-/tBu Peptide Synthesis       back to top
Product Name Mesh Size Loading Polymer Matrix Product # 
Map Core Resins      
Fmoc-Lys(Fmoc)-Lys[Fmoc-Lys(Fmoc)]-b-Ala-O-Wang resin 100-200 ~ 1.0 mmol/g PS crosslinked with 1 % DVB 09850
Fmoc-Lys(Fmoc)-Lys[Fmoc-Lys(Fmoc)]-Lys{Fmoc-Lys(Fmoc)-Lys[Fmoc-Lys(Fmoc)]}-b-Ala-O-Wang resin 200-400 ~ 0.8 mmol/g PS crosslinked with 1 % DVB 09865
Preloaded TentaGel® S Trityl Resins      
TentaGel S Trt-Ala-Fmoc ~ 90 mm ~ 0.18 mmol/g PEG grafted on low cross-linked PS beads 86408
TentaGel S Trt-Arg(Pbf)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86442
TentaGel S Trt-Arg(Pmc)Fmoc ~ 90 mm ~ 0.18 mmol/g PEG grafted on low cross-linked PS beads 86411
TentaGel S Trt-Arg(Mtr)Fmoc ~ 90 mm ~ 0.18 mmol/g PEG grafted on low cross-linked PS beads 86409
TentaGel S Trt-Asn(Trt)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86412
TentaGel S Trt-Asp(t-Bu)Fmoc ~ 90 mm ~ 0.18 mmol/g PEG grafted on low cross-linked PS beads 86413
TentaGel S Trt-Cys(Acm)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86414
TentaGel S Trt-Cys(t-Bu)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86415
TentaGel S Trt-Cys(S-t-Bu)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86416
TentaGel S Trt-Cys(Trt)Fmoc ~ 90 mm ~ 0.18 mmol/g PEG grafted on low cross-linked PS beads 86417
TentaGel S Trt-Gln(Trt)Fmoc ~ 90 mm ~ 0.18 mmol/g PEG grafted on low cross-linked PS beads 86419
TentaGel S Trt-Glu(t-Bu)Fmoc ~ 90 mm ~ 0.18 mmol/g PEG grafted on low cross-linked PS beads 86421
TentaGel S Trt-Gly-Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86422
TentaGel S Trt-His(Trt)Fmoc ~ 90 mm ~ 0.18 mmol/g PEG grafted on low cross-linked PS beads 86423
TentaGel S Trt-Ile-Fmoc ~ 90 mm ~ 0.18 mmol/g PEG grafted on low cross-linked PS beads 86424
TentaGel S Trt-Leu-Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86425
TentaGel S Trt-Lys(Boc)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86426
TentaGel S Trt-Met-Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86427
TentaGel S Trt-Nle-Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86428
TentaGel S Trt-Orn(Boc)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86429
TentaGel S Trt-Phe-Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86431
TentaGel S Trt-Pro-Fmoc ~ 90 mm ~ 0.18 mmol/g PEG grafted on low cross-linked PS beads 86433
TentaGel S Trt-Ser(t-Bu)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86434
TentaGel S Trt-Thr(t-Bu)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86435
TentaGel S Trt-Trp(Boc)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86443
TentaGel S Trt-Trp-Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86436
TentaGel S Trt-Tyr(t-Bu)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86437
TentaGel S Trt-Val-Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86438
Preloaded TentaGel® Resins      
TentaGel S PHB-Ala-Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86367
TentaGel S PHB-Arg(Pbf)Fmoc ~ 90 mm ~ 0.22 mmol/g PEG grafted on low cross-linked PS beads 86381
TentaGel S PHB-Arg(Pmc)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86369
TentaGel S PHB-Asn(Trt)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86378
TentaGel S PHB-Asp(t-Bu)Fmoc ~ 90 mm ~ 0.22 mmol/g PEG grafted on low cross-linked PS beads 86379
TentaGel S PHB-Cys(Acm)Fmoc ~ 90 mm ~ 0.22 mmol/g PEG grafted on low cross-linked PS beads 86383
TentaGel S PHB-Cys(t-Bu)Fmoc ~ 90 mm ~ 0.22 mmol/g PEG grafted on low cross-linked PS beads 86384
TentaGel S PHB-Cys(S-t-Bu)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86385
TentaGel S PHB-Cys(Trt)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86386
TentaGel S PHB-Gln(Trt)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86388
TentaGel S PHB-Glu(t-Bu)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86389
TentaGel S PHB-Gly-Fmoc ~ 90 mm ~ 0.22 mmol/g PEG grafted on low cross-linked PS beads 86391
TentaGel S PHB-His(Trt)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86392
TentaGel S PHB-Ile-Fmoc ~ 90 mm ~ 0.18 mmol/g PEG grafted on low cross-linked PS beads 86393
TentaGel S PHB-Leu-Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86394
TentaGel S PHB-Lys(Boc)Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86395
TentaGel S PHB-Met-Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86396
TentaGel S PHB-Nle-Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86397
TentaGel S PHB-Orn(Boc)Fmoc ~ 90 mm ~ 0.18 mmol/g PEG grafted on low cross-linked PS beads 86398
TentaGel S PHB-Phe-Fmoc ~ 90 mm ~ 0.22 mmol/g PEG grafted on low cross-linked PS beads 86399
TentaGel S PHB-Pro-Fmoc ~ 90 mm ~ 0.20 mmol/g PEG grafted on low cross-linked PS beads 86401
TentaGel S PHB-Ser(t-Bu)Fmoc ~ 90 mm ~ 0.22 mmol/g PEG grafted on low cross-linked PS beads 86402
TentaGel S PHB-Thr(t-Bu)Fmoc ~ 90 mm ~ 0.22 mmol/g PEG grafted on low cross-linked PS beads 86403
TentaGel S PHB-Trp(Boc)Fmoc ~ 90 mm ~ 0.22 mmol/g PEG grafted on low cross-linked PS beads 86418
TentaGel S PHB-Trp-Fmoc ~ 90 mm ~ 0.22 mmol/g PEG grafted on low cross-linked PS beads 86404
TentaGel S PHB-Tyr(t-Bu)Fmoc ~ 90 mm ~ 0.22 mmol/g PEG grafted on low cross-linked PS beads 86405
TentaGel S PHB-Val-Fmoc ~ 90 mm ~ 0.22 mmol/g PEG grafted on low cross-linked PS beads 86406
Preloaded Trityl Resins      
H-Ala-2-ClTrt resin 75-150 mm; 100-200 ~ 0.8 mmol/g   80929
H-Arg(Pbf)-2-ClTrt resin 75-150 mm; 100-200 ~ 0.6 mmol/g   88619
H-Arg(Pmc)-2-ClTrt resin 75-150 mm; 100-200 ~ 0.7 mmol/g   72859
H-Asn(Trt)-2-ClTrt resin 75-150 mm; 100-200 ~ 0.6 mmol/g   94636
H-Asp(OtBu)-2-ClTrt resin 75-150 mm; 100-200 ~ 1.0 mmol/g   92368
H-Cys(Acm)-2-ClTrt resin 75-150 mm; 100-200 ~ 1.0 mmol/g   94399
H-Cys(Trt)-2-ClTrt resin 75-150 mm; 100-200 ~ 0.7 mmol/g   90254
H-Gln(Trt)-2-ClTrt resin 75-150 mm; 100-200 ~ 0.9 mmol/g   95256
H-Glu(OtBu)-2-ClTrt resin 75-150 mm; 100-200 ~ 1.0 mmol/g   82201
H-Gly-2-ClTrt resin 75-150 mm; 100-200 ~ 1.1 mmol/g   92681
H-His(Trt)-2-ClTrt resin 75-150 mm; 100-200 ~ 0.8 mmol/g   93216
H-Ile-2-ClTrt resin 75-150 mm; 100-200 ~ 1.0 mmol/g   82914
H-Leu-2-ClTrt resin 75-150 mm; 100-200 ~ 1.1 mmol/g   82938
H-Lys(Boc)-2-ClTrt resin 75-150 mm; 100-200 ~ 1.0 mmol/g   89861
H-Met-2-ClTrt resin 75-150 mm; 100-200 ~ 1.0 mmol/g   76215
H-Phe-2-ClTrt resin 75-150 mm; 100-200 ~ 0.9 mmol/g   73771
H-Pro-2-ClTrt resin 75-150 mm; 100-200 ~ 0.6 mmol/g   94077
H-Ser(tBu)-2-ClTrt resin 75-150 mm; 100-200 ~ 1.1 mmol/g   73571
H-Thr(tBu)-2-ClTrt resin 75-150 mm; 100-200 ~ 0.9 mmol/g   72574
H-Trp(Boc)-2-ClTrt resin 75-150 mm; 100-200 ~ 0.8 mmol/g   80539
H-Tyr(tBu)-2-ClTrt resin 75-150 mm; 100-200 ~ 1.0 mmol/g   71025
H-Val-2-ClTrt resin 75-150 mm; 100-200 ~ 1.0 mmol/g   86557
Preloaded Wang Resins      
Fmoc-Ala-Wang resin 75-150 mm; 100-200 0.4-0.8 mmol/g PS cross-linked with 1% DVB 47644
Fmoc-Arg(Pbf)-Wang resin 75-150 mm; 100-200 0.4-0.8 mmol/g PS cross-linked with 1% DVB 47362
Fmoc-Arg(Pmc)-Wang resin 75-150 mm; 100-200 0.4-0.6 mmol/g PS cross-linked with 1% DVB 47665
Fmoc-Asn(Trt)-Wang resin 75-150 mm; 100-200 0.4-0.8 mmol/g PS cross-linked with 1% DVB 47388
Fmoc-Asp(OtBu)-Wang resin 75-150 mm; 100-200 0.4-0.9 mmol/g PS cross-linked with 1% DVB 47646
Fmoc-Cys(Acm)-Wang resin 75-150 mm; 100-200 0.4-0.6 mmol/g PS cross-linked with 1% DVB 47613
Fmoc-Cys(StBu)-Wang resin 75-150 mm; 100-200 0.4-0.6 mmol/g PS cross-linked with 1% DVB 47651
Fmoc-Cys(Trt) Wang resin 38-75 mm; 200-400 ~ 0.7 mmol/g PS cross-linked with 1% DVB 18606
Fmoc-Gln(Trt)-Wang resin 75-150 mm; 100-200 0.4-0.8 mmol/g PS cross-linked with 1% DVB 47402
Fmoc-Glu(OtBu)-Wang resin 75-150 mm; 100-200 0.4-0.6 mmol/g PS cross-linked with 1% DVB 47658
Fmoc-Gly-Wang resin 75-150 mm; 100-200 0.4-0.8 mmol/g PS cross-linked with 1% DVB 47659
Fmoc-His(Trt)-Wang resin 38-75 mm; 200-400 ~ 0.7 mmol/g PS cross-linked with 1% DVB 42237
Fmoc-Ile-Wang resin 75-150 mm; 100-200 0.4-0.6 mmol/g PS cross-linked with 1% DVB 47661
Fmoc-Leu-Wang resin 75-150 mm; 100-200 0.4-0.9 mmol/g PS cross-linked with 1% DVB 47662
Fmoc-Lys(Boc)-Wang resin 75-150 mm; 100-200 0.4-0.6 mmol/g PS cross-linked with 1% DVB 47647
Fmoc-Met-Wang resin 75-150 mm; 100-200 0.4-0.8 mmol/g PS cross-linked with 1% DVB 47663
Fmoc-D-Met-Wang resin 75-150 mm; 100-200 ~ 0.8 mmol/g PS cross-linked with 1% DVB 47338
Fmoc-Phe-Wang resin 38-75 mm; 200-400 0.4-0.8 mmol/g PS cross-linked with 1% DVB 47666
Fmoc-D-Phe-Wang resin Capacity 0.6-1.0 mmol/g, 75-150 mm; 100-200 0.6-1.0 mmol/g PS cross-linked with 1% DVB 47379
Fmoc-Pro-Wang resin 75-150 mm; 100-200 0.4-0.8 mmol/g PS cross-linked with 1% DVB 47667
Fmoc-Ser(tBu)-Wang resin 75-150 mm; 100-200 0.4-0.8 mmol/g PS cross-linked with 1% DVB 47648
Fmoc-Ser(Trt)-Wang resin 75-150 mm; 100-200 0.4-0.8 mmol/g PS cross-linked with 1% DVB 47403
Fmoc-Thr(tBu)-Wang resin 75-150 mm; 100-200 0.4-0.6 mmol/g PS cross-linked with 1% DVB 47652
Fmoc-Trp(Boc) Wang resin 38-75 mm; 200-400 ~ 0.7 mmol/g PS cross-linked with 1% DVB 55606
Fmoc-Trp-Wang resin 38-75 mm; 200-400 0.4-0.6 mmol/g PS cross-linked with 1% DVB 47668
Fmoc-D-Trp-Wang resin 75-150 mm; 100-200 0.4-0.8 mmol/g PS cross-linked with 1% DVB 47404
Fmoc-Tyr(tBu)-Wang resin 75-150 mm; 100-200 0.4-0.8 mmol/g PS cross-linked with 1% DVB 47654
Fmoc-Val-Wang resin 38-75 mm; 200-400 ~ 0.6 mmol/g PS cross-linked with 1% DVB 47669
Fmoc-D-Val-Wang resin 75-150 mm; 100-200 0.4-0.8 mmol/g PS cross-linked with 1% DVB 47407
Trityl Resins Preloaded with Amino Alcohols      
L-Alaninol 2-chlorotrityl ether polymer bound 200-400 0.3-1 mmol/g PS crosslinked with 1 % DVB 94476
3-Amino-1-propanol 2-chlorotrityl ether polymer bound 200-400 0.3-1 mmol/g PS crosslinked with 1 % DVB 90846
4-Amino-1-butanol 2-chlorotrityl ether polymer bound, Capacity 0.3-1 mmol/g 200-400 0.3-1 mmol/g PS crosslinked with 1 % DVB 87052
Glycinol 2-chlorotrityl ether polymer bound 200-400 0.3-1 mmol/g PS crosslinked with 1 % DVB 87019
L-Isoleucinol 2-chlorotrityl ether polymer bound 200-400 0.3-1 mmol/g PS crosslinked with 1 % DVB 87023
L-Leucinol 2-chlorotrityl ether polymer bound particle size 200-400 mesh 200-400 0.3-1 mmol/g PS crosslinked with 1 % DVB 90263
L-Phenylalaninol 2-chlorotrityl ether polymer bound 200-400 0.3-1 mmol/g PS crosslinked with 1 % DVB 76068
L-Prolinol 2-chlorotrityl ether polymer bound 200-400 0.3-1 mmol/g PS crosslinked with 1 % DVB 77222
L-Valinol 2-chlorotrityl ether polymer bound 200-400 0.3-1 mmol/g PS crosslinked with 1 % DVB 73198
N-(3-Aminopropyl)tritylamine polymer bound 200-400 0.3-1 mmol/g PS crosslinked with 1 % DVB 74363
N-(6-Aminohexyl)tritylamine polymer bound Capacity 200-400 meq, 200-400 0.3-1 mmol/g PS crosslinked with 1 % DVB 74282
N-[2-(2-(2-Aminoethoxy)ethoxy)ethyl]-tritylamine polymer bound 200-400 0.3-1 mmol/g PS crosslinked with 1 % DVB 76338
Piperazine-trityl polymer bound 200-400 0.3-1 mmol/g PS crosslinked with 1 % DVB 73197
Homopiperazine-trityl polymer bound 200-400 0.3-1 mmol/g PS crosslinked with 1 % DVB 73375
Phoxime Resin       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
Phoxime resin Capacity ~0.8 mmol/g, ~ 0.8 mmol/g 100-300 PS crosslinked with 1 % DVB 17377
Phoxime resin       52,373-9
Resin Matrices       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
PEGA resin ~0.20 mmol/g ~ 0.20 mmol/g 300-500 mm poly[acryloyl-bis(aminopropyl)
polyethylene glycol]
89839
PEGA resin ~0.40 mmol/g ~ 0.40 mmol/g 150-300 mm poly[acryloyl-bis(aminopropyl)
polyethylene glycol]
76153
Poly(styrene-co-divinylbenzene) 99% styrene – 1% DVB   200-400 PS crosslinked with 1 % DVB 27820
Poly(styrene-co-divinylbenzene) 98.5% styrene – 1.5% DVB   150-400 "PS crosslinked with 1,5 % DVB" 27833
Poly(styrene-co-divinylbenzene) 98% styrene – 2% DVB   100-200 PS crosslinked with 2 % DVB 27832
Poly(styrene-co-divinylbenzene) 98% styrene – 2% DVB   200-400 PS crosslinked with 2 % DVB 27821
Poly(styrene-co-divinylbenzene) 97% styrene – 3% DVB   100-200 PS crosslinked with 3 % DVB 27831
Poly(styrene-co-divinylbenzene) 97% styrene – 3% DVB   200-400 PS crosslinked with 3 % DVB 27829
Poly(styrene-co-divinylbenzene) 96.5% styrene – 3.5% DVB   150-400 "PS crosslinked with 3,5 % DVB" 27828
Poly(styrene-co-divinylbenzene) 96% styrene – 4% DVB   200-400 PS crosslinked with 4 % DVB 27822
Poly(styrene-co-divinylbenzene) 94% styrene – 6% DVB   150-400 PS crosslinked with 6 % DVB 27823
Rink Acid Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
Rink acid resin Particle size 100-200 mesh 0.5-1.5 mmol/g 100-200   51,659-7
Rink acid resin Particle size 50-100 mesh 0.6-0.8 mmol/g 50-100   53,452-8
Rink acid-labil (hydroxymethyl)polystyrene Capacity ~1.1 mmol/g, resin ~ 1.1 mmol/g 200-400 PS cross-linked with 1% DVB 83893
Rink Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
HypoGel® 200 RAM ~ 0.6 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked PS beads 79028
Polystyrene AM-RAM Capacity 0.4-0.7 mmol/g, 0.4-0.7 mmol/g 160-200 mm PS cross-linked with 1% DVB 81554
Polystyrene A-RAM Capacity 0.5-0.8 mmol/g, 0.5-0.8 mmol/g 160-200 mm PS cross-linked with 1% DVB 81561
Polystyrene A-RAM Capacity 0.4-0.7 mmol/g, resin 0.4-0.7 mmol/g 500-560 mm PS cross-linked with 1% DVB 81560
Rink amide (aminomethyl)polystyrene Particle size 100-200 mesh 0.4-0.6 mmol/g 100-200   52,429-8
Rink amide (aminomethyl)polystyrene Capacity ~1.1 mmol/g, resin ~ 1.1 mmol/g 200-400 PS cross-linked with 1% DVB 83896
Rink amide (aminomethyl)polystyrene Capacity ~1.1 mmol/g, resin ~ 1.1 mmol/g 100-200 PS cross-linked with 1% DVB 83885
Rink amide (aminomethyl)polystyrene Capacity ~0.65 mmol/g, resin ~ 0.65 mmol/g 200-400 PS cross-linked with 1% DVB 83897
Rink amide 4-methylbenzhydrylamine polymer bound Capacity ~0.5 mmol/g, resin ~ 0.5 mmol/g 200-400 PS cross-linked with 1% DVB 83902
Rink amide 4-methylbenzhydrylamine polymer bound Capacity ~1.1 mmol/g, resin ~ 1.1 mmol/g 200-400 PS cross-linked with 1% DVB 83901
Rink amide 4-methylbenzhydrylamine polymer bound Capacity ~1.1 mmol/g, resin ~ 1.1 mmol/g 100-200 PS cross-linked with 1% DVB 83889
Rink amide 4-methylbenzhydrylamine polymer bound with Norleucine internal standard Capacity 0.3-0.7 mmol/g, resin 0.3-0.7 mmol/g 200-400 PS cross-linked with 1% DVB 83891
Rink amide 4-methylbenzhydrylamine polymer bound Particle size 50-100 mesh 0.3-0.8 mmol/g 50-100   53,394-7
Rink amide benzhydrylamine polymer bound Capacity 0.7-1.2 mmol/g, 0.7-1.2 mmol/g 200-400 PS cross-linked with 1% DVB 83898
Rink amide benzhydrylamine polymer bound Capacity ~0.5 mmol/g, resin ~ 0.5 mmol/g 200-400 PS cross-linked with 1% DVB 83900
Rink amide benzhydrylamine polymer bound Capacity ~1.1 mmol/g, resin ~ 1.1 mmol/g 100-200 PS cross-linked with 1% DVB 83887
Rink acid-labil (hydroxymethyl)polystyrene 0.8-1.0 mmmol/g 70-90   53393-5
Rink amide (aminomethyl)polystyrene with Norleucine internal standard Capacity ~1.1 mmol/g, resin ~ 1.1 mmol/g 200-400 PS cross-linked with 1% DVB 83886
Rink amide benzhydrylamine polymer bound with Norleucine internal standard Capacity ~0.5 mmol/g, resin ~ 0.5 mmol/g 200-400 PS cross-linked with 1% DVB 83888
Rink amine resin Particle size 100-200 mesh 0.5-1.5 mmol/g 100-200   49,729-0
TentaGel HL-RAM Capacity ~0.35 mmol/g, ~ 0.35 mmol/g ~ 75 mm PEG grafted on low cross-linked PS beads 86342
TentaGel MB-RAM Capacity ~0.35 mmol/g, ~ 0.35 mmol/g 140-170 mm PEG grafted on low cross-linked PS beads 86374
TentaGel S RAM Capacity ~0.24 mmol/g, ~ 0.24 mmol/g ~ 90 mm PEG grafted on low cross-linked PS beads 86407
Safety Catch Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
3-[Hydroxy(2-phenyl-1,3-dithian-2-yl)methyl]phenoxymethyl-polystyrene Capacity ~0.6 mmol/g, ~ 0.6 mmol/g 100-300 PS crosslinked with 1 % DVB 56146
N-(4-Sulfamoylbenzoyl)aminomethyl-polystyrene Capacity (S) ~0.9 mmol/g, resin ~ 0.9 mmol/g 200-400 PS cross-linked with 1% DVB 86052
TentaGel® Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
TentaGel HL-PHB Capacity ~0.35 mmol/g, ~ 0.35 mmol/g ~ 75 mm PEG grafted on low cross-linked PS beads 86341
TentaGel MB-PHB Capacity ~0.40 mmol/g, ~ 0.40 mmol/g 140-170 mm PEG grafted on low cross-linked PS beads 86373
TentaGel S PHB Capacity ~0.24 mmol/g, ~ 0.24 mmol/g ~ 90 mm PEG grafted on low cross-linked PS beads 86366
Thiol-Functionalized Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
HypoGel® 200 S-Trt ~ 0.7 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB) PS beads 77044
Polystyrene AM-S-Trityl Capacity 0.6-0.8 mmol/g, 0.6-0.8 mmol/g 160-200 mm PS crosslinked with 1 % DVB 81557
Polystyrene AM-S-Trityl Capacity 0.8-1.0 mmol/g, 0.8-1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 81555
Polystyrene AM-S-Trityl Capacity 0.6-0.8 mmol/g, 0.6-0.8 mmol/g 200-400 PS crosslinked with 1 % DVB 81556
Polystyrene A-S-Trityl Capacity 0.6-0.8 mmol/g, 0.6-0.8 mmol/g 160-200 mm PS crosslinked with 1 % DVB 81562
Polystyrene A-S-Trityl Capacity 0.6-0.8 mmol/g, 0.6-0.8 mmol/g 500-560 mm PS crosslinked with 1 % DVB 81563
TentaGel HL-S-Trityl Capacity ~0.40 mmol/g, ~ 0.40 mmol/g ~ 110 mm PEG grafted on low cross-linked PS beads 86343
TentaGel MB-S-Trityl Capacity ~0.4 mmol/g, ~ 0.4 mmol/g 140-170 mm PEG grafted on low cross-linked PS beads 86376
TentaGel S-S-Trityl Capacity ~0.25 mmol/g, ~ 0.25 mmol/g ~ 90 mm PEG grafted on low cross-linked PS beads 86387
Triazine-Based Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
Aniline, polymer-bound 0.5-1.3 mmol/g 100-200 PS cross-linked with 1% DVB 564761
N-Benzylamine, polymer-bound 1.0-2.0 mmol/g 100-200 PS cross-linked with 1% DVB 538310
Piperazine, polymer-bound ca. 1.5 mmol/g 200-400 PS cross-linked with 2 % DVB 526290
Trityl Amines       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
2-Chlorotrityl amine, polymer-bound 1.3-1.8 mmol/g 100-200 PS cross-linked with 1% DVB 536113
2-Chlorotritylhydrazine polymer bound 0.4-0.6 mmol/g 200-400 PS cross-linked with 1% DVB 530948
Aminotrityl, polymer-bound 1.3-1.7 mmol/g 100-200 PS cross-linked with 1% DVB 542970
Hydrazine trityl, polymer-bound 1.2-1.7 mmol/g 100-200 PS cross-linked with 1% DVB 542997
1,2-Diaminoethane trityl, polymer-bound 0.5-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 543004
1,3-Diaminopropane trityl, polymer-bound 0.5-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 543012
1,4-Diaminobutane trityl, polymer-bound 0.3-1.2 mmol/g 100-200 PS cross-linked with 1% DVB 564273
1,6-Diaminohexane trityl resin 0.3-1.2 mmol/g 100-200 PS cross-linked with 1% DVB 564303
2-Chlorotrityl 2-hydroxyethylamine, polymer-bound 1.22 mmol/g 100-200 PS cross-linked with 1% DVB 543462
2-Chlorotrityl 3-amino-1-propanol, polymer-bound 0.7-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 543888
2-Chlorotrityl 3-hydroxypropylamine, polymer-bound 0.7-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 543489
2-Chlorotrityl 4-aminomethylbenzoic acid, polymer-bound 0.7-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 543799
2-Chlorotrityl 4-cyanobenzoic acid, polymer-bound 0.7-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 543802
2-Chlorotrityl alcohol, polymer-bound 1.2-1.8 mmol/g 200-400 PS cross-linked with 1% DVB 529389
2-Chlorotrityl chloride polymer bound ~ 1.9 mmol/g 100-300 PS cross-linked with 1% DVB 26568
2-Chlorotrityl chloride polymer bound 1.0-1.6 mmol/g 100-200 PS crosslinked with 1 % DVB 73311
2-Chlorotrityl chloride polymer bound 1.0-1.5 mmol/g 200-400 PS cross-linked with 1% DVB 532290
2-Chlorotrityl chloride polymer bound 1.0-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 532304
2-Chlorotrityl cysteamine, polymer-bound 0.4-0.8 mmol/g 200-400 PS cross-linked with 1% DVB 530107
2-Chlorotrityl ethylamine, polymer-bound 0.3-1.2 mmol/g 100-200 PS cross-linked with 1% DVB 564257
2-Chlorotrityl ethyleneglycol, polymer-bound 0.7-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 544671
2-Chlorotrityl glycinol, polymer-bound 0.4-0.8 mmol/g 200-400 PS cross-linked with 1% DVB 530298
2-Chlorotrityl Knorr resin 0.3-1.2 mmol/g 100-200 PS cross-linked with 1% DVB 549401
2-Chlorotrityl-N-Fmoc-hydroxylamine, polymer-bound 0.6-1.0 mmol/g 100-200 PS cross-linked with 1% DVB 544434
2-Chlorotrityl proline polymer-bound 0.4-0.8 mmol/g 200-400 PS cross-linked with 1% DVB 529397
2-Chlorotrityl thiol, polymer-bound 0.3-1.2 mmol/g 100-200 PS cross-linked with 1% DVB 564354
2-Chlorotritylhydrazine polymer bound ~ 0.35 mmol/g 100-200 PS cross-linked with 1% DVB 17753
4-Methoxytrityl alcohol, polymer-bound 1.5-2.5 mmol/g 100-200 PS cross-linked with 1% DVB 564230
4-Methoxytrityl chloride polymer bound 1.0-1.6 mmol/g 100-200 PS crosslinked with 1 % DVB 94679
4-Methoxytrityl chloride polymer bound ~ 0.7 mmol/g Cl resin 200-400 PS cross-linked with 1% DVB 65374
4-Methoxytrityl chloride polymer bound 1.5-2.5 mmol/g 100-200 PS cross-linked with 1% DVB 564249
4-Methyltrityl alcohol, polymer-bound 1.5-2.5 mmol/g 100-200 PS cross-linked with 1% DVB 564222
4-Methyltrityl chloride polymer bound ~ 0.7 mmol/g Cl resin 200-400 PS cross-linked with 1% DVB 69516
4-Methyltrityl chloride polymer bound 1.3-1.7 mmol/g 100-200 PS cross-linked with 1% DVB 542989
Cysteamine 2-chlorotrityl thioether polymer bound 0.3-1 mmol/g 200-400 PS crosslinked with 1 % DVB 74399
1,10-Dihydroxydecane trityl resin 0.3-1.2 mmol/g 100-200 PS cross-linked with 1% DVB 564346
Ethylamine trityl, polymer-bound 0.3-1.2 mmol/g 100-200 PS cross-linked with 1% DVB 564265
O-(2-Chlorotrityl)-N-Fmoc-hydroxylamine polymer bound Capacity ~1.1 mmol/g, (N) ~ 1.1 mmol/g 100-200 PS cross-linked with 1% DVB 17757
Thiol 4-methoxy trityl, polymer-bound 0.3-1.2 mmol/g 100-200 PS cross-linked with 1% DVB 564362
Trityl alcohol, polymer-bound 1.5-2.5 mmol/g 100-200 PS cross-linked with 1% DVB 564214
Trityl chloride polymer bound Capacity (Cl) ~1.1 mmol/g, resin ~ 1.1 mmol/g 100-200 PS cross-linked with 1% DVB 93003
Trityl chloride polymer bound Capacity ~1.7 mmol/g, ~ 1.7 mmol/g 100-200 PS cross-linked with 1% DVB 93005
Trityl chloride polymer bound Capacity 1.0-2.0 mmol/g, 1.0-2.0 mmol/g 200-400 PS crosslinked with 1 % DVB 93118
Trityl chloride polymer bound 1 % cross-linked Pore Size 100-200 mesh 1.0-1.8 mmol/g 100-200 PS cross-linked with 1% DVB 533483
Trityl chloride polymer bound 1 % cross-linked Pore Size 200-400 mesh 1.0-1.8 mmol/g 200-400 PS cross-linked with 1% DVB 533491
Wang       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
4-(Benzyloxy)benzyl bromide polymer bound Capacity ~0.7 mmol/g, ~ 0.7 mmol/g 75-150 PS crosslinked with DVB 17333
4-(Benzyloxy)benzyl chloride polymer bound Capacity (Cl) ~1.1 mmol/g, resin (T) ~ 1.1 mmol/g 200-400 PS crosslinked with 1 % DVB 02377
Wang Derivatives       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
Imidazole carboxylate-Wang resin Capacity ~1.0 mmol/g, ~ 1.0 mmol/g 100-300 PS crosslinked with 1 % DVB 17017
[4-(Trichloroacetimidoyloxymethyl)-phenoxymethyl]polystyrene Capacity ~0.9 mmol/g, ~ 0.9 mmol/g 100-300 PS crosslinked with 1 % DVB 17335
        back to top