| Sigma-Aldrich 2-Propanol Specifications Comparison |
| |
CHROMASOLV LC-MS |
CHROMASOLV Plus |
CHROMASOLV for HPLC |
Anhydrous |
ACS Spectro- photometric |
ACS |
ReagentPlus |
| Purity |
≥99.9% |
≥99.9% |
≥99.9% |
99.5% |
≥99.5% |
≥99.5% |
≥99% |
| UV Absorption |
| λ: 210 nm |
|
|
≤0.70 |
|
≤1.00 |
|
|
| λ: 220 nm |
≤0.30 |
|
≤0.30 |
|
≤0.40 |
|
|
| λ: 230 nm |
≤0.12 |
|
≤0.13 |
|
≤0.20 |
|
|
| λ: 245 nm |
|
≤1.00 |
|
|
≤0.08 |
|
|
| λ: 250 nm |
|
≤0.30 |
|
| λ: 260 nm |
≤0.0088 |
|
≤0.01 |
|
≤0.04 |
|
|
| λ: 275 nm |
|
≤0.005 |
|
|
≤0.03 |
|
|
| λ: 300 nm |
|
≤0.02 |
|
|
| λ: 330 nm |
|
≤0.01 |
|
|
| λ: 400 nm |
|
≤0.005% |
|
|
≤0.01 |
|
|
| Total Impurities |
| Water |
≤0.05% |
≤0.02% |
≤0.05% |
≤0.003% |
≤0.20% |
≤0.20% |
|
| |
|
|
≤0.005% (100mL) |
|
|
|
| Evaporation Residue |
≤0.0005% |
≤0.0001% |
≤0.0005% |
≤0.0003% |
≤0.001% |
≤0.001% |
|
| Halogenated Residue |
|
≤10 ng/L (as Heptachlor epoxide) |
|
| Fluorescence 254 nm |
≤1 ppb |
|
| Fluorescence 365 nm |
≤1 ppb |
≤0.5 ppb |
|
| LC-MS suitability |
passes test |
|
| Proprionaldehyde |
|
≤0.002% |
≤0.002% |
|
| Acetone |
|
≤0.002% |
≤0.002% |
|
| Titratable acid |
≤0.001% free acid (as C6H5COOH) |
|
≤0.001% free acid (as C6H5COOH |
|
≤0.0001 meq/g |
≤0.0001 meq/g |
|
| Titratable base |
|
≤0.0001 meq/g |
≤0.0001 meq/g |
|
| Silver |
0.1 ppm |
|
| Aluminum |
0.5 ppm |
|
| Barium |
0.1 ppm |
|
| Calcium |
0.1 ppm |
|
| Cadmium |
0.05 ppm |
|
| Cobalt |
0.02 ppm |
|
| Chromium |
0.02 ppm |
|
| Copper |
0.01 ppm |
|
| Iron |
0.1 ppm |
|
| Potassium |
0.1 ppm |
|
| Magnesium |
0.1 ppm |
|
| Manganese |
0.01 ppm |
|
| Sodium |
0.1 ppm |
|
| Nickel |
0.02 ppm |
|
| Lead |
0.02 ppm |
|
| Tin |
0.1 ppm |
|
| Zinc |
0.1 ppm |
|
| Packaging Options |
| |
CHROMASOLV LC-MS |
CHROMASOLV Plus |
CHROMASOLV for HPLC |
Anhydrous |
ACS Spectro- photometric |
ACS |
ReagentPlus |
| Glass Bottle |
34965 |
650447 |
34863 |
|
154970 |
190764 |
109827 |
| PVC Coated Bottle |
|
675431 |
439207 |
|
|
437522 |
|
| Sure/Seal™ |
|
|
|
278475 |
|
|
320471 |
| Aluminum Bottle |
|
673773 |
|
| VerSA-Flow™ |
|
|
439207 |
|
| NOWPAK™ |
|
|
439207 |
|
| Pure-Pac™ |
|
|
34863 |
|
| Carbon Steel |
|
190764 |
109827 |
| Drum |
|
|
439207 |
|
|
190764 |
|
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!