|
Whether you need HPLC acetone, ACS acetone or another Sigma-Aldrich grade, we stock the right product for your application. As a leading supplier of high-purity, research grade solvents, we have the acetone to meet your exact needs.
Select a solvent
| Acetone Physical Properties |
| Synonym |
2-Propanone |
Beilstein Registry |
635680 |
| Molecular Formula |
(CH3)COCH3 |
EG/EC Number |
2006622 |
| Molecular Weight |
58.08 |
MDL Number |
MFCD00008765 |
| CAS Number |
67-64-1 |
Vapor Pressure |
184 mm Hg (20°C) |
| Density |
0.791 g/mL at 25ºC |
Dielectric Constant |
21.01 at 20ºC |
| Boiling Point |
56ºC |
Dipole Moment |
2.880 |
| Melting Point |
- 94ºC |
UV cutoff |
330 nm |
| Flash Point |
-17.2ºC |
Refractive Index |
1.359 at 20ºC |
| Viscosity |
0.306 at 25ºC |
Vapor Density |
2 (vs. air) |
| RIDADR |
UN 1090 3/PG 2 |
Risk Statements |
11-36-66-67 |
| Hazard Codes |
F,Xi |
Safety Statements |
9-16-26 |
|
| Acetone Miscibility / Immiscibility Table |
|