| Sigma-Aldrich Acetone Specifications Comparison |
| |
CHROMASOLV Plus |
CHROMASOLV for HPLC |
ACS Spectro- photometric |
ACS |
ReagentPlus |
| Purity |
≥99.9% |
≥99.9% |
≥99.5% |
≥99.5% |
≥99% |
| UV Absorption |
| λ: 330 nm |
≤1.00 |
≤1.00 |
≤1.00 |
|
|
| λ: 340 nm |
≤0.06 |
≤0.10 |
≤0.10 |
|
|
| λ: 350 nm |
≤0.01 |
≤0.02 |
≤0.02 |
|
|
| λ: 375 nm |
≤0.005 |
|
| λ: 400 nm |
≤0.005 |
≤0.01 |
≤0.01 |
|
|
| Total Impurities |
| Water |
≤0.5% |
≤0.5% |
≤0.5% |
≤0.5% |
|
| Evaporation Residue |
≤0.0003% |
≤0.0002% |
≤0.001% |
≤0.001% |
|
| Halogenated Residue |
≤10 ng/L (as Heptachlor epoxide) |
|
| Fluorescence 365 nm |
≤1.0 ppb |
|
| Titratable acid |
|
|
0.0003 meq/g |
0.0003 meq/g |
|
| Titratable base |
|
|
0.0006 meq/g |
0.0006 meq/g |
|
| Formaldehyde |
|
|
≤0.002% |
≤0.002% |
|
| Isopropanol |
|
|
≤0.05% |
≤0.05% |
|
| Methanol |
|
|
≤0.05% |
≤0.05% |
|
| Packaging Options |
| |
CHROMASOLV Plus |
CHROMASOLV for HPLC |
ACS Spectro- photometric |
ACS |
ReagentPlus |
| Glass Bottle |
650501 |
270725 |
154598 |
179124 |
179973 |
| PVC Coated Bottle |
|
439126 |
|
320110 |
|
| Aluminum Bottle |
|
|
|
673781 |
|
| VerSA-Flow™ |
|
439126 |
|
|
|
| NOWPAK™ |
|
439126 |
|
|
|
| Pure-Pac™ |
|
270725 |
|
|
|
| Carbon Steel |
|
|
|
179124 |
179973 |
| Drum |
|
439126 |
|
673781 |
|
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!