Sigma-Aldrich
Chemistry > Solvent Center > Acetonitrile
Solvent Center
Acetonitrile
Whether you need HPLC acetonitrile, BioTech acetonitrile, Anhydrous acetonitrile or another Sigma-Aldrich grade, we stock the right product for your application. As a leading supplier of high-purity, research grade solvents, we have the acetonitrile to meet your exact needs
  •  Acetone 
  •  Acetonitrile 
  •  Chloroform 
  • Dichloromethane 
  • Dimethyl sulfoxide
  • Ethyl Acetate
  •  Hexane 
  •  Methanol 
  •  2-Propanol 
  • Tetrahydrofuran
  • Toluene
  •  Water 
Select a solvent
View and Compare Acetonitrile Products
Sigma-Aldrich Acetonitrile
Synonym Methyl cyanide Beilstein Registry 741857
Molecular Formula CH3CN EG/EC Number 2008352
Molecular Weight 41.05 MDL Number MFCD00001878
CAS Number 75-05-8 Vapor Pressure 72.8 mm Hg (20°C)
Density 0.786 g/mL at 25ºC Dielectric Constant 36.64 at 20ºC
Boiling Point 81-82ºC Dipole Moment 3.924
Melting Point -48ºC UV cutoff 190 nm
Flash Point 5.5ºC Refractive Index 1.344 at 20ºC
Viscosity 0.369 at 25ºC Vapor Density 1.41 (vs. air)
RIDADR UN 1648 3/PG 2 Risk Statements 11-20/21/22-36
Hazard Codes F,Xn Safety Statements 16-36/37
image0011  image003
Acetonitrile Miscibility / Immiscibility Table
acetonitrile_miscibility
Back to Solvent Center