Solvent Center
Anhydrous Solvents
Sigma-Aldrich's offers a full range of high-purity solvents with extremely low water levels specifically manufactured for moisture sensitive Organic and BioTech applications.
Anhydrous Solvents
BioTech Solvents
Sigma-Aldrich Anhydrous & BioTech Solvents Table
Anhydrous Solvents
  Use our anhydrous solvent supplier cross-reference guide to compare grades and packaging options with Sigma-Aldrich quality products. (180 Kb PDF)
Complete list of Anhydrous grade solvents
Water content ranges from typically 10 to 30 ppm and do not require additional drying in your lab due to our low residue levels.
Ideal use in applications requiring extremely low levels of water such as:
Chemical Synthesis
High Throughput Screening
Oligonucleotide Synthesis
Organometallic
Sigma-Aldrich Anhydrous solvents are available in Sure/Seal Bottles and Pure-Pac containers.
Anhydrous 2-Methyltetrahydrofuran - A versatile alternative to Tetrahydrofuran and Dichloromethane
Back to Top
BioTech Solvents
Complete list of BioTech grade solvents
The BioTech line of solvents is characterized by not only low water content, but also minimal residue and clean UV spectra.
Purified for biotechnology applications such as:
Oligonucleotide Synthesis
Sigma-Aldrich BioTech solvents are available in Sure/Seal Bottles and Pure-Pac containers.
Back to Top
Sigma-Aldrich Anhydrous & BioTech Solvents Table
Anhydrous Biotech
Acetonitrile  
Purity
Water
100 mL
Residue
27,100-4
99.8%
< 10 ppm
< 50 ppm
< 0.0005%
  49,444-5
≥99.93%
< 10 ppm
<0.0002%
UV nm
190
210
220
235-400
<0.008 meq/g
<0.0006 meq/g
Max Abs.
1.000
0.030
0.010
0.005
Titr. Acid
Titr. Base
Buytl acetate  
Purity
Water
Residue
28,772-5
≥99%
< 50 ppm
< 0.0005%
  49,616-2
99.7%
< 10 ppm
<0.0005%
UV nm
<0.0016 meq/g
Max Abs.
Titr. Acid
Carbon tetrachloride  
Purity
Water
100 mL
Residue
28,911-6
≥99.5%
< 20 ppm
<50 ppm
< 0.0003%
1-Chlorobutane  
Purity
Water
100 mL
Residue
41,425-5
99.5%
< 30 ppm
< 50 ppm
< 0.0005%
  49,617-0
≥99.5%
< 10 ppm
<0.0003%
UV nm
<0.0016 meq/g
Max Abs.
Titr. Acid
Chloroform
0.5-1.0% ethanol as stabilizer
Purity
Water
100 mL
Residue
40,176-5
99.7%
< 10 ppm
< 50 ppm
< 0.0005%
  49,618-9
≥99.9%
< 10 ppm
<0.0003%
Dichloromethane
50-150 ppm hydrocarbon as stabilizer
Purity
Water
100 mL
Residue
27,099-7
99.0%
< 10 ppm
< 50 ppm
< 0.0005%
  49,445-3
99.9%
< 20 ppm
<0.0003%
UV nm
235
240
250
260
340-400
Max Abs.
1.000
0.200
0.050
0.020
0.010
N,N-Diisopropylethylamine  
Purity
Water
Residue
27,101-2
99.8%
< 50 ppm
< 0.0001%
  49,621-9
99.5%
< 20 ppm
N,N-Dimethylformamide  
Purity
Water
Residue
22,705-6
99.8%
< 50 ppm
< 0.0005%
  49,448-8
≥99.9%
< 50 ppm
<0.0005%
UV nm
275
295
310
340-400
<0.0005 meq/g
<0.0003 meq/g
Max Abs.
0.30
0.10
0.05
0.07
Titr. Acid
Titr. Base
Dimethyl sulfoxide  
Purity
Water
Residue
27,685-5
≥99.9%
< 50 ppm
< 0.0005%
  49,442-9
99.8%
< 50 ppm
<0.0005%
UV nm
270
280
295
350-400
Max Abs.
1.00
0.30
0.10
0.01
1,4-Dioxane  
Purity
Water
100 mL
Residue
29,630-9
99.8%
< 30 ppm
< 50 ppm
< 0.0003%
Ether
1 ppm BHT as stabilzer
Purity
Water
100 mL
Residue
29,608-2
≥ 99.7%
< 30 ppm
< 50 ppm
< 0.0003%
Ethyl actate  
Purity
Water
Residue
27,098-9
99.8%
< 50 ppm
< 0.0005%
  49,451-8
≥99.8%
< 10 ppm
<0.0003%
UV nm
254
263
275-400
<0.0009 meq/g
Max Abs.
1.00
0.05
0.01
Titr. Acid
Heptane  
Purity
Water
100 mL
Residue
24,665-4
99%
< 10 ppm
< 50 ppm
< 0.0005%
  49,452-6
≥99%
< 50 ppm
<0.0002%
UV nm
197
210
220
230
260-400
Max Abs.
1.00
0.40
0.15
0.05
0.01
Methanol  
Purity
Water
100 mL
Residue
32,241-5
99.8%
< 20 ppm
< 50 ppm
< 0.0003%
  49,443-7
99.93%
< 20 ppm
<0.0003%
UV nm
203
220
230
240
260-400
<0.0003 meq/g
<0.0002 meq/g
Max Abs.
1.00
0.60
0.15
0.05
0.01
Titr. Acid
Titr. Base
3-Methyl-1-butanol  
Purity
Water
100 mL
Residue
30,943-5
≥99%
< 30 ppm
< 50 ppm
< 0.0005%
  49,620-0
≥99%
< 50 ppm
<0.0005%
UV nm
<0.002 meq/g
<0.1%
Max Abs.
Titr. Acid
Carbonyl
1-Methyl-2-pyrrolidinone  
Purity
Water
32,863-4
99.5%
< 50 ppm
  49,449-6
≥99.5%
< 50 ppm
UV nm
285
300
325
350-400
<0.01% meq/g
Max Abs.
1.00
0.50
0.10
0.01
Free amines
1-Propanol  
Purity
Water
Residue
27,954-4
99.7%
< 50 ppm
< 0.0003%
  49,619-7
99.7%
<0.0003%
UV nm
<0.0004 meq/g
Max Abs.
Titr. Acid
Pyridine  
Purity
Water
100 mL
Residue
27,097-0
99.8%
< 30 ppm
< 50 ppm
< 0.0005%
  49,441-0
≥99.9%
< 30 ppm
<0.0003%
UV nm
<0.001%
<0.0005%
<0.002%
<0.0001%
Max Abs.
Cl
Cu
NH3
SO4
Tetrahydrofuran
Inhibitor-free
Purity
Water
100 mL
Residue
401757
99.9%
< 20 ppm
< 50 ppm
< 0.0005%
  49,446-1
≥99.9%
< 50 ppm
<0.0005%
UV nm
<0.005%
Max Abs.
Peroxides
Tetrahydrofuran
250 ppm BHT as stabilizer
Purity
Water
100 mL
186562
99.9%
< 20 ppm
< 50 ppm
Toluene  
Purity
Water
100 mL
Residue
24,451-1
99.8%
< 10 ppm
< 50 ppm
<0.0005%
Visit the complete list of Sigma-Aldrich Anydrous Grade Solvents and Biotech Grade Solvents.
Back to Top
Sigma-Aldrich offers a full range of high-purity solvents with extremely low water levels specifically manufactured for moisture sensitive Organic and Biotech applications.
    Anhydrous Solvents
    Biotech Solvents
    Sigma-Aldrich Anhydrous & Biotech Solvents Table
Anhydrous Solvents
Anhydrous Solvent Supplier Cross-Reference Guide   Use our anhydrous solvent supplier cross-reference guide to compare grades and packaging options with Sigma-Aldrich quality products. (180 Kb PDF)
Complete list of Anhydrous grade solvents
Sigma-Aldrich Anhydrous solvents are available in Sure/Seal Bottles and Pure-Pac containers.
Anhydrous 2-Methyltetrahydrofuran - A versatile alternative to Tetrahydrofuran and Dichloromethane
back to top
Biotech Solvents
Complete list of Biotech grade solvents
The Biotech line of solvents is characterized by not only low water content, but also minimal residue and clean UV spectra.
Purified for biotechnology applications such as:
Sigma-Aldrich Biotech solvents are available in Sure/Seal Bottles and Pure-Pac containers.
back to top
Sigma-Aldrich Anhydrous & Biotech Solvents Table
  Anhydrous Biotech
  Acetonitrile Purity
Water
100 mL
Residue
271004
99.8%
< 10 ppm
< 50 ppm
< 0.0005%
494445
≥99.93%
< 10 ppm
<0.0002%
UV nm
190
210
220
235-400
<0.008 meq/g
<0.0006 meq/g
Max Abs.
1.000
0.030
0.010
0.005
Titr. Acid
Titr. Base
  Buytl acetate Purity
Water
Residue
287725
≥99%
< 50 ppm
< 0.0005%
496162
99.7%
< 10 ppm
<0.0005%
UV nm
<0.0016 meq/g
Max Abs.
Titr. Acid
  Carbon tetrachloride Purity
Water
100 mL
Residue
289116
≥99.5%
< 20 ppm
<50 ppm
< 0.0003%
  1-Chlorobutane Purity
Water
100 mL
Residue
414255
99.5%
< 30 ppm
< 50 ppm
< 0.0005%
496170
≥99.5%
< 10 ppm
<0.0003%
UV nm
<0.0016 meq/g
Max Abs.
Titr. Acid
  Chloroform
  0.5-1.0% ethanol as stabilizer
Purity
Water
100 mL
Residue
401765
99.7%
< 10 ppm
< 50 ppm
< 0.0005%
496189
≥99.9%
< 10 ppm
<0.0003%
  Dichloromethane
50-150 ppm hydrocarbon as stabilizer
 Purity
Water
100 mL
Residue
270997
99.0%
< 10 ppm
< 50 ppm
< 0.0005%
494453
99.9%
< 20 ppm
<0.0003%
UV nm
235
240
250
260
340-400
Max Abs.
1.000
0.200
0.050
0.020
0.010
  N,N-Diisopropylethylamine Purity
Water
Residue
271012
99.8%
< 50 ppm
< 0.0001%
496219
99.5%
< 20 ppm
  N,N-Dimethylformamide Purity
Water
Residue
227056
99.8%
< 50 ppm
< 0.0005%
494488
≥99.9%
< 50 ppm
<0.0005%
UV nm
275
295
310
340-400
<0.0005 meq/g
<0.0003 meq/g
Max Abs.
0.30
0.10
0.05
0.07
Titr. Acid
Titr. Base
  Dimethyl sulfoxide Purity
Water
Residue
276855
≥99.9%
< 50 ppm
< 0.0005%
494429
99.8%
< 50 ppm
<0.0005%
UV nm
270
280
295
350-400
Max Abs.
1.00
0.30
0.10
0.01
  1,4-Dioxane Purity
Water
100 mL
Residue
296309
99.8%
< 30 ppm
< 50 ppm
< 0.0003%
  Ether
 1 ppm BHT as stabilzer
Purity
Water
100 mL
Residue
296082
≥ 99.7%
< 30 ppm
< 50 ppm
< 0.0003%
  Ethyl actate Purity
Water
Residue
270989
99.8%
< 50 ppm
< 0.0005%
494518
≥99.8%
< 10 ppm
<0.0003%
UV nm
254
263
275-400
<0.0009 meq/g
Max Abs.
1.00
0.05
0.01
Titr. Acid
  Heptane Purity
Water
100 mL
Residue
246654
99%
< 10 ppm
< 50 ppm
< 0.0005%
494526
≥99%
< 50 ppm
<0.0002%
UV nm
197
210
220
230
260-400
Max Abs.
1.00
0.40
0.15
0.05
0.01
  Methanol Purity
Water
100 mL
Residue
322415
99.8%
< 20 ppm
< 50 ppm
< 0.0003%
494437
99.93%
< 20 ppm
<0.0003%
UV nm
203
220
230
240
260-400
<0.0003 meq/g
<0.0002 meq/g
Max Abs.
1.00
0.60
0.15
0.05
0.01
Titr. Acid
Titr. Base
  3-Methyl-1-butanol  Purity
Water
100 mL
Residue
309435
≥99%
< 30 ppm
< 50 ppm
< 0.0005%
496200
≥99%
< 50 ppm
<0.0005%
UV nm
<0.002 meq/g
<0.1%
Max Abs.
Titr. Acid
Carbonyl
  1-Methyl-2-pyrrolidinone  Purity
Water
328634
99.5%
< 50 ppm
494496
≥99.5%
< 50 ppm
UV nm
285
300
325
350-400
<0.01% meq/g
Max Abs.
1.00
0.50
0.10
0.01
Free amines
  1-Propanol  Purity
Water
Residue
279544
99.7%
< 50 ppm
< 0.0003%
496197
99.7%
<0.0003%
UV nm
<0.0004 meq/g
Max Abs.
Titr. Acid
  Pyridine Purity
Water
100 mL
Residue
270970
99.8%
< 30 ppm
< 50 ppm
< 0.0005%
494410
≥99.9%
< 30 ppm
<0.0003%
UV nm
<0.001%
<0.0005%
<0.002%
<0.0001%
Max Abs.
Cl
Cu
NH3
SO4
  Tetrahydrofuran
 Inhibitor-free
Purity
Water
100 mL
Residue
401757
99.9%
< 20 ppm
< 50 ppm
< 0.0005%
494461
≥99.9%
< 50 ppm
<0.0005%
UV nm
<0.005%
Max Abs.
Peroxides
  Tetrahydrofuran
250 ppm BHT as stabilizer
Purity
Water
100 mL
186562
99.9%
< 20 ppm
< 50 ppm
  Toluene Purity
Water
100 mL
Residue
244511
99.8%
< 10 ppm
< 50 ppm
<0.0005%
Visit the complete list of Sigma-Aldrich Anydrous Grade Solvents and Biotech Grade Solvents
back to top