Sigma-Aldrich
Aldrich - Chemistry logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Solvent Center > Bottles & Cans
Solvents
Bottles & Cans
Sigma-Aldrich offers a variety of packaging options for your application. Whether you need your solvent packaging in glass bottle, PVC coated bottle, Sure/Seal™ bottle or aluminum can, we package and solvent dispensing options to meet your needs.
Amber Glass Bottles
PVC Coated Bottles
Sure/Seal™ Bottles
Aluminum Bottles
Packing for Bottles
Bottle/Packaging Specifications
For Amber Glass, PVC Coated, and Sure/Seal™ Bottles
Capacity Bottle Dimension
(H x D) cm
Closure Case Pack Case Pack Dimension
(L x W x H) cm
100 mL 11.91 x 5.51 33-430 12 x 100 mL 34.93 x 24.13 x 24.77
500 mL 18.75 x 8.81 33-430 6 x 500 mL 34.93 x 24.13 x 24.77
1 L 22.86 x 10.34 33-430 6 x 1 L 39.37 x 26.83 x 27.94
2 L 33.98 x 13.33 38-439 4 x 2 L 33.78 x 37.78 x 40.00
4 L 34.29 x 16.20 38-439 4 x 4 L 33.78 x 37.78 x 40.00
Amber Glass Bottles
Amber_Glass_Bottle Products available in Amber glass bottles
Sigma-Aldrich Solvents are available in a variety of amber glass bottles. The inert composition of glass makes them ideal for storing and dispensing of high purity solvents.
Back to Top
PVC Coated Bottles
  Products available in PVC coated bottles
To help prevent chemical spills, Sigma-Aldrich provides solvents in the shatter-resistant, poly vinyl chloride coated bottles. The plastic coating aids in absorbing some of the shock in the event the bottle is dropped and assists in containing the spill. Selected solvents are available in 100mL, 1L and 4L PVC coated bottles (refer to above Bottle/Packaging Specifications).
Hirschmann_dispenser Pumping
Hirschmann® Ceramus® bottle-top dispenser (Z34,147-9)
  • Chemically inert − unique ceramic dispenser piston in a Duran borosilicate glass barrel
  • Micro-step volume adjustment − ensures reproducibility
  • Rotates on the bottle − for convenience
  • Recirculation system − for fast priming without loss of reagent
  • Lifetime calibration − supplied with individual serial number and Test Certificate
HPLC_Caps HPLC Delivery Caps
For direct solvent delivery to HPLC instruments or simple pressure transfers. Each Delivery Cap includes 10ft. of PFA tubing. Caps require a GL-45 bottle adapter for use with 2L and 4L bottles.
Two-hole Delivery Cap (Z54,384-5)
Three-hole Delivery Cap (Z54,358-6)
Glass_Bottle_Storage Glass Bottles Storage and Dispensing Options
Aldrich 4L Bottle Jacket (Z25,636-6)
Provides virtually unbreakable, secondary containment for transport and storage of chemicals. Meets OSHA Laboratory Standards (29CFR1910-1450).
  • Rugged PE construction
  • Solvent labels and levels clearly visible through jacket
  • Convenient, handle-locking mechanism keeps top and bottom of jacket locked together
Back to Top
Sure/Seal™ Bottles
Sure_Seal Products available in Sure/Seal bottles
The Sure/Seal bottle incorporates the inert composition of our amber glass bottles with a time-tested crimp-top system. This design ensures the exceptional state of dryness achieved at time of purification is maintained throughout long-term storage and use.
Before filling, low levels traces of moisture are removed from the bottle through oven drying. The bottles are then filled under an inert atmosphere of dry nitrogen to prevent moisture build-up and sealed with our unique Sure/Seal liner and crown cap. Anhydrous and Biotech grades are available in 100mL, 1L and 2L Sure/Seal bottles (refer to above Bottle/Packaging Specifications).
Sure_Seal_Accessories Dispensing Methods for Sure/Seal Bottles
Standard syringe and cannula techniques are used to transfer the contents.
See Technical Bulletin AL-134.
Sure/Seal Transfer Accessories
CHEM-FLEX Transfer Line
Two 12-gauge SS needles (6 and 18 in.) are connected to the CHEM-FLEX 106 tubing with nylon clamps. Solvents contact only PTFE and SS during transfers.
Sure_Store_Flasks Aldrich Sure/Stor™ flasks (Z40,449-3)
Designed for safe, reliable storage and dispensing of air-sensitive and odoriferous chemicals, solvents, pyrophorics, alkyl lithiums, Grignards and corrosives.
Features:
  • High-vacuum PTFE valve-to-glass seal eliminates air contamination in storage and septum leakage after initial needle penetration
  • Heavy-wall borosilicate glass
  • Removable side-arm hose connector for easy attachment of vacuum and inert gas lines. For long-term storage, replace hose connector with a rubber septum for secondary protection.
Back to Top
Aluminum Bottles
Products available in Aluminum Bottles
These one-piece seamless aluminum bottles are corrosion resistant and U.N. Approved.
  • Pure 1050A aluminum (min. 99.5%)
  • Polypropylene closure with PTFE liner
  • Anti-back off, tamper evident locking closure
  • Easily recyclable
aluminum-bottle
Capacity Bottle Dimension (H x D) cm Closure Case Pack Case Pack Dimension (L x W x H) cm
1 L 24.3 x 8.8 32 mm 6 x 1 L 39.37 x 26.83 x 27.94
4 L 23.9 x 17.5 45 mm 4 x 4 L 36.20 x 36.20 x 39.37
Back to Top