|
Whether you need HPLC chloroform, ACS chloroform, Anhydrous chloroform or another Sigma-Aldrich grade, we stock the right product for your application. As a leading supplier of high-purity, research grade solvents, we have the chloroform to meet your exact needs.
Select a solvent
| Sigma-Aldrich Chloroform |
| Synonym |
Trichloromethane |
Beilstein Registry |
1731042 |
| Molecular Formula |
CHCI3 |
EG/EC Number |
2006638 |
| Molecular Weight |
119.38 |
MDL Number |
MFCD00000826 |
| CAS Number |
67-66-3 |
Vapor Pressure |
160 mm Hg (20°C) |
| Density |
1.492 g/mL at 25ºC |
Dielectric Constant |
4.81 at 20ºC |
| Boiling Point |
60.5-61.5ºC |
Dipole Moment |
1.040 |
| Melting Point |
-63ºC |
UV cutoff |
245 nm |
| Flash Point |
- |
Refractive Index |
1. 445 at 20ºC |
| Viscosity |
0.117 at 25ºC |
Vapor Density |
4.1 (vs. air) |
| RIDADR |
UN 1888 6.1/PG 3 |
Risk Statements |
22-38-40-48/20/22 |
| Hazard Codes |
Xn |
Safety Statements |
36/37 |
|
| Chlotoform Miscibility / Immiscibility Table |
|