Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Solvent Center > Chloroform > Amylene Specification
Solvents -- Chloroform
Amylene Specification
Amylene Stabilized Chloroform Specification Comparison PDF version (126 kb)
Sigma-Aldrich Chloroform Specifications Comparison
Stabilized with Amylenes
  CHROMASOLV
Plus
CHROMASOLV
for HPLC
Anyhydrous ACS
Spectro-
photometric
ACS
Purity ≥99.9% ≥99.8% ≥99% ≥99.8% ≥99.8%
UV Absorption
λ: 245 nm ≤1.00     ≤1.00  
λ: 250 nm ≤0.30 ≤1.00      
λ: 255 nm       ≤0.25  
λ: 260 nm   ≤0.30   ≤0.15  
λ: 270 nm       ≤0.05  
λ: 275 nm ≤0.005        
λ: 280 nm   ≤0.005      
λ: 290 nm       ≤0.01  
λ: 400 nm ≤0.005     ≤0.01  
Total Impurities
Stabilizer Amylenes Amylenes Amylenes Amylenes Amylenes
Water ≤0.02% ≤0.03% ≤0.001% ≤0.03% ≤0.03%
    ≤0.005% (100mL)    
Evaporation Residue ≤0.0001% ≤0.0005% ≤0.0003% ≤0.001% ≤0.001%
Halogenated Residue ≤10 ng/L (as Heptachlor epoxide)        
Fluorescence 365 nm ≤0.5 ppb        
Acetaldehyde       Passes Test Passes Test
Acetone       Passes Test Passes Test
Titratable acid   ≤0.001% free acid (as HCL)      
Free Chlorine       Passes Test Passes Test
Formaldehyde       Passes Test Passes Test
Lead       ≤0.05 ppm ≤0.05 ppm
Packaging Options
  CHROMASOLV
Plus
CHROMASOLV
for HPLC
Anyhydrous ACS
Spectro-
photometric
ACS
Glass Bottle 650498 34854   154733 472476
PVC Coated Bottle         480150
Sure/Seal     372978    
Aluminum Bottle         676896
NOWPAK   34854      
Pure-Pac     372978    
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!