|
Sigma-Aldrich Chloroform Specifications Comparison
Stabilized with Ethanol
|
| |
CHROMASOLV Plus |
CHROMASOLV for HPLC |
Biotech |
Anyhydrous |
ACS Spectro- photometric |
ACS |
ReagentPlus |
| Purity |
≥99.9% |
≥99.8% |
≥99.8% |
≥99% |
≥99.8% |
≥99.8% |
≥99.8% |
| UV Absorption |
| λ: 245 nm |
≤1.00 |
≤1.00 |
|
|
≤1.00 |
|
|
| λ: 250 nm |
≤0.30 |
|
| λ: 255 nm |
|
≤0.15 |
|
|
≤0.25 |
|
|
| λ: 260 nm |
|
≤0.15 |
|
|
≤0.15 |
|
|
| λ: 270 nm |
|
≤0.02 |
|
|
≤0.05 |
|
|
| λ: 275 nm |
≤0.005 |
|
| λ: 290 nm |
|
≤0.01 |
|
|
≤0.01 |
|
|
| λ: 400 nm |
≤0.005 |
≤0.01 |
|
|
≤0.01 |
|
|
| Total Impurities |
| Stabilizer |
0.5-1.0% Ethanol |
0.5-1.0% Ethanol |
0.5-1.0% Ethanol |
0.5-1.0% Ethanol |
0.5-1.0% Ethanol |
0.5-1.0% Ethanol |
0.5-1.0% Ethanol |
| Water |
≤0.02% |
≤0.01% |
≤0.01% |
≤0.001% |
≤0.03% |
|
≤0.03% |
| |
|
|
≤0.005% (100mL) |
|
|
|
| Evaporation Residue |
≤0.0001% |
≤0.0003% |
≤0.0003% |
≤0.0003% |
≤0.001% |
≤0.001% |
|
| Halogenated Residue |
≤10 ng/L (as Heptachlor epoxide) |
|
| Fluorescence 365 nm |
≤0.5 ppb |
|
| Acetaldehyde |
|
Passes Test |
|
|
Passes Test |
Passes Test |
|
| Acetone |
|
Passes Test |
|
|
Passes Test |
Passes Test |
|
| Titratable acid |
|
Passes Test |
Passes Test |
|
| Free Chlorine |
|
Passes Test |
Passes Test |
|
Passes Test |
Passes Test |
|
| Formaldehyde |
|
Passes Test |
|
|
Passes Test |
Passes Test |
|
| Lead |
|
≤0.05 ppm |
≤0.05 ppm |
|
≤0.05 ppm |
≤0.05 ppm |
|
| Packaging Options |
| |
CHROMASOLV Plus |
CHROMASOLV for HPLC |
Biotech |
Anyhydrous |
ACS Spectro- photometric |
ACS |
ReagentPlus |
| Glass Bottle |
650471 |
366927 |
|
288306 |
366919 |
319988 |
132950 |
| PVC Coated Bottle |
|
439142 |
|
|
|
437581 |
320242 |
| Sure/Seal™ |
|
|
496189 |
|
| Aluminum Bottle |
|
676888 |
|
| Pure-Pac™ |
|
|
496189 |
|
| Carbon Steel |
|
319988 |
132950 |
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!