|
Solvents -- Chloroform
Without UV Specification
|
|
|
PDF version (74 kb)
|
Sigma-Aldrich Chloroform Specifications Comparison
Without UV Specifications
|
| |
Biotech |
Anhydrous |
Anhydrous |
ACS |
ACS |
ReagentPlus |
| Purity |
≥99.8% |
≥99% |
≥99% |
≥99.8% |
≥99.8% |
≥99.8% |
| Total Impurities |
| Stabilizer |
0.5-1.0% Ethanol |
0.5-1.0% Ethanol |
Amylenes |
0.5-1.0% Ethanol |
Amylenes |
0.5-1.0% Ethanol |
| Water |
≤0.01% |
≤0.001% |
≤0.001% |
|
≤0.03% |
≤0.03% |
| |
≤0.005% (100mL) |
≤0.005% (100mL) |
|
|
|
| Evaporation Residue |
≤0.0003% |
≤0.0003% |
≤0.0003% |
≤0.001% |
≤0.001% |
|
| Acetaldehyde |
|
|
|
Passes Test |
Passes Test |
|
| Acetone |
|
|
|
Passes Test |
Passes Test |
|
| Titratable acid |
|
|
|
Passes Test |
|
|
| Free Chlorine |
Passes Test |
|
|
Passes Test |
Passes Test |
|
| Formaldehyde |
|
|
|
Passes Test |
Passes Test |
|
| Lead |
≤0.05 ppm |
|
|
≤0.05 ppm |
≤0.05 ppm |
|
| Packaging Options |
| |
Biotech |
Anhydrous |
Anhydrous |
ACS |
ACS |
ReagentPlus |
| Glass Bottle |
|
288306 |
|
319988 |
472476 |
132950 |
| PVC Coated Bottle |
|
|
|
437581 |
480150 |
320242 |
| Sure/Seal™ |
496189 |
|
372978 |
|
|
|
| Aluminum Bottle |
|
|
|
676888 |
676896 |
|
| Pure-Pac™ |
496189 |
|
372978 |
|
|
|
| Carbon Steel |
|
|
|
319988 |
|
132950 |
For a complete list of all available unit sizes and product specifications, simply click on the product numbers above!