Sigma-Aldrich
Chemistry > Solvent Center > Chloroform > Without UV Specification
Solvents -- Chloroform
Without UV Specification
Chloroform Without UV Specifications Specification Comparison PDF version (74 kb)
Sigma-Aldrich Chloroform Specifications Comparison
Without UV Specifications
  Biotech Anhydrous Anhydrous ACS ACS ReagentPlus
Purity ≥99.8% ≥99% ≥99% ≥99.8% ≥99.8% ≥99.8%
Total Impurities
Stabilizer 0.5-1.0% Ethanol 0.5-1.0% Ethanol Amylenes 0.5-1.0% Ethanol Amylenes 0.5-1.0% Ethanol
Water ≤0.01% ≤0.001% ≤0.001%   ≤0.03% ≤0.03%
  ≤0.005% (100mL) ≤0.005% (100mL)      
Evaporation Residue ≤0.0003% ≤0.0003% ≤0.0003% ≤0.001% ≤0.001%  
Acetaldehyde       Passes Test Passes Test  
Acetone       Passes Test Passes Test  
Titratable acid       Passes Test    
Free Chlorine Passes Test     Passes Test Passes Test  
Formaldehyde       Passes Test Passes Test  
Lead ≤0.05 ppm     ≤0.05 ppm ≤0.05 ppm  
Packaging Options
  Biotech Anhydrous Anhydrous ACS ACS ReagentPlus
Glass Bottle   288306   319988 472476 132950
PVC Coated Bottle       437581 480150 320242
Sure/Seal™ 496189   372978      
Aluminum Bottle       676888 676896  
Pure-Pac™ 496189   372978      
Carbon Steel       319988   132950
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!