CHROMASOLV® LC-MS
|
Complete list of LC-MS grade solvents
The LC-MS solvents are designed specifically with low content of alkaline impurities, such as calcium, magnesium, potassium, and sodium, which can interfere in the analysis by forming artifacts with the analyte.
The CHROMASOLV® LC-MS solvents are also run through specific UV-spectroscopic quality control tests to guarantee the traditional CHROMASOLV® specification.
For more details on LC-MS Solvents or to see our latest researches: View the LC-MS CHROMASOLV® Flyer (491 K PDF)
|
Back to Top
CHROMASOLV® Plus
| Complete list of Plus grade solvents
Manufactured to go beyond our CHROMASOLV® for HPLC to be your high-purity, multipurpose solvent. Tested for suitability in HPLC with gradient analysis, spectrophotometry, environmental testing and some LC-MS applications.
CHROMASOLV® Plus solvents are available in a variety of container types: Glass Bottles and NOWPAK CHROMASOLV® Plus Specification Sheet Acetonitrile Methanol Water
|
Back to Top
CHROMASOLV® Gradient
| Complete list of Gradient grade solvents
The same high quality as our CHROMASOLV® for HPLC, but with added testing for suitability in HPLC with gradient analysis and spectrophotometry. Eliminates any Impurity Peaks which could be caused by the solvent rather than by the compound being tested – Solvent impurity peaks are less than 0.005 absorbent units.
CHROMASOLV® Gradient solvents are available in a variety of container types: Glass Bottles, VerSA-Flow, NOWPAK, PurePac and Drums
|
Back to Top
CHROMASOLV® for HPLC
| Complete list of HPLC grade solvents
Designed for use with HPLC instrumentation and organic synthesis applications. CHROMASOLV® HPLC solvents are glass distilled; submicron filtered and undergoes rigorous specification testing to provide you with lot-to-lot consistency.
CHROMASOLV® for HPLC solvents are available in a variety of container types: Glass Bottles, VerSA-Flow, NOWPAK, PurePac and Drums
|
Back to Top
CHROMASOLV® Mobile Phase
|
Complete list of Pre-Blended solvents
The convenience of accurately blended mobile phase solvents for HPLC and LC-MS applications. Using these precisely blended solvents eliminates time-consuming preparation, and can eliminate lost sample information and instrument downtime caused by impure mobile phases. For consistent baselines and to minimize background noise, these blends are prepared with ultra pure acids.
CHROMASOLV® Mobile Phase solvents are available in a variety of container types: Glass Bottles, VerSA-Flow, NOWPAK, and Drums