Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Solvent Center > Dimethyl sulfoxide > Specification Comparison
Solvents -- Dimethyl sulfoxide
Specification Comparison
Dimethyl sulfoxide (DMSO) Specification Comparison PDF version (116 kb)
Sigma-Aldrich Dimethyl sulfoxide Specifications Comparison
  CHROMASOLV
Plus
Biotech Anhydrous ACS
Spectro-
photometric
ACS ReagentPlus
Purity ≥99.7% 99.98% ≥99.9% ≥99.9% ≥99.9% ≥99.6%
UV Absorption
λ: 270 nm ≤0.70 ≤1.00   ≤0.40    
λ: 280 nm ≤0.30 ≤0.30        
λ: 290 nm       ≤0.18    
λ: 295 nm   ≤0.10        
λ: 300 nm ≤0.10          
λ: 310 nm       ≤0.06    
λ: 330 nm       ≤0.02    
λ: 350 nm ≤0.01 ≤0.01   ≤0.01    
λ: 400 nm   ≤0.01   ≤0.01    
Total Impurities
Water ≤0.2% ≤0.005% ≤0.005% ≤0.1% ≤0.1% ≤0.1%
Evaporation Residue ≤0.002% ≤0.0005% ≤0.0005% ≤0.01% ≤0.01%  
Titratable acid       ≤0.001 meq/g ≤0.001 meq/g  
Packaging Options
  CHROMASOLV
Plus
Biotech Anhydrous ACS
Spectro-
photometric
ACS ReagentPlus
Glass Bottle 34869     154938 472301 471267
Sure/Seal   494429 276855      
VerSA-Flow         472301 471267
NOWPAK 34869          
Pure-Pac   494429 276855      
Drum 34869       472301  
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!