|
Whether you need HPLC ethyl acetate, ACS ethyl acetate, Anhydrous ethyl acetate or another Sigma-Aldrich grade, we stock the right product for your application. As a leading supplier of high-purity, research grade solvents, we have the ethyl acetate to meet your exact needs.
Select a solvent
| Ethyl Acetate Physical Properties |
| Synonym |
Ethyl ethanoate |
Beilstein Registry |
506104 |
| Molecular Formula |
CH3COOC2H5 |
EG/EC Number |
2055004 |
| Molecular Weight |
88.11 |
MDL Number |
MFCD00009171 |
| CAS Number |
141-78-6 |
Vapor Pressure |
73 mm Hg (20°C) |
| Density |
0.902 g/mL at 25ºC |
Dielectric Constant |
6.08 at 20ºC |
| Boiling Point |
76.5-77.5ºC |
Dipole Moment |
1.780 |
| Melting Point |
-84ºC |
UV cutoff |
255 nm |
| Flash Point |
-3.3ºC |
Refractive Index |
1.372 at 20ºC |
| Viscosity |
0.423 at 25ºC |
Vapor Density |
3 (vs. air) |
| RIDADR |
UN 1173 3/PG 2 |
Risk Statements |
11-36-66-67 |
| Hazard Codes |
F,Xi |
Safety Statements |
16-26-33 |
| |
|
| Ethyl Acetate Miscibility / Immiscibility Table |
|