|
Solvents -- Ethyl acetate
Specification Comparison
|
|
|
PDF version (117 kb)
| Sigma-Aldrich Ethyl acetate Specifications Comparison |
| |
CHROMASOLV LC-MS |
CHROMASOLV Plus |
CHROMASOLV for HPLC |
Biotech |
Anhydrous |
ACS Spectro- photometric
|
ACS
|
ReagentPlus |
| Purity |
≥99.7% |
≥99.9% |
≥99.7% |
≥99.8% |
99.8% |
≥99.5% |
≥99.5% |
≥99.5% |
| UV Absorption |
| λ: 254 nm |
|
|
|
≤1.00 |
|
| λ: 255 nm |
|
|
≤0.70 |
|
|
≤1.00 |
|
|
| λ: 256 nm |
|
≤1.00 |
|
| λ: 257 nm |
|
≤0.50 |
|
|
| λ: 260 nm |
≤0.30 |
|
≤0.30 |
|
| λ: 263 nm |
|
|
|
≤0.05 |
|
≤0.10 |
|
|
| λ: 270 nm |
|
|
≤0.10 |
|
| λ: 275 nm |
|
≤0.05 |
|
≤0.01 |
|
≤0.05 |
|
|
| λ: 300 nm |
≤0.009 |
≤0.03 |
≤0.01 |
|
| λ: 325 nm |
|
≤0.005 |
|
| λ: 330 nm |
|
≤0.01 |
|
|
| λ: 400 nm |
|
≤0.005 |
|
≤0.01 |
|
≤0.01 |
|
|
| Total Impurities |
| Water |
≤0.03% |
≤0.03% |
≤0.03% |
≤0.01% |
≤0.005% |
≤0.20% |
≤0.20% |
≤0.005 g / 100mL |
| Evaporation Residue |
≤0.0005% |
≤0.0001% |
≤0.0005% |
≤0.0003% |
≤0.0005% |
≤0.003% |
≤0.003% |
≤0.005% |
| Halogenated Residue |
|
≤10 ng/L (as Heptachlor epoxide) |
|
| Fluorescence 365 nm |
|
≤1.0 ppb |
|
| LC-MS suitability |
Passes Test |
|
| Titratable acid |
|
|
≤0.005% free acid (as CH3COOH) |
0.0009 meq/g |
|
0.0009 meq/g |
0.0009 meq/g |
|
| Alkalinity |
≤0.0005% (NH3) |
|
| Calcium |
0.1 ppm |
|
| Magnesium |
0.1 ppm |
|
| Potassium |
0.1 ppm |
|
| Sodium |
0.1 ppm |
|
| Packaging Options |
| |
CHROMASOLV LC-MS |
CHROMASOLV Plus |
CHROMASOLV for HPLC |
Biotech |
Anhydrous |
ACS Spectro- photometric |
ACS
|
ReagentPlus |
| Glass Bottle |
34972 |
650528 |
34858 |
|
|
154857 |
319902 |
537446 |
| PVC Coated Bottle |
|
|
439169 |
|
|
|
437549 |
|
| Sure/Seal™ |
|
|
|
494518 |
270989 |
|
|
|
| Aluminum Bottle |
|
676810 |
|
| NOWPAK™ |
|
|
439169 |
|
| Pure-Pac™ |
|
|
34858 |
|
270989 |
|
|
|
| Carbon Steel |
|
319902 |
537446 |
| Drum |
|
|
439169 |
|
|
|
319902 |
|
For a complete list of all available unit sizes and product specifications, simply click on the product numbers above!