| Complete list of Reagent & ReagentPlus® solvents
We offer a wide variety of general applications solvents to meet your needs. ReagentPlus solvents have a guaranteed purity of ≥98.5% when higher purities are required such as specialty organic synthesis, preparative scale liquid chromatography or glass washing.
For general laboratory use, try our Reagent grade solvents; ≥95% purity.
Both of our Reagent and ReagentPlus solvents are available are available in a wide variety of containers options including: Amber Glass Bottle, PVC Coated Glass Bottle, Safety Can, Carbon Steel Flex-Spout Can , Carbon Steel Can with NPT Threads, VerSA-Flow Container , Closed Head Steel Drum or Composite Drum.
|