|
Whether you need HPLC hexane, ACS hexane, Anhydrous hexane or another Sigma-Aldrich grade, we stock the right product for your application. As a leading supplier of high-purity, research grade solvents, we have the hexane to meet your exact needs.
Select a solvent
| Hexane Physical Properties |
| Synonym |
n-Hexane |
Beilstein Registry |
1730733 |
| Molecular Formula |
CH3(CH2)4CH3 |
EG/EC Number |
2037776 |
| Molecular Weight |
86.18 |
MDL Number |
MFCD00009520 |
| CAS Number |
110-54-3 |
Vapor Pressure |
132 mm Hg (20°C) |
| Density |
0.659 g/mL at 25ºC |
Dielectric Constant |
1.89 at 20ºC |
| Boiling Point |
69ºC |
UV cutoff |
210 nm |
| Melting Point |
-95ºC |
Refractive Index |
1.375 at 20ºC |
| Flash Point |
-23.3ºC |
Vapor Density |
3 (vs. air) |
| Viscosity |
0.300 at 25ºC |
Risk Statements |
11-38-48/20-51/53-62-65-67 |
| RIDADR |
UN 1208 3/PG 2 |
Safety Statements |
9-16-29-33-36/37-61-62 |
| Hazard Codes |
F,Xn,N |
| Hexane, mixture of isomers Physical Properties |
| Synonym |
Hexanes |
Beilstein Registry |
1730733 |
| Molecular Formula |
C6H14 |
EG/EC Number |
2035234 |
| Molecular Weight |
86.18 |
MDL Number |
MFCD00009520 |
| Density |
0.672 g/mL at 25ºC |
Dielectric Constant |
1.89 at 20ºC |
| Boiling Point |
68-70ºC |
UV cutoff |
195 nm |
| Melting Point |
-95ºC |
Refractive Index |
1.379 at 20ºC |
| Flash Point |
-22.7ºC |
Vapor Density |
3 (vs. air) |
| Viscosity |
0.286 at 25ºC |
Risk Statements |
11-38-48/20-51/53-62-65-67 |
 |
| Hexane Miscibility / Immiscibility Table |