Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Solvent Center > High Purity Solvents
Solvents
High Purity Solvents
Spectrophotometric Solvents
Tax-Paid Ethanol
Pesticide Residue Analysis Solvents
Capillary GC Solvents
NMR Solvents
Electronic Solvents
Spectrophotometric Solvents
Complete list of Spectrophotometric grade solvents
Sigma-Aldrich Spectrophotometric solvents are excellent for applications requiring high-purity, low residue on evaporation and a clean UV spectrum. Each one is rigorously tested during and after production to ensure lot-to-lot consistency is maintained.
Sigma-Aldrich Spectrophotometric solvents are available in Amber Glass Bottles .
Back to Top
Tax-Paid Ethanol
Complete list of Tax-Paid Ethanol
Purchasing 190 and 200 proof ethanol has never been easy, until now! Because these ethanols are federally regulated, an ATF permit is required to purchase 190 and 200 proof ethanol.
Sigma-Aldrich takes the hassle out of this process for you! There is no ATF paperwork needed or records to be kept on your part. Sigma-Aldrich immediately pays the federal excise tax (200-proof pure alcohol = $27.00/gallon).
For more regulatory information regard the tax on ethanol, visit the U.S. Department of the Treasury, Alcohol and Tobacco Tax and Trade Bureau.
We offer tax-paid ethanol in a variety of grades to meet your application and are available in Amber Glass Bottles and Sure/Seal Bottles
Back to Top
Pesticide Residue Analysis Solvents
Complete list of Pesticide Residue Analysis solvents
Sigma-Aldrich PRA solvents are glass-distilled and extensively tested by GC/ECD to ensure halogenated residue levels is kept to a minimum (<10ng/Liter unless otherwise indicated).
All PRA solvents are available in Amber Glass Bottles
Back to Top
Capillary GC Solvents
Complete list of GC solvents
Sigma-Aldrich GC solvents are specially produced for those working with the stringent demands of capillary gas chromatography and are tested by GC/ECD to ensure that no peak greater than 2ng/Liter as lindane. Each solvent is glass-distilled, resulting in exceptional quality and low residue levels.
All GC solvents are available in Amber Glass Bottles
Back to Top
NMR Solvents
Complete list of NMR solvents
Our wide range NMR solvents provide you with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available. The highest deuterium enrichment solvents are available exclusively from Sigma-Aldrich.
Back to Top
Electronic Solvents
Complete list of Electronic Solvents
Electronic grade solvents are used extensively throughout the semiconductor industry including cleaning equipment, drying wafers and substrate deposition or removal.
Back to Top