Sigma-Aldrich
Chemistry > Solvent Center > Methanol
Solvent Center
Methanol
Whether you need HPLC methanol, ACS methanol, Anhydrous methanol or one of our many other grades, we stock the right product for your application. As a leading supplier of high-purity, research grade solvents, we have the methanol to meet your exact needs.
  •  Acetone 
  •  Acetonitrile 
  •  Chloroform 
  • Dichloromethane 
  • Dimethyl sulfoxide
  • Ethyl Acetate
  •  Hexane 
  •  Methanol 
  •  2-Propanol 
  • Tetrahydrofuran
  • Toluene
  •  Water 
Select a solvent
View and Compare Methanol Products
Methanol Physical Properties
Synonym Methyl alcohol Beilstein Registry 1098229
Molecular Formula CH3OH EG/EC Number 2006596
Molecular Weight 32.04 MDL Number MFCD00004595
CAS Number 67-56-1 Vapor Pressure 410 mm Hg (50ºC)
97.68 mm Hg (20ºC)
Density 0.791 g/mL at 25ºC Dielectric Constant 33.00 at 20ºC
Boiling Point 64.7ºC Dipole Moment 1.700
Melting Point -98ºC UV cutoff 210 nm
Flash Point 11.1ºC Refractive Index 1.329 at 20ºC
Viscosity 0.544 at 25ºC Vapor Density 1.11 (vs. air)
RIDADR UN 1230 3/PG 2 Risk Statements 11-23/24/25-39/23/24/25
Hazard Codes F,T Safety Statements 7-16-36/37-45
image0011  image003
Methanol Miscibility / Immiscibility Table
im-miscible
Back to Solvent Center