|
Solvents -- Methanol
Specification Comparison
|
|
|
PDF version (124 kb)
| Sigma-Aldrich Methanol Specifications Comparison |
| |
CHROMASOLV LC-MS |
CHROMASOLV Plus |
CHROMASOLV Gradient |
CHROMASOLV for HPLC |
Biotech |
Anhydrous |
ACS Spectro- photometric |
ACS |
ReagentPlus |
| Purity |
≥99.9% |
≥99.9% |
≥99.9% |
≥99.9% |
99.93% |
99.8% |
≥99.9% |
≥99.8% |
≥99% |
| UV Absorption |
| λ: 203 nm |
|
≤1.00 |
|
|
≤1.00 |
|
≤1.00 |
|
|
| λ: 205 nm |
|
≤1.00 |
|
≤1.00 |
|
|
| λ: 210 nm |
≤0.52 |
≤0.80 |
≤0.50 |
≤0.50 |
≤0.60 |
|
≤0.80 |
|
|
| λ: 220 nm |
≤0.30 |
≤0.20 |
≤0.30 |
≤0.30 |
≤0.30 |
|
≤0.40 |
|
|
| λ: 230 nm |
≤0.12 |
≤0.10 |
|
|
≤0.15 |
|
≤0.20 |
|
|
| λ: 235 nm |
|
|
≤0.10 |
≤0.10 |
|
| λ: 240 nm |
|
≤0.05 |
|
≤0.10 |
|
|
| λ: 250 nm |
|
≤0.02 |
|
| λ: 260 nm |
≤0.009 |
|
≤0.01 |
≤0.01 |
≤0.01 |
|
≤0.04 |
|
|
| λ: 280 nm |
|
≤0.01 |
|
|
| λ: 400 nm |
|
≤0.005 |
|
|
≤0.01 |
|
≤0.01 |
|
|
| Gradient analysis |
| 230 nm |
≤2 mAU |
≤2 mAU |
≤2 mAU |
|
≤2 mAU |
|
| 254 nm |
|
≤5 mAU |
≤5 mAU |
|
≤5 mAU |
|
| Total Impurities |
| Water |
≤0.02% |
≤0.05% |
≤0.02% |
≤0.03% |
≤0.02% |
≤0.002% |
≤0.10% |
≤0.10% |
|
| |
|
≤0.005% (100mL) |
|
|
|
| Evaporation Residue |
≤0.0005% |
≤0.0002% |
≤0.0005% |
≤0.0005% |
≤0.0003% |
≤0.0003% |
≤0.001% |
≤0.001% |
|
| Halogenated Residue |
|
≤10 ng/L (as Heptachlor epoxide) |
|
| Fluorescence 254 nm |
≤1.0 ppb |
≤1.0 ppb |
≤1.0 ppb |
≤1.0 ppb |
|
| Fluorescence 365 nm |
≤1.0 ppb |
≤1.0 ppb |
≤1.0 ppb |
≤1.0 ppb |
|
| LC-MS suitability |
passes test |
|
| Alkalinity |
≤0.0005% (NH3) |
|
≤0.00035% (NH3) |
|
| Acetaldehyde |
|
≤0.001% |
|
≤0.001% |
≤0.001% |
|
| Acetone |
|
≤0.001% |
|
≤0.001% |
≤0.001% |
|
| Titratable acid |
≤0.001% free acid (as HCOOH) |
0.0003 meq/g |
|
|
0.0003 meq/g |
|
0.0003 meq/g |
0.0003 meq/g |
|
| Titratable base |
|
0.0002 meq/g |
|
|
0.0002 meq/g |
|
0.0002 meq/g |
0.0002 meq/g |
|
| Formaldehyde |
|
≤0.001% |
|
≤0.001% |
≤0.001% |
|
| Silver |
0.1 ppm |
|
| Aluminum |
0.5 ppm |
|
| Barium |
0.1 ppm |
|
| Calcium |
0.1 ppm |
|
| Cadmium |
0.05 ppm |
|
| Cobalt |
0.02 ppm |
|
| Chromium |
0.02 ppm |
|
| Copper |
0.01 ppm |
|
| Iron |
0.1 ppm |
|
| Potassium |
0.1 ppm |
|
| Magnesium |
0.1 ppm |
|
| Manganese |
0.01 ppm |
|
| Sodium |
0.1 ppm |
|
| Nickel |
0.02 ppm |
|
| Lead |
0.02 ppm |
|
| Tin |
0.1 ppm |
|
| Zinc |
0.1 ppm |
|
| Packaging Options |
| |
CHROMASOLV LC-MS |
CHROMASOLV Plus |
CHROMASOLV Gradient |
CHROMASOLV for HPLC |
Biotech |
Anhydrous |
ACS Spectro- photometric |
ACS |
ReagentPlus |
| Glass Bottle |
34966 |
646377 |
34885 |
34860 |
|
|
154903 |
179337 |
179957 |
| PVC Coated Bottle |
|
650609 |
439193 |
|
320390 |
|
| Sure/Seal™ |
|
494437 |
322415 |
|
|
|
| Aluminum Bottle |
|
676780 |
|
| VerSA-Flow™ |
|
|
439193 |
|
320390 |
|
| NOWPAK™ |
|
|
439193 |
|
| Pure-Pac™ |
|
|
34885 |
34860 |
|
322415 |
|
|
|
| Carbon Steel |
|
179337 |
179957 |
| Drum |
|
|
439193 |
|
179337 |
|
For a complete list of all available unit sizes and product specifications, simply click on the product numbers above!
CHROMASOLV® Mobile Phase Blends for HPLC & LC-MS Applications