|
Whether you need HPLC tetrahydrofuran, ACS tetrahydrofuran, Anhydrous tetrahydrofuran or one of our many other grades, we stock the right product for your application. As a leading supplier of high-purity, research grade solvents, we have the THF to meet your exact needs.
Select a solvent
| Tetrahydrofuran Physical Properties |
| Synonym |
THF |
Beilstein Registry |
102391 |
| Molecular Formula |
C4H8O |
EG/EC Number |
2037268 |
| Molecular Weight |
72.11 |
MDL Number |
MFCD00005356 |
| CAS Number |
109-99-9 |
Vapor Pressure |
143 mm Hg (20°C) |
| Density |
0. 889 g/mL at 25ºC |
Dielectric Constant |
7.52 at 22ºC |
| Boiling Point |
65-67ºC |
Dipole Moment |
1.750 |
| Melting Point |
-108ºC |
UV cutoff |
220 nm |
| Flash Point |
-17.2ºC |
Refractive Index |
1.407 at 20ºC |
| Viscosity |
0.456 at 25ºC |
Vapor Density |
2.5 (vs. air) |
| RIDADR |
UN 2056 3/PG 2 |
Risk Statements |
11-19-36/37 |
| Hazard Codes |
F,Xi |
Safety Statements |
16-29-33 |
|
| Tetrahydrofuran Miscibility / Immiscibility Table |
|