|
Solvents -- Tetrahydrofuran
Specification Comparison
|
|
|
Printer friendly PDF version (120 kb)
| Sigma-Aldrich Tetrahydrofuran Specifications Comparison |
| |
CHROMASOLV Plus |
Biotech |
Anhydrous |
Anhydrous |
Spectro- photometric |
ACS |
ReagentPlus |
| Purity |
≥99.9% |
≥99.9% |
≥99.9% |
≥99.9% |
≥99.5% |
≥99% |
≥99% |
| UV Absorption |
| λ: 212 nm |
|
≤1.00 |
|
|
≤1.00 |
|
|
| λ: 245 nm |
≤0.30 |
|
| λ: 250 nm |
|
≤0.18 |
|
|
≤0.18 |
|
|
| λ: 275 nm |
≤0.05 |
|
| λ: 300 nm |
|
≤0.02 |
|
|
≤0.02 |
|
|
| λ: 315 nm |
≤0.004 |
|
| λ: 350 nm |
|
≤0.005 |
|
|
≤0.005 |
|
|
| λ: 400 nm |
|
≤0.005 |
|
|
≤0.005 |
|
|
| Total Impurities |
| Inhibitor |
Inhibitor-free |
Inhibitor-free |
Inhibitor-free |
250 ppm BHT |
Inhibitor-free |
250 ppm BHT |
250 ppm BHT |
| Water |
≤0.05% |
≤0.005% |
≤0.002% |
≤0.002% |
≤0.02% |
≤0.05% |
|
| |
|
≤0.005% (100mL) |
≤0.005% (100mL) |
|
|
|
| Evaporation Residue |
≤0.001% |
|
≤0.0005% |
|
≤0.0005% |
≤0.03% |
|
| Titratable acid |
≤0.002% free acid (as CH3COOH) |
|
| Peroxide |
=0.05% peroxide (as H2O2) |
≤0.005% peroxide (as H2O2) |
≤0.002% peroxide (as H2O2) |
|
≤0.015% peroxide (as H2O2) |
≤0.015% peroxide (as H2O2) |
|
| Packaging Options |
| |
CHROMASOLV Plus |
Biotech |
Anhydrous |
Anhydrous |
Spectro- photometric |
ACS |
ReagentPlus |
| Glass Bottle |
34865 |
|
|
|
242888 |
360589 |
178810 |
| PVC Coated Bottle |
439215 |
|
437638 |
|
| Sure/Seal™ |
|
494461 |
401757 |
186562 |
|
|
|
| Aluminum Bottle |
|
676764 |
|
| NOWPAK™ |
34865 |
|
| Pure-Pac™ |
34865 |
494461 |
401757 |
186562 |
|
|
|
| Carbon Steel |
|
360589 |
178810 |
| Drum |
|
360589 |
|
For a complete list of all available unit sizes and product specifications, simply click on the product numbers above!