Sigma-Aldrich
Chemistry > Solvent Center > Toluene
Solvent Center
Toluene
Whether you need HPLC toluene, ACS toluene, Anhydrous toluene or one of our many other grades, we stock the right product for your application. As a leading supplier of high-purity, research grade solvents, we have the toluene to meet your exact needs.
  •  Acetone 
  •  Acetonitrile 
  •  Chloroform 
  • Dichloromethane 
  • Dimethyl sulfoxide
  • Ethyl Acetate
  •  Hexane 
  •  Methanol 
  •  2-Propanol 
  • Tetrahydrofuran
  • Toluene
  •  Water 
Select a solvent
View and Compare Toluene Products
Toluene Physical Properties
Synonym Methylbenzene Beilstein Registry 635760
Molecular Formula C6H5CH3 EG/EC Number 2036259
Molecular Weight 92.14 MDL Number MFCD00008512
CAS Number 108-88-3 Vapor Pressure 22 mm Hg (20°C)
Density 0. 865 g/mL at 25ºC Dielectric Constant 2.38 at 23ºC
Boiling Point 110-111ºC Dipole Moment 0.375
Melting Point -93ºC UV cutoff 286 nm
Flash Point 4.4ºC Refractive Index 1.496 at 20ºC
Viscosity 0.560 at 25ºC Vapor Density 3.2 (vs. air)
RIDADR UN 1294 3/PG 2 Risk Statements 11-38-48/20-63-65-67
Hazard Codes F,Xn Safety Statements 36/37-46-62
image0011  image003
Toluene Miscibility / Immiscibility Table
toluene_miscibility
Back to Solvent Center