Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Solvent Center > Toluene > Specification Comparison
Solvents -- Toluene
Specification Comparison
Toluene Specification Comparison Printer friendly PDF version (118 kb)
Sigma-Aldrich Toluene Specifications Comparison
  CHROMASOLV
Plus
CHROMASOLV
for HPLC
Anhydrous ACS
Spectro-
photometric
ACS ReagentPlus
Purity ≥99.9% 99.9% 99.8% ≥99.5% ≥99.5% 99%
UV Absorption
λ: 284 nm ≤1.00          
λ: 286 nm       ≤1.00    
λ: 288 nm       ≤0.50    
λ: 290 nm   ≤0.30        
λ: 293 nm       ≤0.20    
λ: 300 nm ≤0.10     ≤0.10    
λ: 310 nm   ≤0.05   ≤0.05    
λ: 325 nm ≤0.02          
λ: 335 nm       ≤0.02    
λ: 350 nm ≤0.005 ≤0.004   ≤0.01    
λ: 400 nm ≤0.005     ≤0.01    
Total Impurities
Water ≤0.02% ≤0.02% ≤0.001% ≤0.03% ≤0.03%  
    ≤0.005% (100mL)      
Evaporation Residue ≤0.0001% ≤0.0005% ≤0.0005% ≤0.0004% ≤0.001%  
Halogenated Residue ≤10 ng/L (as Heptachlor epoxide)          
Titratable acid   ≤0.001% free acid (as HCl)        
Sulfur compounds       ≤0.003% ≤0.003%  
Thiophene   ≤0.0005%        
Packaging Options
  CHROMASOLV
Plus
CHROMASOLV
for HPLC
Anhydrous ACS
Spectro-
photometric
ACS ReagentPlus
Glass Bottle 650579 34866   155004 179418 179965
PVC Coated Bottle   648566     320552  
Sure/Seal     244511      
Aluminum Bottle         676756  
NOWPAK   34866        
Pure-Pac   34866 244511      
Carbon Steel         179418 179965
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!