Sigma-Aldrich
Chemistry > Solvent Center > Water
Solvent Center
Water
Whether you need LC-MS water, HPLC water or ACS water, we stock the right product for your application. As a leading supplier of high-purity, research grade solvents, we have the water to meet your exact needs.
  •  Acetone 
  •  Acetonitrile 
  •  Chloroform 
  • Dichloromethane 
  • Dimethyl sulfoxide
  • Ethyl Acetate
  •  Hexane 
  •  Methanol 
  •  2-Propanol 
  • Tetrahydrofuran
  • Toluene
  •  Water 
Select a solvent
View and Compare Water Products
Water Physical Properties
Molecular Formula H2O Beilstein Registry 2050024
Molecular Weight 18.02 EG/EC Number 2317912
CAS Number 7732-18-5 MDL Number MFCD00011332
Density 1.000 g/mL at 3.98 °C Dielectric Constant 80.10 at 20°C
Boiling Point 100°C Dipole Moment 1.82
Melting Point 0.0°C UV cutoff 191 nm
Viscosity 1.000 at 20°C Refractive Index 1.340 at 20°C
water_miscibility
Back to Solvent Center