| Sigma-Aldrich Water Specifications Comparison |
| |
CHROMASOLV LC-MS |
CHROMASOLV Plus |
CHROMASOLV for HPLC |
ACS |
| UV Absorption |
| λ: 200 nm |
≤0.022 |
|
|
|
| λ: 205 nm |
|
|
≤0.01 |
|
| λ: 210 nm |
|
|
≤0.01 |
|
| λ: 230 nm |
≤0.0044 |
|
|
|
| λ: 250 nm |
|
|
≤0.005 |
|
| λ: 400 nm |
|
|
≤0.005 |
|
| Gradient Analysis |
| 210 nm |
≤5 mAU |
≤5 mAU |
≤10 mAU |
|
| 254 nm |
≤1 mAU |
≤1 mAU |
≤2 mAU |
|
| Total Impurities |
| Evaporation Residue |
≤0.0001% |
≤0.0003% |
≤0.0003% |
|
| Specific Conductance |
|
|
|
2.0 uOhm/cm (Max) @ 25C |
| Fluorescence 254 nm |
≤1.0 ppb |
≤1.0 ppb |
|
|
| Fluorescence 365 nm |
≤1.0 ppb |
≤1.0 ppb |
|
|
| LC-MS suitability |
Passes Test |
|
|
|
| Heavy metals |
|
|
|
≤0.01 ppm (as Pb) |
| Chloride |
≤0.01 ppm |
≤0.01 ppm |
|
≤0.4 ppm |
| Fluoride |
≤0.01 ppm |
≤0.01 ppm |
|
|
| Nitrate |
≤0.1 ppm |
≤0.1 ppm |
|
≤0.4 ppm |
| Phosphate |
|
|
|
≤1.0 ppm |
| Silica |
|
|
|
≤0.01 ppm |
| Sulfate |
≤0.1 ppm |
≤0.1 ppm |
|
≤1.0 ppm |
| Silver |
≤0.1 ppm |
|
|
|
| Aluminum |
≤0.5 ppm |
|
|
|
| Barium |
≤0.1 ppm |
|
|
|
| Calcium |
≤0.1 ppm |
|
|
|
| Cadmium |
≤0.05 ppm |
|
|
|
| Cobalt |
≤0.02 ppm |
|
|
|
| Chromium |
≤0.02 ppm |
|
|
|
| Copper |
≤0.02 ppm |
|
|
|
| Iron |
≤0.1 ppm |
|
|
|
| Potassium |
≤0.1 ppm |
|
|
|
| Magnesium |
≤0.1 ppm |
|
|
|
| Manganese |
≤0.02 ppm |
|
|
|
| Sodium |
≤0.1 ppm |
|
|
|
| Nickel |
≤0.02 ppm |
|
|
|
| Lead |
≤0.1 ppm |
|
|
|
| Tin |
≤0.1 ppm |
|
|
|
| Zinc |
≤0.1 ppm |
|
|
|
| Packaging Options |
| |
CHROMASOLV LC-MS |
CHROMASOLV Plus |
CHROMASOLV for HPLC |
ACS |
| Glass Bottle |
39253 |
34877 |
270733 |
320072 |
| VerSA-Flow™ |
|
|
270733 |
|
| NOWPAK™ |
|
34877 |
270733 |
|
| Drum |
|
|
270733 |
|
For a complete list of all available unit sizes and product specifications,
simply click on the product numbers above!
CHROMASOLV® Mobile Phase Blends for HPLC & LC-MS Applications