Chemistry > Special Offers > BioTech Solvent Savings from Aldrich®
Chemistry
BioTech Solvent Savings from Aldrich®
Use Promo Code 410
US Customers Only
Expires November 30, 2009
Save up to 65% Off List Price
Reference promo code 410 when placing your order.
Offer expires November 30, 2009
Biotech solvents are characterized by low water content, minimal residue and clean UV spectra. They are purified for biotechnology applications such as peptide synthesis, oligonucleotide synthesis and protein sequencing.
Product Description List Price Discount Promo Price
494445-1L Acetonitrile, biotech grade, ≥99.93% $96.40 65% $33.74
494445-2L Acetonitrile, biotech grade, ≥99.93% $134.50 65% $47.08
496162-1L Butyl acetate, biotech grade, 99.7% $97.30 65% $34.06
496162-2L Butyl acetate, biotech grade, 99.7% $142.00 65% $49.70
496170-1L 1-Chlorobutane, biotech grade, ≥99.5% $93.60 65% $32.76
496170-2L 1-Chlorobutane, biotech grade, ≥99.5% $137.00 65% $47.95
496189-1L Chloroform, biotech grade, ≥99.8%,
contains 0.5-1.0% ethanol as stabilizer
$54.40 65% $19.04
496189-2L Chloroform, biotech grade, ≥99.8%,
contains 0.5-1.0% ethanol as stabilizer
$77.50 65% $27.13
494453-1L Dichloromethane, biotech grade, 99.9%,
contains 50-150 ppm amylene as stabilizer
$49.20 65% $17.22
494453-2L Dichloromethane, biotech grade, 99.9%,
contains 50-150 ppm amylene as stabilizer
$70.10 65% $24.54
494488-1L N,N-Dimethylformamide, biotech grade, ≥99.9% $77.80 65% $27.23
494488-2L N,N-Dimethylformamide, biotech grade, ≥99.9% $107.50 65% $37.63
494429-1L Dimethyl sulfoxide, biotech grade, 99.8% $102.50 65% $35.88
494429-2L Dimethyl sulfoxide, biotech grade, 99.8% $146.50 65% $51.28
494526-1L Heptane, biotech grade, ≥99% $65.10 60% $26.04
494526-2L Heptane, biotech grade, ≥99% $95.40 60% $38.16
494437-1L Methanol, biotech grade, 99.93% $44.10 65% $15.44
494437-2L Methanol, biotech grade, 99.93% $58.90 65% $20.62
496200-1L 3-Methyl-1-butanol, biotech grade, ≥99% $179.00 60% $71.60
496200-2L 3-Methyl-1-butanol, biotech grade, ≥99% $225.00 60% $90.00
494496-1L 1-Methyl-2-pyrrolidinone, biotech grade, ≥99.5% $97.10 60% $38.84
494496-2L 1-Methyl-2-pyrrolidinone, biotech grade, ≥99.5% $142.00 60% $56.80
496197-1L 1-Propanol, biotech grade, 99.7% $70.30 60% $28.12
496197-2L 1-Propanol, biotech grade, 99.7% $97.60 60% $39.04
494410-1L Pyridine, biotech grade, ≥99.9% $141.50 65% $49.53
494410-2L Pyridine, biotech grade, ≥99.9% $198.00 65% $69.30
494461-1L Tetrahydrofuran, biotech grade, ≥99.9%, Inhibitor-free $86.10 65% $30.14
494461-2L Tetrahydrofuran, biotech grade, ≥99.9%, Inhibitor-free $122.00 65% $42.70
299537-100g Trifluoroacetic acid, ≥99%, purified by redistillation
for protein sequencing
$103.00 50% $51.50
299537-500g Trifluoroacetic acid, ≥99%, purified by redistillation
for protein sequencing
$294.50 50% $147.25