Chemistry > Stable Isotopes > 13C-labeled Acyl Coenzyme A Derivatives
13C-labeled Acyl Coenzyme A Derivatives
Coenzyme A plays an important role in lipid biosynthesis, catabolic/oxidation processes and enzyme regulation. Coenzyme A is a key cofactor that aids in transferring acyl groups during enzymatic reactions. Researchers use isotopic tracer methods to study the kinetics of acyl coenzyme A activity. For example, a recent paper demonstrates a sensitive, accurate and precise method for the quantification of long chain acyl coenzyme A concentrations and enrichment using  13C16-palmitoyl coenzyme A as a tracer.(1) Radiolabeled acyl coenzyme A is also used in developing assays to quantify or measure enzymatic activity. An article describes an efficient purification method for coenzyme A using radiolabeled 14C-acyl coenzyme A.(2) On the other hand, another paper illustrates the use of 13C-malonyl coenzyme A to measure the activity of malonyl coenzyme decarboxylase (3) in an effort to offer a safer, nonradioactive and sensitive method.
  Coenzyme A
Until now, researchers had to rely on synthesizing their desired 13C-labeled acyl coenzyme A or use radiolabeled 14C-coenzyme A to perform their experiments. With increased interest in isotopically labeled coenzyme A tracers, we now offer a range of short-, medium- and long chain 13C-acyl coenzyme A products for your convenience. As a leader in the stable isotope industry, we are the first to provide this unique range of 13C-acyl coenzyme A products with the highest isotopic enrichment and purity to our customers. 
Products of Interest:
Product No. Description Isotopic Enrichment
658650
Acetyl-1, 2-13C2 coenzyme A, lithium salt
99 atom % 13C
685518 
Malonyl coenzyme A, lithium salt
655759 
Malonyl-13C3 coenzyme A, lithium salt, 95 % CP
99 atom % 13C
703885
Octanoyl-2,4,6,8 -13Ccoenzyme A, lithium salt, 95 % CP
99 atom % 13C
675768 
Oleoyl-13C18 coenzyme A, lithium salt, 95 % CP
99 atom % 13C
658200 
Palmitoyl-1-13C coenzyme A, lithium salt, 95 % CP
99 atom % 13C
655716
Palmitoyl-13C16 coenzyme A, lithium salt, 95 % CP
99 atom % 13C
675776 
Stearoyl-13C18 coenzyme A, lithium salt, 95 % CP
99 atom % 13C
References:
1.Sun D, Cree  MG, Wolfe RR. Quantification of the concentration and 13C tracer enrichment of long-chain fatty acyl-coenzyme A in muscle by liquid chromatography/mass spectrometry. Analytical Biochemistry 349 (2006) 87-95.
2. Minkler PE, Kerner J, Ingalls ST, Hoppel CL. Novel isolation procedure for short-, medium-, and long-chain acyl-coenzyme A esters from tissue.  Analytical Biochemistry 376 (2008) 275-276.
3.  Wang  X, Stanley  WC,  Brunengraber  H ,  Kasumov, T. Assay of the activity of malonyl-coenzyme A decarboxylase by gas chromatography-mass spectrometry. Analytical Biochemistry  363 (2007) 169-174.