Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Stable Isotopes in Biomolecular NMR
Isotopically Enriched Components for Biomolecular NMR
ISOTEC offers a wide selection of isotopically enriched components for minimal media such as 15N ammonium salts, D-Glucose-13C6 and D-Glucose-13C6-d7, as well as ISOGRO, a complex growth medium capable of yielding cell-masses comparable to that obtained using LB. Additionally, we have an array of fully labeled amino acids for cell-free synthesis applications.