|
Aldrich Spectral Viewer Instructional Edition is an electronic reference book on CD-ROM that contains FT-IR and FT-NMR spectra for over 500 representative compounds from a variety of chemical functional groups. Users can practice spectral interpretation by comparing chemical shifts and absorptions for related compounds.
The Instructional Edition Contains:
- 506 total compounds
- FT-IR spectra for 428 compounds
- 300 MHz1H FT-NMR spectra for 437 compounds
- 75 MHz13C FT-NMR spectra for 440 compounds
- Both FT-IR and FT-NMR spectra for 359 compounds
Spectral Viewer Program Features:
- Displays FT-NMR and FT-IR Spectra along with chemical structure and technical data
- Text Searching by Chemical Name, Aldrich Catalog Number, CAS Number, and Molecular Formula
- Data Field Searching on Chemical Category and physical properties such as Boiling Point, Melting Point, Density, Flash Point, Molecular Weight, and Refractive Index
- Spectral Processing using Zoom, Peak Labeling, and Peak Integration (FT-NMR only)
- Prints Spectra, structure, technical data, peak listings, and integral table (FT-NMR only)
- Exports Spectrato a file (Bitmap, Metafile, or JPEG formats) or to the clipboard (Bitmap or Metafile formats)
- Web Link Button provides additional technical data from the Sigma-Aldrich website
|
|
| Version |
Cat. No. |
Qty |
| Instructional Edition, single user |
Z53,643-1 |
1 |
| Instructional Edition, single user |
Z53,643-1 |
10 pack |
| Instructional Edition, Network |
Z53,633-4 |
1 |
For more information email labequipment@sial.com.
System Requirements: Pentium 200MHz, Windows 95 / 98 / NT4-SP5 / 2000 / ME / XP, CD-ROM drive, 300MB hard disk space, 64MB RAM, disk space will vary with version installed.
Spectral Viewer is a trademark and Aldrich is a registered trademark of Sigma-Aldrich Biotechnology, LP Chemical Abstracts is a registered trademark of the American Chemical Society Pentium is a registered trademark of Intel Corp. Windows is a registered trademark of Microsoft Corp.
|