Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Stockroom Reagents > Product Highlights > Semi-Bulk Solvents
Chemicals
Semi-Bulk Solvents
Carbon Steel Flex-Spout Can
Carbon Steel NPT Thread Can
VerSA-Flow™ Containers
NOWPAK® Container
Stainless Steel Pure-Pac®
Packaging Specifications
Carbon Steel Flex-Spout Can
20L_Flex_Spout 20L_Flex_Spout_with_Zinc_
Carbon Steel NPT Thread Can
20L_with_pour_out_adapter
VerSA-Flow™ Containers
  • Available in 18L metal containers with HDPE liner
  • Double containment design for Safety. Can be recycled or used to hold waste solvents.
  • Used for many ACS, CHROMASOLV® for HPLC, Reagent Grade and Mobile Phase Solvent Blends
  • For added convenience, use VerSA-Flow™ Dispensing Options – See Technical Bulletin AL-210
  • UN and DOT approved
  • Products available in VerSA-Flow™ containers
18L_VerSA_Flow_Can
NOWPAK® Container
20L_NOWPAK
Semi-Bulk Drums
50L_Drum
Stainless Steel Pure-Pac™
  • Available in 18L containers for Laboratory, development and production scale (Inquire for product availability in 56 L, 200 L and 400 L containers)
  • Standard packaging selected Anhydrous and Biotech grade solvents
  • Stainless steel construction features ultra-clean and corrosion-resistant environment for high purity solvents
  • Larger sized returnable container reduces storage space requirements, analysis costs and solid and hazardous waste disposal costs
  • Enhances safety by eliminating glass bottles and allowing for a closed transfer system that minimizes exposure to the worker and the environment
  • Complete container specifications and "how-to" manual provided
  • Container is product-dedicated
  • No rental fee; only a refundable deposit
  • See Technical Bulletin AL-201
  • Products available in 18L stainless steel Pure-Pac™ containers
Pure_Pack1