Sigma-Aldrich
Mainland China - Phone Number
China (Mainland) > 中国专卖产品 > ECOS Competent Cell
Sigma-Aldrich中国
ECOS Competent Cell
ecos comptent cell   yeastern logo 
— Fastest One in the World 转化只需1分钟
  • 细胞不再脆弱—转化效率更稳定
  • 节省更多时间—无需SOC培养基及恢复过程
  • 转化效率高—效率可达10 7 -101 0 个克隆/μg PUC19
ecos
ecos table
相关技术资料:
ECOSTM Blue Competent Cells[XL1-Blue]
ECOSTM 9-5 Competent Cells  [JM109]
ECOSTM 101 Competent Cells [DH5α]
yT&A Cloning Kit
ecos flow
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page