细胞分析
细胞增殖&细胞计数检测
细胞增殖是生物体生长、发育、繁殖以及遗传的基础,是生物体的重要生命特征。检测细胞在培养基或组织中的生长速率,对于细胞生长和分化研究至关重要。在药物开发过程中也常被用于评估药物的毒性和对癌细胞生长的抑制情况。细胞增殖检测通常是基于DNA含量或细胞代谢实现的,主要的研究方向为对活细胞的代谢活性的检测(基于WST、XTT、MTT等)及对DNA合成的检测(基于BrdU的免疫法或EdU的点击化学法),可通过体内成像、微孔板或流式细胞术检测等多种技术进行细胞示踪。
使用四唑盐进行代谢活性检测
通过添加四咪唑盐到细胞中可进行线粒体内酶代谢活性的测定。活细胞能将四咪唑盐(如MTT、XTT、WST-1)还原成有色的甲瓒或甲瓒盐(Formazan),可通过分光光度计或酶标仪进行检测。Cell Counting Kit–8 (96992)和Cell Proliferation Reagent WST-1(11644807001)中的WST-8被细胞内脱氢酶生物还原后生成的橙色甲臜染料能够溶解在组织培养基中,生成的甲臜量与活细胞数量成正比。基于CCK-8(WST-8)的试剂盒能高度准确地检测细胞增殖,比MTT法及XTT法的灵敏度高5倍,能检测更低的细胞数目。该试剂盒可适用于96孔板培养的贴壁或悬浮细胞。
测定ATP水平检测细胞增殖
活细胞需要不断以ATP的形式输入自由能,因此通过检测ATP的增加水平可检测细胞增殖。ATP Bioluminescence Assay Kit HS II(11699709001)及ATP Bioluminescence Assay Kit CLS II(11699695001)是通过经典的荧光素酶发光对ATP检测定量的试剂盒。前者主要应用于高灵敏度检测极低浓度ATP的实验,后者主要应用于当有持续信号产生而需要高重复性结果的实验。
DNA的合成的检测: DNA的新组装合成是细胞生长的必要条件,故经常被用来进行细胞增殖、细胞活力及凋亡的检测。 BrdU及EdU,都是胸腺嘧啶的非放射性类似物,能掺入增殖细胞的DNA中,可通过对BrdU及EdU的检测,从而对DNA的合成量进行测定。
可通过Cell Proliferation ELISA,BrdU (比色法.) 11647229001或Cell Proliferation ELISA, BrdU (发光法) 11669915001进行细胞群落中DNA合成测定(BrdU based)。优点是:实验结果同增殖细胞数目紧密相关,可一步完成细胞的温和固定和变性,操作方便稳定,不破坏细胞形态。5-Bromo-2´-dU Labeling/Detection Kit I(荧光法)11296736001或Kit II(AP)11299964001可使用试剂盒提供的BrdU单抗,以及酶或荧光偶联二抗,检测单细胞水平的DNA合成(BrdU based)。前者通过免疫荧光检测,后者通过荧光显微镜检测。优点是:使用核酸酶变性DNA,在不伤害细胞完整性的情况下使抗体结合BrdU。体内外均可进行标记,且可同时使用其他标记进行检测,操作安全简便。适用于贴壁或悬浮细胞,冰冻或福尔马林包埋组织切片。
EdU与BrdU检测法不同,EdU-Click 检测,如EdU-Click 594(BCK-EDU594)无需进行DNA变性来检测掺入的EdU,而是通过一次快速、高度特异性的点击反应,将EdU高效地掺入到新合成的DNA之中, 通过EdU与不同荧光染料的特异性反应检测DNA复制活性,从而确地反映细胞的增殖情况。这一类细胞增殖用荧光标记采用温和的点击实验方案,准确且兼容各种抗体实验方案,整个实验可在30分钟内完成,在结果的数据丰富程度方面,堪比多重分析方法。此外,该方法适用于培养的细胞或组织切片,可用流式细胞术分析检测EdU Flow Cytometry Kit 594(BCK-FC594-100);可进行HCS分析或进行细胞裂解液微孔板检测EdU HTS Kit 594(BCK-HTS594-20);也可适用于培养中的细胞,或通过注射方法进行活体实验的小动物样本,进行体内成像分析In Vivo EdU Click Kit 594(BCK594-IV-IM-M)、流式细胞检测或HCS高通量检测。(共有488、555、594、647四种荧光染料可选)。
细胞增殖&细胞计数试剂盒
产品货号 产品名称 产品描述 次数
03285 Cell Counting Kit 细胞计数试剂盒 500
96992 Cell Counting Kit - 8 细胞计数试剂盒(CCK-8) 500
11296736001 5-Bromo-2´-dU Labeling/Detection Kit I BrdU标记细胞检测试剂盒I(免疫荧光) 1000
11299964001 5-Bromo-2´-dU Labeling/Detection Kit II BrdU标记细胞检测试剂盒II(光学显微镜) 1000
11444611001 5-Bromo-2´-dU Labeling/Detection Kit III BrdU标记细胞检测试剂盒III(ELISA) 1000
11810740001 In Situ Cell Proliferation Kit, FLUOS 细胞增殖原位检测试剂盒,IF,FACS 1000
11669915001 Cell Proliferation ELISA, BrdU chem.lum. 细胞增殖ELISA定量检测试剂盒(BrdU化学发光法) 1000
11647229001 Cell Proliferation ELISA,BrdU (colorim.) 细胞增殖ELISA定量检测试剂盒(BrdU比色法) 1000
11465007001 Cell Proliferation Kit I (MTT) 细胞增殖检测试剂盒I (MTT) 2500
11465015001 Cell Proliferation Kit II (XTT) 细胞增殖检测试剂盒 II (MTT) 2500
11644807001 Cell Proliferation Reagent WST-1 细胞增殖检测试剂 WST-1 25ml
05015944001 Cell Proliferation Reagent WST-1, 8 ml 细胞增殖检测试剂 WST-1,8ml 8ml
06432379001 Cell Viability Imaging Kit 细胞活性成像试剂盒 5x96
AEC101 AEC Staining Kit liquid AEC细胞染色试剂盒(细胞增殖检测) 1000
EXTRA2 Mouse ExtrAvidin® Peroxidase Staining Kit Mouse ExtrAvidin®过氧化物酶染色试剂盒 2000
BCK-EDU488 EdU-Click 488 Baseclick增殖检测试剂盒(488) 100
BCK-EDU555 EdU-Click 555 Baseclick增殖检测试剂盒(555) 100
BCK-EDU594 EdU-Click 594 Baseclick增殖检测试剂盒(594) 100
BCK-EDU647 EdU-Click 647 Baseclick增殖检测试剂盒(647) 100
BCK-FC488-100 EdU Flow Cytometry Kit 488 Baseclick流式细胞术增殖检测试剂盒(488) 100
BCK-FC488-50 EdU Flow Cytometry Kit 488 Baseclick流式细胞术增殖检测试剂盒(488) 50
BCK-FC555-100 EdU Flow Cytometry Kit 555 Baseclick流式细胞术增殖检测试剂盒(555) 100
BCK-FC555-50 EdU Flow Cytometry Kit 555 Baseclick流式细胞术增殖检测试剂盒(555) 50
BCK-FC594-100 EdU Flow Cytometry Kit 594 Baseclick流式细胞术增殖检测试剂盒(594) 100
BCK-FC594-50 EdU Flow Cytometry Kit 594 Baseclick流式细胞术增殖检测试剂盒(594) 50
BCK-FC647-100 EdU Flow Cytometry Kit 647 Baseclick流式细胞术增殖检测试剂盒(647) 100
BCK-FC647-50 EdU Flow Cytometry Kit 647 Baseclick流式细胞术增殖检测试剂盒(647) 50
BCK-HTS488-2 EdU HTS Kit 488 Baseclick增殖高通量检测试剂盒(488) 200
BCK-HTS488-20 EdU HTS Kit 488 Baseclick增殖高通量检测试剂盒(488) 2000
BCK-HTS488-4 EdU HTS Kit 488 Baseclick增殖高通量检测试剂盒(488) 400
BCK-HTS555-2 EdU HTS Kit 555 Baseclick增殖高通量检测试剂盒(555) 200
BCK-HTS555-20 EdU HTS Kit 555 Baseclick增殖高通量检测试剂盒(555) 2000
BCK-HTS555-4 EdU HTS Kit 555 Baseclick增殖高通量检测试剂盒(555) 400
BCK-HTS594-2 EdU HTS Kit 594 Baseclick增殖高通量检测试剂盒(594) 200
BCK-HTS594-20 EdU HTS Kit 594 Baseclick增殖高通量检测试剂盒(594) 2000
BCK-HTS594-4 EdU HTS Kit 594 Baseclick增殖高通量检测试剂盒(594) 400
BCK-HTS647-2 EdU HTS Kit 647 Baseclick增殖高通量检测试剂盒(647) 200
BCK-HTS647-20 EdU HTS Kit 647 Baseclick增殖高通量检测试剂盒(647) 2000
BCK-HTS647-4 EdU HTS Kit 647 Baseclick增殖高通量检测试剂盒(647) 400
BCK488-IV-IM-L In Vivo EdU Click Kit 488 Baseclick增殖体内成像检测试剂盒(488) 100
BCK488-IV-IM-M In Vivo EdU Click Kit 488 Baseclick增殖体内成像检测试剂盒(488) 100
BCK488-IV-IM-S In Vivo EdU Click Kit 488 Baseclick增殖体内成像检测试剂盒(488) 100
BCK555-IV-IM-L In Vivo EdU Click Kit 555 Baseclick增殖体内成像检测试剂盒(555) 100
BCK555-IV-IM-M In Vivo EdU Click Kit 555 Baseclick增殖体内成像检测试剂盒(555) 100
BCK555-IV-IM-S In Vivo EdU Click Kit 555 Baseclick增殖体内成像检测试剂盒(555) 100
BCK594-IV-IM-L In Vivo EdU Click Kit 594 Baseclick增殖体内成像检测试剂盒(594) 100
BCK594-IV-IM-M In Vivo EdU Click Kit 594 Baseclick增殖体内成像检测试剂盒(594) 100
BCK594-IV-IM-S In Vivo EdU Click Kit 594 Baseclick增殖体内成像检测试剂盒(594) 100
BCK647-IV-IM-L In Vivo EdU Click Kit 647 Baseclick增殖体内成像检测试剂盒(647) 100
BCK647-IV-IM-M In Vivo EdU Click Kit 647 Baseclick增殖体内成像检测试剂盒(647) 100
BCK647-IV-IM-S In Vivo EdU Click Kit 647 Baseclick增殖体内成像检测试剂盒(647) 100
BCK488-IV-FC-L In Vivo EdU Flow Cytometry 50 Kit 488 Baseclick流式细胞术体内增殖检测试剂盒(488) 50
BCK488-IV-FC-M In Vivo EdU Flow Cytometry 50 Kit 488 Baseclick流式细胞术体内增殖检测试剂盒(488) 50
BCK488-IV-FC-S In Vivo EdU Flow Cytometry 50 Kit 488 Baseclick流式细胞术体内增殖检测试剂盒(488) 50
BCK555-IV-FC-L In Vivo EdU Flow Cytometry Kit 555 Baseclick流式细胞术体内增殖检测试剂盒(555) 50
BCK555-IV-FC-M In Vivo EdU Flow Cytometry Kit 555 Baseclick流式细胞术体内增殖检测试剂盒(555) 50
BCK555-IV-FC-S In Vivo EdU Flow Cytometry Kit 555 Baseclick流式细胞术体内增殖检测试剂盒(555) 50
BCK594-IV-FC-L In Vivo EdU Flow Cytometry Kit 594 Baseclick流式细胞术体内增殖检测试剂盒(594) 50
BCK594-IV-FC-M In Vivo EdU Flow Cytometry Kit 594 Baseclick流式细胞术体内增殖检测试剂盒(594) 50
BCK594-IV-FC-S In Vivo EdU Flow Cytometry Kit 594 Baseclick流式细胞术体内增殖检测试剂盒(594) 50
BCK647-IV-FC-L In Vivo EdU Flow Cytometry Kit 647 Baseclick流式细胞术体内增殖检测试剂盒(647) 50
BCK647-IV-FC-M In Vivo EdU Flow Cytometry Kit 647 Baseclick流式细胞术体内增殖检测试剂盒(647) 50
BCK647-IV-FC-S In Vivo EdU Flow Cytometry Kit 647 Baseclick流式细胞术体内增殖检测试剂盒(647) 50
BCK488-IV-HTS-L In Vivo EdU HTS 2x96 Kit 488 Baseclick增殖体内高通量检测试剂盒(488) 200
BCK488-IV-HTS-M In Vivo EdU HTS 2x96 Kit 488 Baseclick增殖体内高通量检测试剂盒(488) 200
BCK488-IV-HTS-S In Vivo EdU HTS 2x96 Kit 488 Baseclick增殖体内高通量检测试剂盒(488) 200
BCK555-IV-HTS-L In Vivo EdU HTS 2x96 Kit 555 Baseclick增殖体内高通量检测试剂盒(555) 200
BCK555-IV-HTS-M In Vivo EdU HTS 2x96 Kit 555 Baseclick增殖体内高通量检测试剂盒(555) 200
BCK555-IV-HTS-S In Vivo EdU HTS 2x96 Kit 555 Baseclick增殖体内高通量检测试剂盒(555) 200
BCK594-IV-HTS-L In Vivo EdU HTS 2x96 Kit 594 Baseclick增殖体内高通量检测试剂盒(594) 200
BCK594-IV-HTS-M In Vivo EdU HTS 2x96 Kit 594 Baseclick增殖体内高通量检测试剂盒(594) 200
BCK594-IV-HTS-S In Vivo EdU HTS 2x96 Kit 594 Baseclick增殖体内高通量检测试剂盒(594) 200
BCK647-IV-HTS-L In Vivo EdU HTS 2x96 Kit 647 Baseclick增殖体内高通量检测试剂盒(647) 200
BCK647-IV-HTS-M In Vivo EdU HTS 2x96 Kit 647 Baseclick增殖体内高通量检测试剂盒(647) 200
BCK647-IV-HTS-S In Vivo EdU HTS 2x96 Kit 647 Baseclick增殖体内高通量检测试剂盒(647) 200
11699695001 ATP Bioluminescence Assay Kit CLS II ATP生物发光检测试剂盒 CLS II 1600
11699709001 ATP Bioluminescence Assay Kit HS II ATP生物发光检测试剂盒 HS II 1000
12209136001 TeloTAGGG Telomere Length Assay 端粒DNA长度检测试剂盒 50
11854666910 TeloTAGGG Telomerase PCR ELISA 端粒酶PCR ELISA试剂盒 96
细胞增殖检测试剂
产品货号 产品名称 产品描述
B8434 Monoclonal Anti-BrdU antibody produced in mouse ~1.5 mg/mL, clone BU-33, purified immunoglobulin, buffered aqueous solution Anti-BrdU 抗体(细胞增殖检测)
B2531 Monoclonal Anti-BrdU antibody produced in mouse clone BU-33, ascites fluid, Immunohistology Grade Anti-BrdU 抗体(细胞增殖检测)
P8825 Monoclonal Anti-Proliferating Cell Nuclear Antigen antibody produced in mouse clone PC 10, ascites fluid 增殖细胞核抗原单抗(细胞增殖检测)
P6834 Monoclonal Anti-Proliferating Cell Protein Ki-67 antibody produced in mouse clone PP-67, ascites fluid 增殖细胞Ki-67单抗(细胞增殖检测)
GG1 Glycerol Gelatin aqueous slide mounting medium 水溶性封片胶
D2650 Dimethyl sulfoxide 二甲基亚砜Hybri-Max™,过滤灭菌,≥99.7%
B5002 5-Bromo-2′-deoxyuridine≥99% (HPLC) 5-溴-2′-脱氧尿苷 ≥99% (HPLC)
B9285 5-Bromo-2′-deoxyuridine BioUltra,≥99% 5-溴-2′-脱氧尿苷 BioUltra, ≥99%
P3563 Phosphate buffered saline 磷酸盐缓冲盐溶液pH 7.4,含TWEEN® 20
P7012 Pepsin from porcine gastric mucosa 胃蛋白酶,来源于猪胃粘膜冻干粉
T7409 Trypsin from porcine pancreas 胰蛋白酶,来源于猪胰腺