Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
癌症干细胞假说
 癌症干细胞假说
传统观点认为,癌症是环境或内部因素诱导正常细胞内的关键致癌基因和肿瘤抑制基因突变导致的一种疾病。当这些突变导致细胞向更原始、高度增殖状态转化,克隆扩张形成白血病或实体肿瘤时,就会出现癌症的临床表现1。然而,这种模式不能够完全解释许多肿瘤和转移瘤的长发育延迟,产生细胞去分化和细胞永生的起始诱导机制,或肿瘤本身内的细胞功能和表型多样性起源。过去十年中,越来越多的证据表明,恶性肿瘤起源于组织干细胞在突变作用下的转化,这些突变会导致控制干细胞生长和增殖的正常机制的调节异常1-9
成体干细胞填充每个器官内的离散区位。它们具有长生命期和多能性能,可以重新转化成该器官内的所有类型的细胞。干细胞的两个显著特性是:
  1. 在长期的区位环境中保持休眠的能力,
  2. 不对称细胞分裂能力,可分裂为一个干细胞(称作自我更新)和一个分化的子代。
支持CSC假说的例证
支持CSC假说的例证
此外,干细胞具有细胞凋亡耐受性,具有增强的端粒酶和DNA修复活性,并且具有膜结合的ABC转运蛋白,能够排除异物,从而使干细胞相对耐受化疗剂和其他外源性毒物的损伤作用9-11 。自我更新加上长休眠期,使接触损伤剂的干细胞能积聚可能随着时间推移导致恶性转化的突变3
 癌症干细胞鉴定
研究人员已经从大多数白血病2,4,13以及许多实体瘤(如脑胶质母细胞瘤和成神经管细胞瘤4,5,10,14,15 、乳腺3,16 、子宫颈、结肠直肠17,18 、胃肠道、肝细胞、肺、胰腺、前列腺和皮肤癌6,9,19,20中分离得到少量的干细胞样细胞。癌症干细胞与其对应的正常干细胞具有相同的细胞表面标记物(见表1),但却表现出不受控制的增殖,这可能是由于对负生长调节因子或对接触抑制和间隙连接细胞间通讯(GJIC)缺失的响应性降低所致。1,10
产品特点
AldeRed ALDH检测分析
癌症干细胞醛脱氢酶(ALDH)的表达量升高,被认为是所有CSCs的标志。ALDEFLUOR™检测试剂盒过去一直用于测量ALDH的表达水平,并广泛用于研究罕见的CSC群体。ALDEFLUOR检测试剂盒的发射波长位于电磁光谱的绿色区域(512nm)。因此,其中的试剂不能同时用于表达绿色荧光蛋白(发射绿色荧光光谱)的细胞或小鼠。AldeRed是用于标记活ALDH阳性细胞的ALDH红移荧光底物,从而释放出绿色发射通道以帮助同步完成其他检测。AldeRed克服了ALDEFLUOR检测试剂盒目前的局限。
阅读《Nature》中关于AldeRed的文章
AldeRed的特性和优点:
  • 红移检测释放出其他实验所需的绿色通道
  • 活体干细胞鉴定技术,能够对罕见的细胞群体进行流式分选
  • 快速酶反应检测方案
Aldered detection
Deab
AldeRed 588-A是一种无毒的ALDH荧光底物,能够自由进入完整活细胞但其被ALDH转化成酸产物后仍将滞留在细胞内。
 癌症干细胞信号转导通路
干细胞巢被充满时,正常干细胞的自我更新将终止。与正常干细胞不同,癌症干细胞和干细胞源癌祖细胞的不受控自我更新会超过其区位并渗入周围组织13。Hedgehog,Notch,Wnt,和PTEN是控制正常和癌症干细胞自我更新、增殖和存活的部分途径。在大多数侵袭性癌症中,都可观察到导致这些途径不同程度激活的突变。
Hedgehog 和 Bmi1通路
Hedgehog(Hh)通路能够调节成体干细胞的休眠和自我更新。研究人员已经在哺乳动物中鉴定了三种Hh配体,即Sonic hedgehog(Shh)、Desert hedgehog(Dhh)和Indian hedgehog(Ihh),其中对Shh的研究最多。在没有配体的情况下,Hh受体PTCH1通过催化抑制跨膜蛋白Smoothened(SMO),从而抑制信号转导。配体与PTCH1可使受体失活,并活化SMO,继而诱导Gli转录因子;Gli1和Gli2是Hh信号通路的阳性信号介导子,而Gli3是负调节子3,20
Bmi1的诱导能够促进许多Hh活化的效应。Bmi1 是一个的多梳基因,通过染色质重塑抑制转录,并能够下调Ink-4A / ADP核糖基化因子(ARF)复合物(如p16 Ink4A 和p19 ARF)的基因表达——这些基因是细胞周期的负调节物并且参与干细胞的休眠和分化3,4。这使得Hh通路元件PTCH1、Gli1和Gli2表达上调19,20, 以及Gli1和Gli2诱导的生长促进基因cyclin D1、Myc和Snail表达来实现干细胞分化和自我更新。
Notch-γ-分泌酶通路
Notch信号转导能够促进正常神经干细胞的存活和增殖,抑制它们的分化。许多癌细胞系(包括T细胞白血病,以及脑、乳腺、卵巢、宫颈、结肠直肠、胰腺、唾液腺和肺部癌症3,6,9,15 )的干细胞样细胞中,Notch信号转导通路被高度激活。其中,成神经管细胞瘤和T细胞淋巴细胞白血病是最具Notch依赖性的恶性肿瘤15
Notch通路激活包括通过γ-分泌酶对Notch配体/受体复合物进行蛋白水解切割,从而释放Notch胞内结构域片段(NICD),移位到细胞核并上调Myc、Hes1和其他基因的表达。当用NICD2转染DAOY成神经管细胞瘤细胞系使Notch通路持续激活时,转染后的细胞比非转染的DAOY细胞产生更多的异种移植肿瘤,培养物中CD133 +和侧群干细胞样细胞也增加了。相反,抑制γ-分泌酶可将侧群细胞减少至总细胞计数的0.01%,并且抑制90%的细胞形成软琼脂集落及在免疫受损的小鼠中形成肿瘤异种移植物的能力。NICD2转染可保护细胞免受γ-分泌酶抑制的影响15。因此,在一些肿瘤类型中,抑制Notch信号转导可以消除肿瘤起始所需的细胞群。
Wnt-β-连环蛋白途径
Wnt包含结合Frizzled(Fz)受体家族的19种胞外糖蛋白家族,其可激活抑制β-连环蛋白的蛋白水解降解途径,这使得β-连环蛋白在细胞质中积累并向细胞核转运。在细胞核中,β-连环蛋白通过Tcf / Lef转录因子促进基因转录。在Wnt未激活的情况下,轴蛋白(Axin)和腺瘤性息肉病大肠杆菌蛋白(APC)与糖原合酶激酶3(Gsk3)形成复合物,从而促进细胞质β-连环蛋白磷酸化,继而靶向β-连环蛋白以进行蛋白水解降解。
Wnt通路的激活需要Fz与其共同受体(LDL受体样蛋白5/6(Lrp5 / 6))的结合。Wnt / Fz / Lrp5 / 6复合物的形成激活磷蛋白Dishevelled(Dvl)(Fz受体的组成成分),继而招募Axin / Gsk3,从而解离Axin / APC / Gsk3复合物。Lrp5 / 6被Gsk3和酪蛋白激酶1γ(Ck1γ)依次磷酸化。 磷酸化的Lrp5 / 6结合Axin / Gsk3形成复合体,防止β-连环蛋白的磷酸化从而稳定β-连环蛋白。
癌症干细胞中关键发育信号转导通路的激活
图1. 癌症干细胞中关键发育信号转导通路的激活。Shh、Wnt和Notch信号转导途径与维持癌症干细胞的致瘤潜力有关。通过与Bmi1信号转导介体和其他自我更新基因(如Nanog和Sox-2)的相互作用,这些新的调节机制可以促进或抑制自我更新,从而提供癌症治疗干预的方法。
材料
 参考文献
  1. Trosko, J.E., et al., Ignored hallmarks of carcinogenesis: stem cells and cell-cell communication. Ann. NY Acad. Sci., 1028, 192-201 (2004).
  2. Dick, J.E., Acute myeloid leukemia stem cells. Ann. NY Acad. Sci., 1044, 1-5 (2005).
  3. Liu, S., et al., Hedgehog signaling and Bmi-1 regulate self-renewal of normal and malignant human mammary stem cells. Cancer Res., 66, 6063-6071 (2006).
  4. Massard, C., et al., Tumor stem cell-targeted treatment: elimination or differentiation. Ann. Oncol., 17, 1620-1624 (2006).
  5. Meletis, K., et al., p53 suppresses the self-renewal of adult neural stem cells. Development, 133, 363-369 (2006).
  6. Mimeault, M., and Batra, S.K., Concise review: recent advances on the significance of stem cells in tissue regeneration and cancer therapies. Stem Cells, 24, 2319-2345 (2006).
  7. Shen, R., et al., Precancerous stem cells can serve as tumor vasculogenic progenitors. PLOS One, 3, e1652 (2008).
  8. Vermeulen, L., et al., Cancer stem cells - old concepts, new insights. Cell. Death Differ., e-pub Feb 15. 2008.
  9. Wicha, M.S., et al., Cancer stem cells: an old idea - a paradigm shift. Cancer Res., 66, 1883-1890 (2006).
  10. Sakariassen, P.O., et al., Cancer stem cells as mediators of treatment resistance in brain tumors: status and controversies. Neoplasia, 9, 882-892 (2007).
  11. Styczynski, J., and Drewa, T., Leukemic stem cells: from metabolic pathways and signaling to a new concept of drug resistance targeting. Acta Biochim. Pol., 54, 717-726 (2007).
  12. Bailey, P., and Cushing, H., A Classification of the Tumors of the Glioma Group on a Histogenetic Basis with a Correlated Study of Prognosis. Philadelphia: J.P. Lippincott, (1926).
  13. Strewler, G.J., The stem cell niche and bone metastasis. BoneKEy-Osteovision, 3, 19-29 (2006).
  14. Singh, S.K., et al. Identification of a cancer stem cell in human brain tumors. Cancer Res., 63, 5821-5828 (2003).
  15. Fan, X., et al., Notch pathway inhibition depletes stem-like cells and blocks engraftment in embryonal brain tumors. Cancer Res., 66, 7445-7452 (2006).
  16. Benhaj, K., et al., Redundant expression of canonical Wnt ligands in human breast cancer cell lines. Oncol. Rep., 15, 701-707 (2006).
  17. Fodde, R., and Brabletz, T., Wnt/β-catenin signaling in cancer stemness and malignant behavior. Curr. Opin. Cell Biol., 19, 150-158 (2007).
  18. Zhou, J., et al., Activation of the PTEN/mTOR/STAT3 pathway in breast cancer stemlike cells is required for viability and maintenance. Proc. Natl. Acad. Sci. USA, 104, 16158-16163 (2007).
  19. Mimeault, M., and Batra, S.K., Interplay of distinct growth factors during epithelialmesenchymal transition of cancer progenitor cells and molecular targeting as novel cancer therapies. Ann. Oncol., 18, 1605-1619 (2007).
  20. Peacock, C.D., et al., Hedgehog signaling maintains a tumor stem cell compartment in multiple myeloma. Proc. Natl. Acad. Sci. USA, 104, 4048-4053 (2007).
  21. Bao, S., et al., Stem cell-like glioma cells promote tumor angiogenesis through vascular endothelial growth factor. Cancer Res., 66, 7843-7848 (2006).
  22. O’Neil, J., et al., FBW7 mutations in leukemic cells mediate NOTCH pathway activation and resistance to γ-secretase inhibitors. J. Exp. Med., 204, 1813-1824 (2007).
  23. He, X., and Axelrod, J.D., A WNTer wonderland in Snowbird. Development, 133, 2597-2603 (2006).
  24. Zeng, X., et al., Initiation of Wnt signaling: control of Wnt coreceptor Lrp6 phosphorylation/activation via frizzled, disheveled and axin functions. Development, 135, 367-375 (2008).
  25. Groszer, M., et al., PTEN negatively regulates neural stem cell self-renewal by modulating G0-G1 cell cycle entry. Proc. Natl. Acad. Sci. USA, 103, 111-116 (2006).
  26. Hede, K., PTEN takes center stage in cancer stem cell research, works as tumor suppressor. J. Nat. Cancer Inst., 98, 808-809 (2006).
  27. A red-shifted fluorescent substrate for aldehyde dehydrogenase. Martin G. Pomper et al. Nature Communications 5, Article number: 3662 23 April 2014
back to top 
Explore Our Cell Culture Offering Technology