Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Supelco推出Ascentis® Express 5μm颗粒液相色谱柱
Sigma-Aldrich宣布其Supelco品牌发布Ascentis® Express C18,5μm颗粒的液相色谱柱——以加快分析速度,提升高效液相色谱柱分离性能。与传统5μm颗粒的色谱柱相比,新型色谱柱是以熔融核颗粒技术Fused-Core®为基础,其分析速度更快,柱效更高。Ascentis® Express 5μm系列色谱柱提供给科学家们及广大分析工作者在传统HPLC仪器上提高性能的第一选择,并消除了在使用小粒径颗粒色谱柱时的担忧。
新型5μm颗粒熔融核色谱柱的速度和分离能力与3μm颗粒色谱柱相当,而与亚2μm颗粒色谱柱相比,则更稳固耐用寿命更长。新系列的色谱柱操作的背压与传统5μm颗粒色谱柱相当,确保了不会超过传统HPLC压力限制。Ascentis® Express系列产品在ΜHPLC及HPLC上均能使用。
“Supelco新上市的Ascentis® Express产品线使得科学工作者们能够提升现有HPLC系统的性能,非常轻松地在现场或合同研究组织(CRO)实现方法转移,而不用担心兼容性问题。”Supelco公司的HPLC/GC市场经理Wayne Way博士说,“这些色谱柱在LC/MS的生物分析领域用处极大,因为它们能实现高流速和高通量,以满足生物分析方法的需求。此外,大颗粒也令HPLC色谱柱足够坚固。”Way博士总结道。
现有推出的Ascentis® Express 5μm液相色谱柱,固定相有C18和F5两种。将现有方法转换至新型5μm色谱柱,非常简单。传统HPLC使用者能非常便利地用其替换已有色谱柱,无需更变色谱柱尺寸,流速或样品制备。
此外,近日Sigma-Aldrich还推出了HPLC转换计算器器,目前该应用已经上传至Sigma-Aldrich网站,点击这里访问:http://www.sigmaaldrich.com/hplccalculator。该应用使得切换色谱柱和方法,变得非常容易。输入相应色谱柱变量(柱长、柱内径、粒径)和现有方法(流速、进样量、压力、运行时间、平衡时间),能将等度或梯度的方法从一根色谱柱转移至另一根色谱柱。该转换器也能提供从分析柱放大到制备柱的色谱分析方法转换,同时可计算节省的时间和溶剂。该应用也可在 iPad®, iPhone® 及 Android™ 等移动设备上使用。
技术资料下载
Ascentis® Express 技术资料
点击这里下载
HPLC 方法转移计算器
HPLC 方法转移计算器工具
点击这里访问网页版
Google Play marketplace
iTunes marketplace