Hazard and Precautionary Statements
HAZARD STATEMENTS
There are 72 individual and 17 combined Hazard statements - these are assigned a unique alphanumerical code which consists of one letter and three numbers as follows:
  1. the letter "H" (for "hazard statement");
  2. a number designating the type of hazard as follows:
    - "2" for physical hazards
    - "3" for health hazards
    - "4" for environmental hazards
  3. two numbers corresponding to the sequential numbering of hazards arising from the intrinsic properties of the substance or mixture, such as explosive properties (codes from 200 to 210), flammability (codes from 220 to 230), etc.
>>> Hazard statement overview
PRECAUTIONARY STATEMENTS
There are 116 individual and 33 combined Precautionary statements – these are assigned a unique alphanumerical code which consists of one letter and three numbers as follows:
  1. the letter "P" (for "precautionary statement");
  2. one number designating the type of precautionary statement as follows: - "1" for general precautionary statements
    - "2" for prevention precautionary statements
    - "3" for response precautionary statements
    - "4" for storage precautionary statements
    - "5" for disposal precautionary statements
  3. two numbers (corresponding to the sequential numbering of precautionary statements)
>>> Precautionary statement overview
PICTOGRAMS AND HAZARD CODES
Compared to the current EU system the most noticeable change are the pictograms (formerly: hazard symbols). While the most of the GHS pictograms have an equivalent in the old system, the pictograms GHS 04, GHS 07 and GHS 08 are completely new.
The GHS System is built on 16 physical, 10 health and 3 environmental hazard classes and comprises the following communication elements:
9 Pictograms
Description
Pictogram
Hazard class and hazard category:
Exploding Bomb
GHS01
Unstable explosives
Explosives of Divisions 1.1, 1.2, 1.3, 1.4
Self reactive substances and mixtures, Types A,B
Organic peroxides, Types A,B
Flame
GHS02
Flammable gases, category 1
Flammable aerosols, categories 1,2
Flammable liquids, categories 1,2,3
Flammable solids, categories 1,2
Self-reactive substances and mixtures, Types B,C,D,E,F
Pyrophoric liquids, category 1
Pyrophoric solids, category 1
Self-heating substances and mixtures, categories 1,2
Substances and mixtures, which in contact with water,
emit flammable gases, categories 1,2,3
Organic peroxides, Types B,C,D,E,F
Flame Over Circle
GHS03
Oxidizing gases, category 1
Oxidizing liquids, categories 1,2,3
Gas Cylinder
GHS04
Gases under pressure:
- Compressed gases
- Liquefied gases
- Refrigerated liquefied gases 
- Dissolved gases
Corrosion
GHS05
Corrosive to metals, category 1
Skin corrosion, categories 1A,1B,1C
Serious eye damage, category 1
Skull and Crossbones
GHS06
Acute toxicity (oral, dermal, inhalation), categories 1,2,3
Exclamation Mark
GHS07
Acute toxicity (oral, dermal, inhalation), category 4
Skin irritation, category 2
Eye irritation, category 2
Skin sensitisation, category 1
Specific Target Organ Toxicity – Single exposure, category 3
Health Hazard
GHS08
Respiratory sensitization, category 1
Germ cell mutagenicity, categories 1A,1B,2
Carcinogenicity, categories 1A,1B,2
Reproductive toxicity, categories 1A,1B,2
Specific Target Organ Toxicity – Single exposure, categories 1,2
Specific Target Organ Toxicity – Repeated exposure, categories 1,2
Aspiration Hazard, category 1
Environment
GHS09
Hazardous to the aquatic environment
- Acute hazard, category1
- Chronic hazard, categories 1,2