Glassware Catalog
Bottles - Sigma-Aldrich Glassware Catalog
On this page:
Dropping bottles Reagent bottlesAldrich graduated cylinder • Storage bottlesFiltration bottlesAldrich 75mm O-ring bottles
Dropping bottles with graduated dropper
Clear glass dropping bottle with ground glass joint. The 1mL dropper has 0.10mL graduations.
Cap (mL)
S.T. Joint
Catalog Number
30
14/20
60
14/20
125
19/22
Back to Top
Reagent bottles
Reagent bottles come with a ground glass hex head stopper.
Cap (mL)
S.T. Joint
Catalog Number
Clear
30
14/20
60
14/20
125
19/22
Amber
30
14/20
60
14/20
125
19/22
Back to Top
Aldrich graduated cylinder with S.T. joint, Class A
Single metric scale. Useful as a distillation receiver and for manipulation of air-
sensitive materials with the use of a septum inlet adapter.
Cap
(mL)
S.T.
Joint
Grad.
(mL)
Tol.
(+/- mL)
Catalog Number
5
14/20
0.1
0.05
10
14/20
0.2
0.10
25
14/20
0.5
0.25
50
14/20
1.0
0.50
100
24/40
1.0
0.50
250
24/40
2.0
1.00
500
24/40
5.0
2.50
1,000
24/40
10.0
5.00
Back to Top
Storage bottles without joints
Clear glass, one piece design with stopcock equipped septum-inlet.
Cap. (mL)
Stopcock bore (mm)
PTFE stopcock Catalog Number
Glass stopcock
Catalog Number
125
2
250
2
500
2
1,000
4
-
2,000
4
-
Back to Top
Storage bottles with threaded joints
Cap. (mL)
PTFE stopcock
Catalog Number
Glass stopcock
Catalog Number
125
250
500
1,000
2,000
Replacement parts
Cap. (mL)
Storage bottle
Catalog Number
125
250
500
1,000
2,000
Description
Septum adapter
Catalog Number
Back to Top
Filtration bottles
Available with standard hose barb or threaded 6mm glass stopcock. Use a number 12 stopper and 3/8 in. i.d. tubing with all versions.
Cap (L)
Diam. x H (mm)
Hose barb
Catalog Number
Stopcock
Catalog Number
Graduated bottles - double scale (L and gal) in 500mL increments
9.5
187 x 476
13.25
238 x 438
19
292 x 508
Ungraduated bottles
9.5
187 x 476
13.25
238 x 438
19
292 x 508
45.5
406 x 584
Polycoating is available upon request, please contact the Aldrich Glass Shop at aldglass@sial.com for more details.
Back to Top
Aldrich 75mm O-ring bottles
Ideal for controlled vacuum filtration or transfer situations without the need for a stopper
Cap. (L)
Diam. x H (mm)
Catalog Number
Graduated bottles - double scale (L and gal) in 500mL increments
9.5
187 x 476
13.25
238 x 438
19
292 x 508
Ungraduated bottles
9.5
187 x 476
13.25
238 x 438
19
292 x 508
45.5
406 x 584
Aldrich 75mm O-ring tops
Each top comes with a 4mm PTFE stopcock. Use the stainless steel clamp listed below.
Torion inlet style - Use 1/2in. hard-wall transfer tubing (e.g. PP or PTFE) for the airtight transfer of air-sensitive materials.
Standard Taper - The S.T. 24/40 and S.T. 29/32 joint can accommodate a transfer adapter or a filter funnel.
Description
Catalog Number
Torion inlet top
S.T. 24/40 top
S.T. 29/32 top
Stainless Steel 75mm Clamp
Replacement Viton O-Ring
Back to top