Glassware Catalog
Filtration Flasks - Sigma-Aldrich Glassware Catalog
Filtration  •  Flat-Bottom  •  Round-Bottom  •  Multi-Neck  •  Schlenk  •  Septum-Inlet  •  Accessories
On this page:
Aldrich filter flasksPyrex heavy-wall filtering flasks
Filtration bottlesAldrich 75mm o-ring bottles
Aldrich filter flasks
A. 1,000mL reservoir flasks with hose barb.
S.T. Joint
Catalog Number
24/40
29/32
B. 1,000mL, with 3-way glass stopcock. Use with NMR tube cleaners, Gooch crucibles, and Buchner funnels. With threaded S.T. 24/40 joint.
Z17,481-5
Back to top
Pyrex heavy-wall filtering flasks
Blown in special molds to give maximum strength. Graduated to show approximate capacity.
Cap.
(mL)
Stopper
Number
Grad.
range (mL)
Catalog
Number
Without hose connection (Corning 5320)
250
6
75 - 250
500
7
150 - 500
1,000
8
300 - 1,000
2,000
9
600 - 2,000
4,000
12
1,500 - 4,000
10mm o.d. hose-connection for sizes up to 1,000mL and 12.5mm o.d. for 2,000 and 4,000mL sizes (Corning 5340)
250
6
75 - 250
500
7
150 - 500
1,000
8
300 - 1,000
2,000
9
600 - 2,000
4,000
12
1,500 - 4,000
Back to top
Filtration bottles
Available with standard hose barb or threaded 6mm glass stopcock. Use a number 12 stopper and 3/8in. i.d. tubing with all versions.
Cap. (L)
Diam. x H (mm)
Hose Barb
Catalog Number
Stopcock
Catalog Number
Graduated bottles - double scale (L and gal) in 500mL increments
9.5
187 x 476
13.25
238 x 438
19
292 x 508
Ungraduated bottles
9.5
187 x 476
13.25
238 x 438
19
292 x 508
45.5
406 x 584
Polycoating is available upon request, please contact the Aldrich Glass Shop at aldglass@sial.com for more details.
Back to top
Aldrich 75mm o-ring bottles
Ideal for controlled vacuum filtration or transfer situations without the need for a stopper
Cap. (L)
Diam. x H (mm)
Catalog Number
Graduated bottles - double scale (L and gal) in 500mL increments
9.5
187 x 476
13.25
238 x 438
19
292 x 508
Ungraduated bottles
9.5
187 x 476
13.25
238 x 438
19
292 x 508
45.5
406 x 584
Aldrich 75mm o-ring tops
Each top comes with a 4mm PTFE stopcock. Use the stainless steel clamp listed below.
Torion inlet style - Use 1/2in. hard-wall transfer tubing (e.g. PP or PTFE) for the airtight transfer of air-sensitive materials.
Standard Taper - The S.T. 24/40 and S.T. 29/32 joint can accommodate a transfer adapter or a filter funnel.
Description
Catalog Number
Torion inlet top Z51,712-7
S.T. 24/40 top Z51,713-5
S.T. 29/32 top Z51,739-9
Stainless Steel 75mm Clamp
Replacement Viton O-Ring Z54,300-4
Back to top