Figure 1. Two genes are expressed from a single bicistronic mRNA.
For the construct illustrated (pBICEP-CMV-1), the target gene is expressed in the typical cap-dependent fashion while the neoR gene is translated in a cap-independent manner via the EMCV IRES element.
Close Window