KiCqStart® qPCR Product Line Overview
The KiCqStart line of qPCR products provide high-quality, easy-to-use tools to measure gene expression. KiCqStart qPCR ReadyMixes are 2X concentrated, ready-to-use reaction cocktails containing all components, except primers and template, needed for real-time quantitative PCR (qPCR). This unique combination of proprietary buffer, stabilizers, and hot-start Taq DNA polymerase delivers maximum PCR efficiency, sensitivity, specificity using fast or conventional cycling protocols. Compatible with any thermal cycler, KiCqStart Primers are ready-to-order, pre-designed primer pairs for quantifying gene expression by SYBR Green I RT-qPCR (Reverse Transcription Quantitative Real-Time PCR). To comply with MIQE, they have been developed using sophisticated bioinformatics tools and validated in silico to avoid off-target amplification.
  • ReadyMixes enable cycling in fast or conventional qPCR mode
  • Preformatted ReadyMixes include instrument-specific reference dye
  • MIQE-compliant pre-designed SYBR Green primers and arrays