Rotor Guide - Amicon® Ultra-2 and 4
  • Use any centrifuge and rotor suitable for 15ml tubes.
  • Max g-Force 4,000 g in swinging bucket rotor or 7,500 g in fixed angle rotor for the concentration spin and 1,000 g for the reverse spin with the Amicon® Ultra 2 mL.
Learn more about Amicon® Ultra-2
Learn more about Amicon® Ultra-4
 Select Manufacturer to See Recommendations
Note: Centrifuges and rotors listed are not available from us. Please confirm product availabilty with the manufacturer.
Beckman
Centrifuge Model Rotor Adapter Required
Allegra X12 SX4750 359472
Allegra X12 SX4750A 359472
back to top
Eppendorf
Centrifuge Model Rotor Adapter Required
5702/R/RH A-4-38 22639188
5702/R/RH F-35-30-17 22639447
5702/R/RH A-8-17  
5804 / 5804R F-34-6-38 22637274
5804 / 5804R A-44-4 22637606
5810 / 5810R F-34-6-38 22637274
5810 / 5810R A-4-62 22638289
back to top
Hermle
Centrifuge Model Rotor Adapter Required
C0200-A C0200-96  
back to top
Hettich
Centrifuge Model Rotor Adapter Required
EBA20 Included  
EBA21 1416  
EBA21 1116 1631
ROTANTA460/460R 5624 5625/4739
ROTANTA460/460R 5624 5620/4839
ROTANTA460/460R 5615 1466
ROTINA35/35R 1724 1430/1347
ROTINA35/35R 1724 1425/1736
ROTINA35/35R 1794 1790/5133
ROTINA35/35R 1717 1462
ROTINA35/35R 1792 1466
ROTINA35/35R 1720 1466
ROTOFIX32 1624 1381/1347
ROTOFIX32 1619 1462
ROTOFIX32 1617 1462
ROTOFIX32 1613  
ROTOFIX32 1620A 1451
UNIVERSAL32/32R 1624 1381/1347
UNIVERSAL32/32R 1619 1462
UNIVERSAL32/32R 1617 1462
UNIVERSAL32/32R 1620A 1451
UNIVERSAL32/32R 1613  
back to top
Kendro
Centrifuge Model Rotor Adapter Required
Biofuge Primo/Primo R 75007588 75003095
Biofuge Primo/Primo R 75007555 75007557
Biofuge Stratos 75003057 75003095
Biofuge Stratos 75008172 75008187
Legend Mach 1.6, Multifuge 1/1R 75002006 75003095
Legend Mach 1.6, Multifuge 1/1R 75002002 75007621
Legend Mach 1.6, Multifuge 1/1R 75002001 75002020
Legend T/RT, Multifuge 3/3R 75003057 75003095
Legend T/RT, Multifuge 3/3R 75006441 75008137
Legend T/RT, Multifuge 3/3R 75006446 75006456
back to top
Thermo/Forma
Centrifuge Model Rotor Adapter Required
5528 400ML GP 5852151, 5852152, 5858012, 5852368, 5801106
5529 400ML GP  
5530 400ML GP 5800236 5802092
5530/5532 1L GP/GPR 5800224, 5852242 5807230
5530/5532 1L GP/GPR 5852151, 5852152, 5858012, 5858162 5801106
5530/5532 1L GP/GPR   5801106
5530/5532 1L GP/GPR 5852151, 5852152,5859581, 5858012, 5858162 5801106, (5850968 - for use with rotors listed to left)
5530/5532 1L GP/GPR 5800243 5806561
5677/5678 1L MP/MPR 5852151, 5852152, 58012, 58162 5801106
5540/5542 1L MP/MPR 5806555 5806561
5694/5696 1L HP/HPR 5807685 5806651/5806715
5694/5696 1L HP/HPR 5808685 5806651/5806715
5694/5696 1L HP/HPR 5808850 5806715
5694/5696 1L HP/HPR 5808947 5806561
5660/5662 3L 2500 GP/GPR 5800825 5801106/571
5660/5662 3L 2500 GP/GPR 5806554 5807012 (sealed), 5805712 (unsealed)
5681/5682 3L 4500 GP/GPR (bench), 5687/5688 3L 4500 GP/GPR (floor) 5800218 5805712
5681/5682 3L 4500 GP/GPR (bench), 5687/5688 3L 4500 GP/GPR (floor) 5800825 5801106/571
back to top
Thermo/IEC
Centrifuge Model Rotor Adapter Required
Clinical 52151, 58012 1106
Centra® CL2 52151, 52152, 58012 1106
Centra CL2 236 2092S
HN-SII, Centra® HN, Centra® CL4 958; 52152; 59582 326/320/1106
HN-SII, Centra HN, Centra CL4 245, 268 7230
HN-SII, Centra HN, Centra CL4 58012, 58162 1106
HN-SII, Centra HN, Centra CL4 215 325/320/1106
HN-SII, Centra HN, Centra CL4 215 326/320/1106
Centra® CL3/CL3R 224, 52242 7230
Centra CL3/CL3R 52151, 52152, 58012, 58162 1106
Centra CL3/CL3R 52151, 52152, 59581,58012, 58162 1106, (50968 - for use with rotors listed to left)
Centra CL3/CL3R 243 6561E
Centra® MP4/MP4R 52151, 52152, 58012, 58162 1106
Centra® CL5/CL5R 6555C 6561E
Multi/ Multi RF 7685C 6651E/6715
Multi/ Multi RF 8685 6651E/6715
Multi/ Multi RF 8850 6715
Multi/ Multi RF 8947 6561E
Centra® 7/7R, Centra® 8/8R 210, 216 7012E (sealed), 5712 (unsealed)
Centra 7/7R, Centra 8/8R 825S 1106/571
Centra 7/7R, Centra 8/8R 210, 216 7012E (sealed), 5712 (unsealed)
Centra® GP6/GP6R 825S 1106/571
Centra GP6/GP6R 6554C 7012E (sealed), 5712 (unsealed)
Centra® GP8/GP8R (Bench),GP8K/GP8RK (Kneewell), GP8F/GP8RF (Floor) 216, 228 5712
Centra GP8/GP8R (Bench),GP8K/GP8RK (Kneewell), GP8F/GP8RF (Floor) 218A 5712
Centra GP8/GP8R (Bench),GP8K/GP8RK (Kneewell), GP8F/GP8RF (Floor) 825S 1106/571
Model K, EXD 219A 256, 949 5712
Model K, EXD 284, 259 384S/7230
Model K, EXD 921, 269, 253 325/305/1106
Model K, EXD 921, 269, 253 325/320/1106
Model K, EXD 921, 269, 253 326/305/1106
Model K, EXD 921, 269, 253 326/320/1106
PR-7000M 219A, 256, 949, 966, 981 5712
PR-7000M 284, 259 384S/7230
PR-7000M 269, 253 325/305/1106
PR-7000M 269, 253 325/320/1106
PR-7000M 269, 253 326/305/1106
PR-7000M 269, 253 326/320/1106
B-22M 877, 963 7230
back to top