Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Purification and Detection
Enzyme Inhibitor Cocktail
 Protease Inhibitor Cocktails
Product Number P2714 S8820 P1860 P8465 P8340 P8849 S8830 P8215 P9599 I3911 I3786 MSSAFE
  General
Use
General
Use
Tissue
Culture
Bacterial
Cells
Mammalian
Tissues
Histidine-
Tagged
Proteins
Histidine-
Tagged
Proteins
Fungal/
Yeast
Cells
Plant
Cells
General
Use
(Animal
Component
Free/ACF)
Mammalian
Cells and
Tissues
(Animal
Component
Free/ACF)
Mammalian
Cells and
Tissues
  Lyophilized
powder -
Water
Soluble
Tablet -
Water
Soluble
Solution
in
DMSO
Lyophilized
powder -
Vial of
DMSO
Solution
in DMSO
Solution
in DMSO
Tablet -
Water
Soluble
Solution
in DMSO
Solution
in DMSO
Lyophilized
powder
(water
soluble)
Solution
in DMSO
Lyophilized
powder
(water
soluble)
AEBSF Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes  
Aprotinin Yes Yes Yes   Yes        
Bestatin Yes Yes Yes Yes Yes Yes Yes   Yes
EDTA Yes Yes   Yes              
E-64 Yes Yes Yes Yes Yes Yes Yes Yes Yes  
Leupeptin Yes Yes Yes   Yes       Yes
Pepstatin A     Yes Yes Yes Yes Yes Yes Yes  
Phosphoramidon           Yes Yes        
1,10-Phenanthroline               Yes Yes      
Elastinal                      
Nafamostat mesylate                      
Antipain                      
 Mix Your Own Cocktails
Create your own broad-spectrum protease inhibitor cocktails or screen your extracts for proteolytic activity. Panel includes inhibitors for serine, cysteine, calpains and metalloproteinases.
Protease Inhibitor Panel (INHIB1)
AEBSF 25 mg
Epsilon-AminoCaproic Acid 25 g
Antipain 5 mg
Aprotinin 5 mg
Benzamidine HCl 5 G
Bestatin 5 mg
Chymostatin 5 mg
E-64 5 mg
EDTA 50 g
N-Ethylmaleimide 5 g
Leupeptin 5 mg
Pepstatin 5 mg
Phosphoramidon 5 mg
Trypsin Inhibitor 100 mg
Priced at a fraction of the cost when purchased separately!
Includes detailed instructions for formulating your own cocktails.
 Phosphatase Inhibitor Cocktails
Product Number P5726 P0044 MSSAFE
  Tyrosine Protein Phosphatases and Acid and Alkaline Phosphatases Ser/Thr Protein Phosphatases and Alkaline Phosphatase L-Isozymes Tyrosine Protein Phosphatases, Ser/Thr Protein Phosphatases, Acid and Alkaline Phosphatases
(--)-p-Bromotetramisole   Yes
Calyculin A   Yes  
Cantharidin    
Imidazole Yes    
Okadaic acid    
Sodium fluoride    
Sodium molybdate Yes    
Sodium orthovanadate Yes  
Sodium tartrate Yes    
back to top
back to Enzyme Explorer