Cesium
 Cesium
Metal
Salts
Solution Deposition Precursors
Atomic Number: 55
Electronic Configuration: [Xe]6s1
Atomic Weight: 132.9055
Did you know? Cesium is the most electropositive of all naturally occurring elements. It will burst in to flames when exposed to air and explode when touched by water.