Thorium
 Thorium
Salts
Atomic Number: 90
Electronic Configuration: [Rn]6d27s2
Atomic Weight: 232.0381
Did you know? Thorium is an alloying agent used in GTAW (Gas Tungsten Arc Welding) to increase the melting temperature of tungsten electrodes and improve arc stability.